Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30668680-30669180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
UniGene
Hs.247186
Gene Chromosome Location
Chr.16: 30658142-30670810 on Build GRCh38
Transcripts
NM_001105079.2 XM_011545916.2 XM_011545917.2 XM_011545919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4809105 c.2276G>A p.R759Q Chr.16: 30668889-30668889 on Build GRCh38
5811878 c.2418G>T p.K806N Chr.16: 30669120-30669120 on Build GRCh38
4789385 c.2193A>T p.P731P Chr.16: 30668806-30668806 on Build GRCh38
5766138 c.494G>A p.R165Q Chr.16: 30668931-30668931 on Build GRCh38
5160639 c.2236C>T p.R746C Chr.16: 30668849-30668849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371959874 Chr.16: 30668941-30668941 on Build GRCh38
rs750561710 Chr.16: 30668788-30668788 on Build GRCh38
rs369543989 Chr.16: 30669014-30669014 on Build GRCh38
rs750958105 Chr.16: 30668843-30668843 on Build GRCh38
rs932134661 Chr.16: 30669112-30669112 on Build GRCh38
rs780832620 Chr.16: 30669098-30669098 on Build GRCh38
rs773293582 Chr.16: 30668813-30668813 on Build GRCh38
rs780671515 Chr.16: 30668858-30668858 on Build GRCh38
rs1026671866 Chr.16: 30668974-30668974 on Build GRCh38
rs139099586 Chr.16: 30669082-30669082 on Build GRCh38
rs374541307 Chr.16: 30668739-30668739 on Build GRCh38
rs1021383996 Chr.16: 30669046-30669046 on Build GRCh38
rs774348487 Chr.16: 30669133-30669135 on Build GRCh38
rs771816183 Chr.16: 30668929-30668929 on Build GRCh38
rs779469767 Chr.16: 30669086-30669086 on Build GRCh38
rs751560671 Chr.16: 30668857-30668857 on Build GRCh38
rs772919466 Chr.16: 30668749-30668749 on Build GRCh38
rs777763974 Chr.16: 30668957-30668957 on Build GRCh38
rs541586289 Chr.16: 30668903-30668903 on Build GRCh38
rs767518644 Chr.16: 30669058-30669058 on Build GRCh38
rs35567484 Chr.16: 30669078-30669079 on Build GRCh38
rs758475880 Chr.16: 30668793-30668793 on Build GRCh38
rs111835702 Chr.16: 30668850-30668850 on Build GRCh38
rs759545282 Chr.16: 30668948-30668948 on Build GRCh38
rs754679103 Chr.16: 30669092-30669092 on Build GRCh38
rs201724059 Chr.16: 30669116-30669116 on Build GRCh38
rs757017507 Chr.16: 30669072-30669072 on Build GRCh38
rs967723639 Chr.16: 30668711-30668711 on Build GRCh38
rs772983220 Chr.16: 30668742-30668743 on Build GRCh38
rs752903003 Chr.16: 30668889-30668889 on Build GRCh38
rs769820389 Chr.16: 30668747-30668747 on Build GRCh38
rs981286158 Chr.16: 30668716-30668716 on Build GRCh38
rs539112912 Chr.16: 30669030-30669030 on Build GRCh38
rs745762130 Chr.16: 30668991-30668991 on Build GRCh38
rs865936430 Chr.16: 30668913-30668913 on Build GRCh38
rs200130245 Chr.16: 30668968-30668968 on Build GRCh38
rs773632675 Chr.16: 30669008-30669008 on Build GRCh38
rs753001934 Chr.16: 30668954-30668954 on Build GRCh38
rs750347547 Chr.16: 30669078-30669078 on Build GRCh38
rs771228797 Chr.16: 30669124-30669124 on Build GRCh38
rs764411493 Chr.16: 30669063-30669063 on Build GRCh38
rs755132317 Chr.16: 30668804-30668804 on Build GRCh38
rs757822798 Chr.16: 30668779-30668779 on Build GRCh38
rs908504790 Chr.16: 30668902-30668902 on Build GRCh38
rs774731584 Chr.16: 30669131-30669131 on Build GRCh38
rs367793438 Chr.16: 30668799-30668799 on Build GRCh38
rs570030199 Chr.16: 30668777-30668777 on Build GRCh38
rs776239889 Chr.16: 30668740-30668740 on Build GRCh38
rs745367456 Chr.16: 30668916-30668916 on Build GRCh38
rs774722034 Chr.16: 30669055-30669055 on Build GRCh38
rs770598435 Chr.16: 30668824-30668824 on Build GRCh38
rs767564835 Chr.16: 30668951-30668951 on Build GRCh38
rs572117442 Chr.16: 30669007-30669007 on Build GRCh38
rs765018102 Chr.16: 30669073-30669073 on Build GRCh38
rs753744419 Chr.16: 30669070-30669070 on Build GRCh38
rs777120720 Chr.16: 30668836-30668836 on Build GRCh38
rs753658772 Chr.16: 30668960-30668960 on Build GRCh38
rs200647927 Chr.16: 30668943-30668943 on Build GRCh38
rs769908280 Chr.16: 30668810-30668810 on Build GRCh38
rs758689250 Chr.16: 30668722-30668722 on Build GRCh38
rs780243756 Chr.16: 30668729-30668729 on Build GRCh38
rs1011340513 Chr.16: 30668708-30668708 on Build GRCh38
rs779565387 Chr.16: 30668996-30668996 on Build GRCh38
rs760279494 Chr.16: 30668931-30668931 on Build GRCh38
rs575660613 Chr.16: 30669062-30669062 on Build GRCh38
rs765730952 Chr.16: 30668783-30668783 on Build GRCh38
rs1011705263 Chr.16: 30669009-30669009 on Build GRCh38
rs1050608454 Chr.16: 30669126-30669126 on Build GRCh38
rs371557670 Chr.16: 30668700-30668700 on Build GRCh38
rs774060423 Chr.16: 30668775-30668775 on Build GRCh38
rs777673949 Chr.16: 30668899-30668899 on Build GRCh38
rs567855481 Chr.16: 30668955-30668955 on Build GRCh38
rs908104029 Chr.16: 30668785-30668785 on Build GRCh38
rs763491230 Chr.16: 30668942-30668942 on Build GRCh38
rs781077356 Chr.16: 30668809-30668809 on Build GRCh38
rs759072873 Chr.16: 30668776-30668776 on Build GRCh38
rs991162041 Chr.16: 30668752-30668752 on Build GRCh38
rs781253246 Chr.16: 30668872-30668872 on Build GRCh38
rs368020065 Chr.16: 30668930-30668930 on Build GRCh38
rs778529104 Chr.16: 30668907-30668907 on Build GRCh38
rs770165391 Chr.16: 30668933-30668933 on Build GRCh38
rs770974158 Chr.16: 30668906-30668906 on Build GRCh38
rs746413394 Chr.16: 30669146-30669146 on Build GRCh38
rs2919433 Chr.16: 30668999-30668999 on Build GRCh38
rs765927124 Chr.16: 30668766-30668766 on Build GRCh38
rs563247495 Chr.16: 30668995-30668995 on Build GRCh38
rs561229767 Chr.16: 30668748-30668749 on Build GRCh38
rs780065741 Chr.16: 30668797-30668797 on Build GRCh38
rs868005057 Chr.16: 30669074-30669074 on Build GRCh38
rs759285388 Chr.16: 30668835-30668835 on Build GRCh38
rs371395419 Chr.16: 30668837-30668837 on Build GRCh38
rs773689008 Chr.16: 30668825-30668825 on Build GRCh38
rs758333525 Chr.16: 30669083-30669083 on Build GRCh38
rs756616817 Chr.16: 30668849-30668849 on Build GRCh38
rs778702468 Chr.16: 30668988-30668988 on Build GRCh38
rs748165740 Chr.16: 30668743-30668743 on Build GRCh38
rs371377757 Chr.16: 30668812-30668812 on Build GRCh38
rs765528853 Chr.16: 30668842-30668842 on Build GRCh38
rs2972815 Chr.16: 30669056-30669056 on Build GRCh38
rs775692051 Chr.16: 30669060-30669060 on Build GRCh38
rs893214162 Chr.16: 30669145-30669145 on Build GRCh38
rs770829251 Chr.16: 30668995-30668996 on Build GRCh38
rs868317279 Chr.16: 30669139-30669139 on Build GRCh38
rs771266045 Chr.16: 30669022-30669022 on Build GRCh38
rs11538956 Chr.16: 30668818-30668818 on Build GRCh38
rs762535917 Chr.16: 30668762-30668762 on Build GRCh38
rs991757309 Chr.16: 30669129-30669129 on Build GRCh38
rs755290850 Chr.16: 30668859-30668859 on Build GRCh38

View Full Product Details