Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31428022-31428523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX6A2
Entrez Gene ID
1339
Gene Name
cytochrome c oxidase subunit 6A2
Gene Aliases
COX6AH, COXVIAH
UniGene
Hs.250760
Gene Chromosome Location
Chr.16: 31427731-31428428 on Build GRCh38
Transcripts
NM_005205.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409652 c.199A>C p.I67L Chr.16: 31428026-31428026 on Build GRCh38
6634351 c.41G>T p.S14I Chr.16: 31428285-31428285 on Build GRCh38
4466190 c.141C>T p.S47S Chr.16: 31428084-31428084 on Build GRCh38
7297064 c.153G>A p.S51S Chr.16: 31428072-31428072 on Build GRCh38
7303828 c.73+2T>C p.? Chr.16: 31428251-31428251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892424165 Chr.16: 31428366-31428366 on Build GRCh38
rs765055668 Chr.16: 31428311-31428311 on Build GRCh38
rs778776784 Chr.16: 31428248-31428248 on Build GRCh38
rs777368777 Chr.16: 31428146-31428146 on Build GRCh38
rs745458434 Chr.16: 31428249-31428249 on Build GRCh38
rs540842412 Chr.16: 31428122-31428122 on Build GRCh38
rs909602856 Chr.16: 31428253-31428253 on Build GRCh38
rs774556987 Chr.16: 31428176-31428177 on Build GRCh38
rs755017137 Chr.16: 31428094-31428094 on Build GRCh38
rs760787848 Chr.16: 31428298-31428298 on Build GRCh38
rs890216794 Chr.16: 31428406-31428406 on Build GRCh38
rs1038680299 Chr.16: 31428450-31428450 on Build GRCh38
rs1050921508 Chr.16: 31428134-31428134 on Build GRCh38
rs200780049 Chr.16: 31428324-31428324 on Build GRCh38
rs897536746 Chr.16: 31428193-31428193 on Build GRCh38
rs752729509 Chr.16: 31428297-31428297 on Build GRCh38
rs1009427334 Chr.16: 31428150-31428150 on Build GRCh38
rs868550507 Chr.16: 31428073-31428073 on Build GRCh38
rs779669707 Chr.16: 31428262-31428262 on Build GRCh38
rs572961091 Chr.16: 31428083-31428083 on Build GRCh38
rs748711535 Chr.16: 31428267-31428267 on Build GRCh38
rs779900385 Chr.16: 31428492-31428492 on Build GRCh38
rs543950432 Chr.16: 31428071-31428071 on Build GRCh38
rs778686952 Chr.16: 31428155-31428155 on Build GRCh38
rs941118349 Chr.16: 31428271-31428271 on Build GRCh38
rs757046966 Chr.16: 31428247-31428247 on Build GRCh38
rs1031463975 Chr.16: 31428281-31428281 on Build GRCh38
rs767619130 Chr.16: 31428296-31428296 on Build GRCh38
rs1051501357 Chr.16: 31428391-31428391 on Build GRCh38
rs968130651 Chr.16: 31428182-31428182 on Build GRCh38
rs1037995114 Chr.16: 31428354-31428354 on Build GRCh38
rs773396385 Chr.16: 31428212-31428212 on Build GRCh38
rs374752640 Chr.16: 31428370-31428370 on Build GRCh38
rs755670336 Chr.16: 31428421-31428421 on Build GRCh38
rs749005782 Chr.16: 31428148-31428148 on Build GRCh38
rs1002300977 Chr.16: 31428388-31428388 on Build GRCh38
rs988670822 Chr.16: 31428080-31428080 on Build GRCh38
rs866320644 Chr.16: 31428140-31428140 on Build GRCh38
rs749748326 Chr.16: 31428353-31428353 on Build GRCh38
rs752639598 Chr.16: 31428240-31428240 on Build GRCh38
rs759672607 Chr.16: 31428229-31428229 on Build GRCh38
rs926879837 Chr.16: 31428154-31428154 on Build GRCh38
rs772329947 Chr.16: 31428375-31428375 on Build GRCh38
rs771706165 Chr.16: 31428173-31428173 on Build GRCh38
rs988229644 Chr.16: 31428283-31428283 on Build GRCh38
rs755953945 Chr.16: 31428144-31428144 on Build GRCh38
rs746537698 Chr.16: 31428338-31428338 on Build GRCh38
rs899720410 Chr.16: 31428116-31428116 on Build GRCh38
rs770249533 Chr.16: 31428268-31428268 on Build GRCh38
rs751684403 Chr.16: 31428088-31428088 on Build GRCh38
rs562235961 Chr.16: 31428187-31428187 on Build GRCh38
rs767531048 Chr.16: 31428239-31428239 on Build GRCh38
rs753811433 Chr.16: 31428300-31428300 on Build GRCh38
rs770441908 Chr.16: 31428149-31428149 on Build GRCh38
rs760166259 Chr.16: 31428192-31428192 on Build GRCh38
rs771269329 Chr.16: 31428274-31428274 on Build GRCh38
rs1007193996 Chr.16: 31428410-31428410 on Build GRCh38
rs753626288 Chr.16: 31428244-31428244 on Build GRCh38
rs777289661 Chr.16: 31428186-31428186 on Build GRCh38
rs781170355 Chr.16: 31428106-31428106 on Build GRCh38
rs763291425 Chr.16: 31428273-31428273 on Build GRCh38
rs115502256 Chr.16: 31428192-31428192 on Build GRCh38
rs998530419 Chr.16: 31428133-31428133 on Build GRCh38
rs960957773 Chr.16: 31428205-31428205 on Build GRCh38
rs763978974 Chr.16: 31428299-31428299 on Build GRCh38
rs115157197 Chr.16: 31428241-31428241 on Build GRCh38
rs750662079 Chr.16: 31428070-31428070 on Build GRCh38
rs73538329 Chr.16: 31428376-31428376 on Build GRCh38
rs200721543 Chr.16: 31428308-31428308 on Build GRCh38
rs766498062 Chr.16: 31428075-31428075 on Build GRCh38
rs1033792906 Chr.16: 31428431-31428431 on Build GRCh38
rs370508400 Chr.16: 31428259-31428259 on Build GRCh38
rs140129800 Chr.16: 31428292-31428292 on Build GRCh38
rs779830938 Chr.16: 31428333-31428333 on Build GRCh38
rs868488221 Chr.16: 31428118-31428118 on Build GRCh38
rs1041641454 Chr.16: 31428217-31428217 on Build GRCh38
rs750129888 Chr.16: 31428318-31428318 on Build GRCh38
rs552611628 Chr.16: 31428286-31428286 on Build GRCh38
rs368585114 Chr.16: 31428361-31428361 on Build GRCh38
rs200209655 Chr.16: 31428328-31428328 on Build GRCh38
rs773734363 Chr.16: 31428270-31428270 on Build GRCh38
rs763100609 Chr.16: 31428126-31428128 on Build GRCh38
rs115517818 Chr.16: 31428466-31428466 on Build GRCh38
rs745413427 Chr.16: 31428164-31428164 on Build GRCh38
rs60662915 Chr.16: 31428227-31428227 on Build GRCh38
rs956100607 Chr.16: 31428282-31428282 on Build GRCh38
rs766437850 Chr.16: 31428174-31428187 on Build GRCh38
rs993255073 Chr.16: 31428214-31428214 on Build GRCh38
rs367655225 Chr.16: 31428350-31428350 on Build GRCh38
rs917916804 Chr.16: 31428111-31428111 on Build GRCh38
rs754503300 Chr.16: 31428342-31428342 on Build GRCh38
rs370671772 Chr.16: 31428216-31428216 on Build GRCh38
rs376029051 Chr.16: 31428360-31428360 on Build GRCh38
rs752619419 Chr.16: 31428126-31428126 on Build GRCh38

View Full Product Details