Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89320870-89321370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 NR_045839.1 XM_006721181.1 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1 NM_001242885.1

Gene Symbol
LOC100287036
Entrez Gene ID
100287036
Gene Name
uncharacterized LOC100287036
Gene Aliases
-
UniGene
Hs.656995
Gene Chromosome Location
Chr.16: 89321133-89325110 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 NR_045839.1 XM_006721181.1 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1 NM_001242885.1

SNP Detail

SNP ID Chromosome Location
rs941446649 Chr.16: 89321096-89321096 on Build GRCh38
rs1035845120 Chr.16: 89321053-89321053 on Build GRCh38
rs934895140 Chr.16: 89320895-89320895 on Build GRCh38
rs149969108 Chr.16: 89320913-89320913 on Build GRCh38
rs970707989 Chr.16: 89321028-89321028 on Build GRCh38
rs1043439018 Chr.16: 89321337-89321337 on Build GRCh38
rs1029592986 Chr.16: 89321116-89321116 on Build GRCh38
rs967822172 Chr.16: 89321134-89321134 on Build GRCh38
rs560115431 Chr.16: 89320931-89320931 on Build GRCh38
rs993379535 Chr.16: 89321213-89321213 on Build GRCh38
rs1037095279 Chr.16: 89321296-89321296 on Build GRCh38
rs889667005 Chr.16: 89321119-89321119 on Build GRCh38
rs3114906 Chr.16: 89321189-89321189 on Build GRCh38
rs943248635 Chr.16: 89320945-89320945 on Build GRCh38
rs943262128 Chr.16: 89321111-89321111 on Build GRCh38
rs147699189 Chr.16: 89321306-89321306 on Build GRCh38
rs531833309 Chr.16: 89321036-89321036 on Build GRCh38
rs924763145 Chr.16: 89321045-89321045 on Build GRCh38
rs540885312 Chr.16: 89320925-89320925 on Build GRCh38
rs991922505 Chr.16: 89321075-89321075 on Build GRCh38
rs144371705 Chr.16: 89321179-89321179 on Build GRCh38
rs901959397 Chr.16: 89320961-89320961 on Build GRCh38
rs142175962 Chr.16: 89321061-89321061 on Build GRCh38
rs180705095 Chr.16: 89321243-89321243 on Build GRCh38
rs901696257 Chr.16: 89321123-89321123 on Build GRCh38
rs956254127 Chr.16: 89321202-89321202 on Build GRCh38
rs1021965707 Chr.16: 89321133-89321133 on Build GRCh38
rs1035917498 Chr.16: 89321222-89321222 on Build GRCh38
rs554479194 Chr.16: 89321224-89321224 on Build GRCh38
rs549460381 Chr.16: 89321296-89321297 on Build GRCh38
rs964379646 Chr.16: 89321310-89321310 on Build GRCh38
rs539096280 Chr.16: 89321284-89321284 on Build GRCh38
rs888534611 Chr.16: 89321090-89321090 on Build GRCh38
rs780732653 Chr.16: 89321180-89321180 on Build GRCh38
rs1053135567 Chr.16: 89321142-89321142 on Build GRCh38
rs889747627 Chr.16: 89321321-89321321 on Build GRCh38
rs570888571 Chr.16: 89320992-89320992 on Build GRCh38
rs933119765 Chr.16: 89321129-89321129 on Build GRCh38
rs974638204 Chr.16: 89321311-89321311 on Build GRCh38
rs984497364 Chr.16: 89321307-89321307 on Build GRCh38
rs1011611638 Chr.16: 89321169-89321169 on Build GRCh38
rs918968405 Chr.16: 89321086-89321086 on Build GRCh38
rs984853608 Chr.16: 89321106-89321106 on Build GRCh38
rs766793048 Chr.16: 89321207-89321207 on Build GRCh38
rs967749891 Chr.16: 89321034-89321034 on Build GRCh38
rs1038904136 Chr.16: 89321121-89321121 on Build GRCh38
rs887686377 Chr.16: 89320944-89320944 on Build GRCh38
rs1044506354 Chr.16: 89320921-89320921 on Build GRCh38
rs186270141 Chr.16: 89321309-89321309 on Build GRCh38
rs1005350406 Chr.16: 89320948-89320948 on Build GRCh38
rs1006559654 Chr.16: 89321132-89321132 on Build GRCh38
rs933194997 Chr.16: 89321323-89321323 on Build GRCh38
rs928930461 Chr.16: 89320916-89320916 on Build GRCh38
rs377344706 Chr.16: 89320962-89320962 on Build GRCh38
rs761921473 Chr.16: 89321055-89321055 on Build GRCh38
rs1011953802 Chr.16: 89320994-89320994 on Build GRCh38
rs948763627 Chr.16: 89321076-89321076 on Build GRCh38
rs998404454 Chr.16: 89321114-89321114 on Build GRCh38
rs1001807005 Chr.16: 89321221-89321221 on Build GRCh38
rs763685052 Chr.16: 89320989-89320989 on Build GRCh38
rs929025420 Chr.16: 89321087-89321087 on Build GRCh38
rs895801581 Chr.16: 89320912-89320912 on Build GRCh38
rs868163333 Chr.16: 89321322-89321322 on Build GRCh38
rs1057449641 Chr.16: 89320906-89320908 on Build GRCh38
rs189422910 Chr.16: 89320951-89320951 on Build GRCh38
rs909157985 Chr.16: 89321109-89321109 on Build GRCh38
rs1031936901 Chr.16: 89321299-89321299 on Build GRCh38
rs776530661 Chr.16: 89321024-89321024 on Build GRCh38
rs953755029 Chr.16: 89321010-89321010 on Build GRCh38
rs1026096602 Chr.16: 89321287-89321287 on Build GRCh38
rs1031686728 Chr.16: 89320986-89320986 on Build GRCh38
rs1021917727 Chr.16: 89321026-89321026 on Build GRCh38
rs891872597 Chr.16: 89321159-89321159 on Build GRCh38

View Full Product Details