Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81121568-81122071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AATK
Entrez Gene ID
9625
Gene Name
apoptosis associated tyrosine kinase
Gene Aliases
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP
UniGene
Hs.514575
Gene Chromosome Location
Chr.17: 81117295-81166072 on Build GRCh38
Transcripts
NM_001080395.2 NM_004920.2 XM_006722193.2 XM_006722194.2 XM_006722195.2 XM_006722196.2 XM_011525509.2 XM_011525510.2 XM_011525511.2 XM_017025421.1 XM_017025422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130793 c.2194C>A p.Q732K Chr.17: 81121742-81121742 on Build GRCh38
6589362 c.1976C>T p.T659I Chr.17: 81121960-81121960 on Build GRCh38
6408858 c.1964C>T p.P655L Chr.17: 81121972-81121972 on Build GRCh38
6589360 c.2044G>A p.V682M Chr.17: 81121892-81121892 on Build GRCh38
1480153 c.2107G>T p.G703C Chr.17: 81121829-81121829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200480348 Chr.17: 81121871-81121871 on Build GRCh38
rs748039722 Chr.17: 81121779-81121779 on Build GRCh38
rs778797422 Chr.17: 81121887-81121887 on Build GRCh38
rs767788616 Chr.17: 81121912-81121912 on Build GRCh38
rs541794558 Chr.17: 81122023-81122023 on Build GRCh38
rs373425173 Chr.17: 81122046-81122046 on Build GRCh38
rs756137049 Chr.17: 81121876-81121876 on Build GRCh38
rs762845072 Chr.17: 81121978-81121978 on Build GRCh38
rs758692493 Chr.17: 81121743-81121743 on Build GRCh38
rs779863844 Chr.17: 81121926-81121926 on Build GRCh38
rs763220135 Chr.17: 81121824-81121824 on Build GRCh38
rs755262818 Chr.17: 81121961-81121962 on Build GRCh38
rs543807684 Chr.17: 81121678-81121678 on Build GRCh38
rs574125102 Chr.17: 81121994-81121994 on Build GRCh38
rs745633908 Chr.17: 81121888-81121888 on Build GRCh38
rs758940818 Chr.17: 81122051-81122051 on Build GRCh38
rs979618973 Chr.17: 81121681-81121681 on Build GRCh38
rs753655906 Chr.17: 81121884-81121884 on Build GRCh38
rs887357987 Chr.17: 81121599-81121599 on Build GRCh38
rs764669123 Chr.17: 81121613-81121613 on Build GRCh38
rs776612070 Chr.17: 81121972-81121972 on Build GRCh38
rs780107147 Chr.17: 81121761-81121761 on Build GRCh38
rs770687787 Chr.17: 81121802-81121802 on Build GRCh38
rs571083150 Chr.17: 81121813-81121813 on Build GRCh38
rs1031630259 Chr.17: 81121651-81121651 on Build GRCh38
rs779847368 Chr.17: 81121891-81121891 on Build GRCh38
rs773578536 Chr.17: 81121864-81121864 on Build GRCh38
rs773864534 Chr.17: 81121803-81121803 on Build GRCh38
rs758161250 Chr.17: 81121890-81121890 on Build GRCh38
rs866925925 Chr.17: 81121614-81121614 on Build GRCh38
rs766387357 Chr.17: 81122040-81122040 on Build GRCh38
rs751104783 Chr.17: 81121928-81121928 on Build GRCh38
rs747364906 Chr.17: 81122019-81122019 on Build GRCh38
rs373446518 Chr.17: 81122032-81122032 on Build GRCh38
rs181074632 Chr.17: 81121608-81121608 on Build GRCh38
rs762375873 Chr.17: 81121597-81121597 on Build GRCh38
rs758253045 Chr.17: 81121923-81121923 on Build GRCh38
rs993179686 Chr.17: 81121782-81121782 on Build GRCh38
rs769599692 Chr.17: 81121783-81121783 on Build GRCh38
rs527352561 Chr.17: 81122033-81122033 on Build GRCh38
rs777570783 Chr.17: 81121836-81121836 on Build GRCh38
rs377604606 Chr.17: 81121660-81121660 on Build GRCh38
rs747665089 Chr.17: 81121946-81121946 on Build GRCh38
rs948564678 Chr.17: 81121671-81121671 on Build GRCh38
rs751911292 Chr.17: 81121874-81121876 on Build GRCh38
rs780616846 Chr.17: 81121941-81121941 on Build GRCh38
rs765636578 Chr.17: 81121817-81121817 on Build GRCh38
rs771408924 Chr.17: 81121948-81121948 on Build GRCh38
rs779986234 Chr.17: 81121966-81121966 on Build GRCh38
rs562437913 Chr.17: 81121980-81121980 on Build GRCh38
rs758434134 Chr.17: 81121653-81121653 on Build GRCh38
rs560006855 Chr.17: 81122024-81122024 on Build GRCh38
rs907616671 Chr.17: 81121932-81121932 on Build GRCh38
rs780129026 Chr.17: 81121658-81121658 on Build GRCh38
rs763550537 Chr.17: 81122047-81122047 on Build GRCh38
rs865962912 Chr.17: 81121664-81121664 on Build GRCh38
rs930671942 Chr.17: 81121765-81121765 on Build GRCh38
rs199534891 Chr.17: 81121878-81121878 on Build GRCh38
rs368788026 Chr.17: 81122045-81122045 on Build GRCh38
rs201872744 Chr.17: 81121687-81121687 on Build GRCh38
rs750838086 Chr.17: 81121821-81121821 on Build GRCh38
rs747036680 Chr.17: 81121659-81121659 on Build GRCh38
rs749709313 Chr.17: 81121901-81121901 on Build GRCh38
rs1031641443 Chr.17: 81121778-81121778 on Build GRCh38
rs761945776 Chr.17: 81121973-81121973 on Build GRCh38
rs61738829 Chr.17: 81121907-81121907 on Build GRCh38
rs754322629 Chr.17: 81121627-81121627 on Build GRCh38
rs775634996 Chr.17: 81121913-81121913 on Build GRCh38
rs764758940 Chr.17: 81121714-81121714 on Build GRCh38
rs995371771 Chr.17: 81121595-81121595 on Build GRCh38
rs762617969 Chr.17: 81121703-81121703 on Build GRCh38
rs760756271 Chr.17: 81121967-81121967 on Build GRCh38
rs770397466 Chr.17: 81121861-81121861 on Build GRCh38
rs1049587752 Chr.17: 81121773-81121773 on Build GRCh38
rs781210369 Chr.17: 81121962-81121967 on Build GRCh38
rs747486246 Chr.17: 81121717-81121717 on Build GRCh38
rs968733057 Chr.17: 81121649-81121649 on Build GRCh38
rs765655662 Chr.17: 81121736-81121736 on Build GRCh38
rs374375526 Chr.17: 81121709-81121709 on Build GRCh38
rs779382765 Chr.17: 81121953-81121953 on Build GRCh38
rs369359391 Chr.17: 81121921-81121921 on Build GRCh38
rs765028859 Chr.17: 81121774-81121774 on Build GRCh38
rs769513007 Chr.17: 81121680-81121680 on Build GRCh38
rs769926715 Chr.17: 81122030-81122030 on Build GRCh38
rs374945846 Chr.17: 81121920-81121920 on Build GRCh38
rs764044234 Chr.17: 81121918-81121918 on Build GRCh38
rs745474725 Chr.17: 81121847-81121847 on Build GRCh38
rs763388197 Chr.17: 81121865-81121865 on Build GRCh38
rs997315692 Chr.17: 81121622-81121622 on Build GRCh38
rs386799844 Chr.17: 81121829-81121842 on Build GRCh38
rs905788705 Chr.17: 81121996-81121996 on Build GRCh38
rs773773077 Chr.17: 81121900-81121900 on Build GRCh38
rs767537196 Chr.17: 81121833-81121833 on Build GRCh38
rs774578188 Chr.17: 81121870-81121870 on Build GRCh38
rs753320625 Chr.17: 81122049-81122049 on Build GRCh38
rs766613950 Chr.17: 81121866-81121866 on Build GRCh38
rs866080761 Chr.17: 81121727-81121727 on Build GRCh38
rs772452179 Chr.17: 81122018-81122018 on Build GRCh38
rs779399465 Chr.17: 81122011-81122011 on Build GRCh38
rs746338931 Chr.17: 81122012-81122012 on Build GRCh38
rs369982677 Chr.17: 81122001-81122001 on Build GRCh38
rs746483226 Chr.17: 81121852-81121852 on Build GRCh38
rs748280166 Chr.17: 81121972-81121972 on Build GRCh38
rs905941355 Chr.17: 81121589-81121589 on Build GRCh38
rs767993072 Chr.17: 81121897-81121897 on Build GRCh38
rs978748500 Chr.17: 81121650-81121650 on Build GRCh38
rs772773081 Chr.17: 81121688-81121688 on Build GRCh38
rs746573327 Chr.17: 81121895-81121895 on Build GRCh38
rs1053228822 Chr.17: 81121979-81121979 on Build GRCh38
rs975769352 Chr.17: 81121952-81121952 on Build GRCh38
rs7503604 Chr.17: 81121829-81121829 on Build GRCh38
rs940248016 Chr.17: 81121718-81121718 on Build GRCh38
rs149568038 Chr.17: 81121789-81121789 on Build GRCh38
rs370949748 Chr.17: 81121706-81121706 on Build GRCh38
rs757686596 Chr.17: 81121632-81121632 on Build GRCh38
rs1028859405 Chr.17: 81121758-81121758 on Build GRCh38
rs1014674990 Chr.17: 81121982-81121982 on Build GRCh38
rs544139724 Chr.17: 81121968-81121968 on Build GRCh38
rs756087199 Chr.17: 81121834-81121834 on Build GRCh38
rs763033967 Chr.17: 81122039-81122039 on Build GRCh38
rs1028644393 Chr.17: 81121830-81121830 on Build GRCh38
rs917049012 Chr.17: 81121666-81121666 on Build GRCh38
rs771638560 Chr.17: 81121808-81121808 on Build GRCh38
rs750589133 Chr.17: 81121648-81121648 on Build GRCh38
rs772360634 Chr.17: 81121958-81121958 on Build GRCh38
rs1018775772 Chr.17: 81121631-81121631 on Build GRCh38
rs762270035 Chr.17: 81121726-81121726 on Build GRCh38
rs56157111 Chr.17: 81121707-81121707 on Build GRCh38
rs200822708 Chr.17: 81122041-81122041 on Build GRCh38
rs984814728 Chr.17: 81121892-81121892 on Build GRCh38
rs750628893 Chr.17: 81121738-81121738 on Build GRCh38
rs558671948 Chr.17: 81121922-81121922 on Build GRCh38
rs772864214 Chr.17: 81121593-81121593 on Build GRCh38
rs1004340211 Chr.17: 81121602-81121602 on Build GRCh38
rs754342730 Chr.17: 81121720-81121720 on Build GRCh38
rs767696896 Chr.17: 81121872-81121872 on Build GRCh38
rs767093144 Chr.17: 81122042-81122042 on Build GRCh38
rs780895354 Chr.17: 81121995-81121995 on Build GRCh38
rs200015615 Chr.17: 81121672-81121672 on Build GRCh38
rs779287877 Chr.17: 81121639-81121639 on Build GRCh38
rs765873050 Chr.17: 81121605-81121605 on Build GRCh38
rs1040218204 Chr.17: 81121795-81121795 on Build GRCh38
rs560172011 Chr.17: 81121771-81121771 on Build GRCh38
rs775724904 Chr.17: 81121960-81121960 on Build GRCh38
rs372359503 Chr.17: 81121977-81121977 on Build GRCh38
rs55846463 Chr.17: 81121842-81121842 on Build GRCh38
rs771726396 Chr.17: 81121851-81121851 on Build GRCh38
rs774739437 Chr.17: 81121910-81121910 on Build GRCh38
rs751726040 Chr.17: 81121762-81121762 on Build GRCh38
rs538497193 Chr.17: 81121815-81121815 on Build GRCh38
rs754539605 Chr.17: 81121930-81121930 on Build GRCh38
rs753167913 Chr.17: 81121607-81121607 on Build GRCh38
rs778635339 Chr.17: 81121843-81121843 on Build GRCh38
rs759891280 Chr.17: 81121911-81121911 on Build GRCh38
rs752207629 Chr.17: 81121999-81121999 on Build GRCh38
rs376402794 Chr.17: 81121788-81121788 on Build GRCh38
rs554007171 Chr.17: 81121875-81121875 on Build GRCh38
rs55881512 Chr.17: 81121956-81121956 on Build GRCh38
rs768825150 Chr.17: 81122022-81122022 on Build GRCh38
rs765321357 Chr.17: 81121976-81121976 on Build GRCh38

View Full Product Details