Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62715562-62716078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP1
Entrez Gene ID
23239
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
PHLPP, PLEKHE1, PPM3A, SCOP
UniGene
Hs.465337
Gene Chromosome Location
Chr.18: 62715439-62980443 on Build GRCh38
Transcripts
NM_194449.3 XM_011525886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5446119 c.78_119del42 p.A27_A40del14 Chr.18: 62715761-62715802 on Build GRCh38
7292172 c.344G>C p.G115A Chr.18: 62716027-62716027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs921375996 Chr.18: 62715898-62715898 on Build GRCh38
rs1016774262 Chr.18: 62715793-62715793 on Build GRCh38
rs952173463 Chr.18: 62715765-62715797 on Build GRCh38
rs973861142 Chr.18: 62715806-62715806 on Build GRCh38
rs960391075 Chr.18: 62715832-62715832 on Build GRCh38
rs113217057 Chr.18: 62715583-62715583 on Build GRCh38
rs962823841 Chr.18: 62715794-62715794 on Build GRCh38
rs754224940 Chr.18: 62715771-62715773 on Build GRCh38
rs11152356 Chr.18: 62715838-62715838 on Build GRCh38
rs768418881 Chr.18: 62716044-62716044 on Build GRCh38
rs916227851 Chr.18: 62715878-62715878 on Build GRCh38
rs948962744 Chr.18: 62715934-62715934 on Build GRCh38
rs891860721 Chr.18: 62716014-62716014 on Build GRCh38
rs867992189 Chr.18: 62715808-62715808 on Build GRCh38
rs780740012 Chr.18: 62715695-62715695 on Build GRCh38
rs578087800 Chr.18: 62715650-62715650 on Build GRCh38
rs918325255 Chr.18: 62715829-62715829 on Build GRCh38
rs1000287404 Chr.18: 62715748-62715748 on Build GRCh38
rs926181674 Chr.18: 62715953-62715953 on Build GRCh38
rs140365112 Chr.18: 62715656-62715657 on Build GRCh38
rs556506288 Chr.18: 62715992-62715992 on Build GRCh38
rs558200606 Chr.18: 62715638-62715638 on Build GRCh38
rs1007188094 Chr.18: 62715791-62715791 on Build GRCh38
rs745311627 Chr.18: 62715698-62715698 on Build GRCh38
rs998096661 Chr.18: 62715592-62715592 on Build GRCh38
rs779924561 Chr.18: 62715760-62715760 on Build GRCh38
rs953981343 Chr.18: 62715786-62715794 on Build GRCh38
rs936158178 Chr.18: 62715997-62715997 on Build GRCh38
rs948890983 Chr.18: 62715708-62715708 on Build GRCh38
rs566190899 Chr.18: 62715600-62715600 on Build GRCh38
rs1029556181 Chr.18: 62715783-62715783 on Build GRCh38
rs1045009351 Chr.18: 62715935-62715935 on Build GRCh38
rs756780211 Chr.18: 62715692-62715692 on Build GRCh38
rs576395177 Chr.18: 62715998-62715998 on Build GRCh38
rs998127429 Chr.18: 62715769-62715769 on Build GRCh38
rs953908160 Chr.18: 62715604-62715604 on Build GRCh38
rs972863614 Chr.18: 62715684-62715684 on Build GRCh38
rs962503323 Chr.18: 62715664-62715664 on Build GRCh38
rs534795618 Chr.18: 62715602-62715602 on Build GRCh38
rs1057416530 Chr.18: 62715699-62715699 on Build GRCh38
rs956194675 Chr.18: 62715590-62715590 on Build GRCh38
rs756047746 Chr.18: 62715700-62715700 on Build GRCh38
rs934052860 Chr.18: 62716020-62716020 on Build GRCh38
rs904661724 Chr.18: 62715745-62715745 on Build GRCh38
rs751189889 Chr.18: 62715634-62715634 on Build GRCh38
rs111833539 Chr.18: 62715729-62715729 on Build GRCh38
rs1031684749 Chr.18: 62715589-62715589 on Build GRCh38
rs1044588713 Chr.18: 62715730-62715730 on Build GRCh38
rs768316835 Chr.18: 62715606-62715607 on Build GRCh38
rs543794884 Chr.18: 62715842-62715842 on Build GRCh38
rs766674717 Chr.18: 62715770-62715771 on Build GRCh38

View Full Product Details