Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62716513-62717001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP1
Entrez Gene ID
23239
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases
PHLPP, PLEKHE1, PPM3A, SCOP
UniGene
Hs.465337
Gene Chromosome Location
Chr.18: 62715439-62980443 on Build GRCh38
Transcripts
NM_194449.3 XM_011525886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017053 c.884C>T p.P295L Chr.18: 62716567-62716567 on Build GRCh38
3756193 c.1066G>A p.A356T Chr.18: 62716749-62716749 on Build GRCh38
4957998 c.1201G>T p.A401S Chr.18: 62716884-62716884 on Build GRCh38
6382571 c.1295A>G p.E432G Chr.18: 62716978-62716978 on Build GRCh38
5813261 c.1296G>T p.E432D Chr.18: 62716979-62716979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747843451 Chr.18: 62716650-62716650 on Build GRCh38
rs778653099 Chr.18: 62716897-62716897 on Build GRCh38
rs951858596 Chr.18: 62716881-62716881 on Build GRCh38
rs955642336 Chr.18: 62716869-62716869 on Build GRCh38
rs746927706 Chr.18: 62716910-62716910 on Build GRCh38
rs769959686 Chr.18: 62716721-62716721 on Build GRCh38
rs945107674 Chr.18: 62716966-62716966 on Build GRCh38
rs904958396 Chr.18: 62716629-62716629 on Build GRCh38
rs571730566 Chr.18: 62716960-62716960 on Build GRCh38
rs935785132 Chr.18: 62716802-62716802 on Build GRCh38
rs1051393057 Chr.18: 62716968-62716968 on Build GRCh38
rs1053536557 Chr.18: 62716853-62716853 on Build GRCh38
rs780291195 Chr.18: 62716980-62716980 on Build GRCh38
rs1010481368 Chr.18: 62716695-62716695 on Build GRCh38
rs896354097 Chr.18: 62716622-62716622 on Build GRCh38
rs1013411693 Chr.18: 62716623-62716623 on Build GRCh38
rs758082051 Chr.18: 62716908-62716908 on Build GRCh38
rs1020487909 Chr.18: 62716706-62716706 on Build GRCh38
rs945675054 Chr.18: 62716927-62716927 on Build GRCh38
rs984585153 Chr.18: 62716892-62716892 on Build GRCh38
rs867055774 Chr.18: 62716679-62716679 on Build GRCh38
rs540790768 Chr.18: 62716544-62716544 on Build GRCh38
rs955569313 Chr.18: 62716718-62716718 on Build GRCh38
rs904988349 Chr.18: 62716846-62716846 on Build GRCh38
rs779380478 Chr.18: 62716723-62716723 on Build GRCh38
rs535194355 Chr.18: 62716884-62716884 on Build GRCh38
rs1046977032 Chr.18: 62716599-62716599 on Build GRCh38
rs974151861 Chr.18: 62716776-62716776 on Build GRCh38
rs894196098 Chr.18: 62716823-62716823 on Build GRCh38
rs763536706 Chr.18: 62716736-62716736 on Build GRCh38
rs1018527303 Chr.18: 62716981-62716981 on Build GRCh38
rs762042568 Chr.18: 62716830-62716830 on Build GRCh38
rs949288369 Chr.18: 62716943-62716943 on Build GRCh38
rs919925107 Chr.18: 62716780-62716781 on Build GRCh38
rs1055192840 Chr.18: 62716961-62716961 on Build GRCh38
rs974603382 Chr.18: 62716899-62716899 on Build GRCh38
rs892198400 Chr.18: 62716964-62716964 on Build GRCh38
rs980672301 Chr.18: 62716945-62716945 on Build GRCh38
rs566159756 Chr.18: 62716867-62716867 on Build GRCh38
rs1032430416 Chr.18: 62716659-62716659 on Build GRCh38
rs529519334 Chr.18: 62716704-62716704 on Build GRCh38
rs551589785 Chr.18: 62716958-62716958 on Build GRCh38
rs752571717 Chr.18: 62716902-62716902 on Build GRCh38
rs936861188 Chr.18: 62716852-62716852 on Build GRCh38
rs1047042335 Chr.18: 62716807-62716807 on Build GRCh38
rs1044859533 Chr.18: 62716624-62716624 on Build GRCh38
rs948458342 Chr.18: 62716974-62716974 on Build GRCh38
rs776201614 Chr.18: 62716734-62716734 on Build GRCh38
rs12957017 Chr.18: 62716749-62716749 on Build GRCh38
rs754927182 Chr.18: 62716874-62716874 on Build GRCh38
rs532772124 Chr.18: 62716560-62716560 on Build GRCh38
rs1054128849 Chr.18: 62716808-62716808 on Build GRCh38
rs368493874 Chr.18: 62716909-62716909 on Build GRCh38
rs1001024913 Chr.18: 62716851-62716851 on Build GRCh38
rs929991204 Chr.18: 62716799-62716799 on Build GRCh38
rs377619069 Chr.18: 62716665-62716665 on Build GRCh38
rs1038795971 Chr.18: 62716548-62716548 on Build GRCh38
rs978740996 Chr.18: 62716949-62716949 on Build GRCh38
rs760220288 Chr.18: 62716843-62716843 on Build GRCh38
rs76447412 Chr.18: 62716694-62716694 on Build GRCh38
rs1024370338 Chr.18: 62716854-62716854 on Build GRCh38
rs915682044 Chr.18: 62716970-62716970 on Build GRCh38
rs549353310 Chr.18: 62716593-62716593 on Build GRCh38
rs750057150 Chr.18: 62716838-62716838 on Build GRCh38
rs774782660 Chr.18: 62716805-62716805 on Build GRCh38
rs771192395 Chr.18: 62716684-62716684 on Build GRCh38
rs907143304 Chr.18: 62716861-62716861 on Build GRCh38
rs745991504 Chr.18: 62716708-62716708 on Build GRCh38
rs923244955 Chr.18: 62716920-62716920 on Build GRCh38
rs770054324 Chr.18: 62716978-62716978 on Build GRCh38
rs187210763 Chr.18: 62716554-62716554 on Build GRCh38

View Full Product Details