Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2512653-2513144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG7
Entrez Gene ID
2788
Gene Name
G protein subunit gamma 7
Gene Aliases
-
UniGene
Hs.515544
Gene Chromosome Location
Chr.19: 2511219-2702748 on Build GRCh38
Transcripts
NM_052847.2 XM_017026606.1 XM_005259695.2

Gene Symbol
LOC101929097
Entrez Gene ID
101929097
Gene Name
uncharacterized LOC101929097
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.19: 2511219-2513571 on Build GRCh38
Transcripts
NM_052847.2 XM_017026606.1 XM_005259695.2

SNP Detail

SNP ID Chromosome Location
rs939818667 Chr.19: 2512898-2512898 on Build GRCh38
rs968806968 Chr.19: 2512863-2512863 on Build GRCh38
rs749817491 Chr.19: 2512724-2512724 on Build GRCh38
rs377750648 Chr.19: 2512781-2512781 on Build GRCh38
rs1009965052 Chr.19: 2513000-2513000 on Build GRCh38
rs930098429 Chr.19: 2512733-2512733 on Build GRCh38
rs1051250846 Chr.19: 2512983-2512983 on Build GRCh38
rs958647756 Chr.19: 2512892-2512892 on Build GRCh38
rs951222018 Chr.19: 2513054-2513054 on Build GRCh38
rs909888975 Chr.19: 2512765-2512765 on Build GRCh38
rs983291669 Chr.19: 2512928-2512928 on Build GRCh38
rs374553428 Chr.19: 2512727-2512727 on Build GRCh38
rs914032170 Chr.19: 2513097-2513097 on Build GRCh38
rs547884595 Chr.19: 2513125-2513125 on Build GRCh38
rs987067703 Chr.19: 2513085-2513085 on Build GRCh38
rs892650927 Chr.19: 2512995-2512995 on Build GRCh38
rs917111087 Chr.19: 2512902-2512902 on Build GRCh38
rs951441217 Chr.19: 2512925-2512925 on Build GRCh38
rs1014140279 Chr.19: 2513033-2513033 on Build GRCh38
rs552449495 Chr.19: 2512888-2512888 on Build GRCh38
rs7248417 Chr.19: 2512797-2512797 on Build GRCh38
rs1019919560 Chr.19: 2512862-2512862 on Build GRCh38
rs909846968 Chr.19: 2512939-2512939 on Build GRCh38
rs962866396 Chr.19: 2513065-2513065 on Build GRCh38
rs934202487 Chr.19: 2513076-2513076 on Build GRCh38
rs889331686 Chr.19: 2512705-2512705 on Build GRCh38
rs116102055 Chr.19: 2512849-2512849 on Build GRCh38
rs1041859783 Chr.19: 2513002-2513002 on Build GRCh38
rs927123851 Chr.19: 2512691-2512691 on Build GRCh38
rs990117689 Chr.19: 2512833-2512833 on Build GRCh38
rs934255053 Chr.19: 2512982-2512982 on Build GRCh38
rs535587793 Chr.19: 2513098-2513098 on Build GRCh38
rs921294837 Chr.19: 2513074-2513074 on Build GRCh38
rs921512686 Chr.19: 2512973-2512973 on Build GRCh38
rs1034686603 Chr.19: 2512880-2512880 on Build GRCh38
rs981750032 Chr.19: 2512872-2512872 on Build GRCh38
rs1036757159 Chr.19: 2512908-2512908 on Build GRCh38
rs982630329 Chr.19: 2513011-2513011 on Build GRCh38
rs377174635 Chr.19: 2513027-2513027 on Build GRCh38
rs1047040366 Chr.19: 2512758-2512758 on Build GRCh38
rs970453453 Chr.19: 2513041-2513041 on Build GRCh38
rs948705486 Chr.19: 2512871-2512871 on Build GRCh38
rs940555233 Chr.19: 2513029-2513029 on Build GRCh38
rs992615598 Chr.19: 2512900-2512900 on Build GRCh38
rs909951255 Chr.19: 2513058-2513058 on Build GRCh38
rs564812342 Chr.19: 2513015-2513015 on Build GRCh38
rs1011666645 Chr.19: 2513032-2513032 on Build GRCh38
rs116227271 Chr.19: 2512678-2512678 on Build GRCh38
rs537756931 Chr.19: 2513023-2513023 on Build GRCh38
rs116711730 Chr.19: 2513016-2513016 on Build GRCh38
rs930986026 Chr.19: 2512935-2512935 on Build GRCh38
rs889445948 Chr.19: 2512981-2512981 on Build GRCh38
rs992836649 Chr.19: 2512722-2512722 on Build GRCh38
rs759689482 Chr.19: 2513044-2513045 on Build GRCh38
rs569084738 Chr.19: 2512798-2512798 on Build GRCh38
rs941599464 Chr.19: 2512769-2512769 on Build GRCh38
rs890191248 Chr.19: 2512845-2512845 on Build GRCh38
rs1037583469 Chr.19: 2513035-2513035 on Build GRCh38
rs982853424 Chr.19: 2513056-2513056 on Build GRCh38
rs746539625 Chr.19: 2513073-2513073 on Build GRCh38
rs1024071240 Chr.19: 2513042-2513042 on Build GRCh38
rs1040363264 Chr.19: 2512785-2512785 on Build GRCh38
rs917322426 Chr.19: 2512729-2512729 on Build GRCh38
rs557748979 Chr.19: 2513059-2513059 on Build GRCh38
rs1027863642 Chr.19: 2512702-2512702 on Build GRCh38
rs1039742932 Chr.19: 2512960-2512960 on Build GRCh38
rs756614831 Chr.19: 2512677-2512677 on Build GRCh38
rs941367483 Chr.19: 2512944-2512944 on Build GRCh38
rs919751219 Chr.19: 2513091-2513091 on Build GRCh38
rs17755100 Chr.19: 2512878-2512878 on Build GRCh38
rs979099409 Chr.19: 2512690-2512690 on Build GRCh38
rs961237216 Chr.19: 2513058-2513059 on Build GRCh38
rs977546776 Chr.19: 2512740-2512740 on Build GRCh38
rs898209731 Chr.19: 2512920-2512920 on Build GRCh38
rs778301188 Chr.19: 2512721-2512721 on Build GRCh38
rs538969090 Chr.19: 2512819-2512819 on Build GRCh38
rs975801429 Chr.19: 2513068-2513068 on Build GRCh38

View Full Product Details