Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11553869-11554401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOF1
Entrez Gene ID
84337
Gene Name
elongation factor 1 homolog
Gene Aliases
ELF1
UniGene
Hs.631633
Gene Chromosome Location
Chr.19: 11553043-11559236 on Build GRCh38
Transcripts
NM_032377.3 XM_017027356.1 XM_017027357.1 XM_017027358.1 XM_017027359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980644 c.11G>A p.R4K Chr.19: 11554337-11554337 on Build GRCh38
5685269 c.58C>A p.L20I Chr.19: 11554290-11554290 on Build GRCh38
2814674 c.177G>A p.T59T Chr.19: 11554021-11554021 on Build GRCh38
990953 c.163G>A p.E55K Chr.19: 11554035-11554035 on Build GRCh38
2814679 c.8G>A p.R3H Chr.19: 11554340-11554340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747766890 Chr.19: 11554317-11554317 on Build GRCh38
rs140013085 Chr.19: 11554367-11554367 on Build GRCh38
rs1016766084 Chr.19: 11554150-11554150 on Build GRCh38
rs775440532 Chr.19: 11554069-11554069 on Build GRCh38
rs781592525 Chr.19: 11554022-11554022 on Build GRCh38
rs373549538 Chr.19: 11553947-11553947 on Build GRCh38
rs769360384 Chr.19: 11554318-11554318 on Build GRCh38
rs572276283 Chr.19: 11554348-11554348 on Build GRCh38
rs776866141 Chr.19: 11554358-11554358 on Build GRCh38
rs1054619391 Chr.19: 11553971-11553971 on Build GRCh38
rs750085439 Chr.19: 11554233-11554233 on Build GRCh38
rs758957440 Chr.19: 11553891-11553891 on Build GRCh38
rs372944360 Chr.19: 11554063-11554063 on Build GRCh38
rs943558132 Chr.19: 11554159-11554159 on Build GRCh38
rs548002884 Chr.19: 11554047-11554047 on Build GRCh38
rs762056453 Chr.19: 11554360-11554360 on Build GRCh38
rs569923731 Chr.19: 11554114-11554114 on Build GRCh38
rs373143619 Chr.19: 11554188-11554188 on Build GRCh38
rs768030543 Chr.19: 11554245-11554245 on Build GRCh38
rs760046806 Chr.19: 11554103-11554103 on Build GRCh38
rs779955819 Chr.19: 11554196-11554196 on Build GRCh38
rs569671018 Chr.19: 11554164-11554164 on Build GRCh38
rs778920880 Chr.19: 11554183-11554183 on Build GRCh38
rs745744285 Chr.19: 11554184-11554184 on Build GRCh38
rs776021530 Chr.19: 11554247-11554247 on Build GRCh38
rs554721120 Chr.19: 11554379-11554379 on Build GRCh38
rs766886574 Chr.19: 11554012-11554012 on Build GRCh38
rs910288369 Chr.19: 11554142-11554142 on Build GRCh38
rs770472199 Chr.19: 11554325-11554325 on Build GRCh38
rs761001102 Chr.19: 11554252-11554252 on Build GRCh38
rs150001226 Chr.19: 11554312-11554312 on Build GRCh38
rs773964581 Chr.19: 11553954-11553954 on Build GRCh38
rs769852920 Chr.19: 11553926-11553926 on Build GRCh38
rs767464156 Chr.19: 11553957-11553957 on Build GRCh38
rs761429233 Chr.19: 11554216-11554216 on Build GRCh38
rs753393563 Chr.19: 11553977-11553977 on Build GRCh38
rs1047764976 Chr.19: 11554197-11554197 on Build GRCh38
rs760464507 Chr.19: 11553961-11553961 on Build GRCh38
rs778038400 Chr.19: 11554132-11554132 on Build GRCh38
rs375925429 Chr.19: 11554070-11554070 on Build GRCh38
rs780860283 Chr.19: 11554315-11554315 on Build GRCh38
rs774246380 Chr.19: 11553967-11553967 on Build GRCh38
rs11539252 Chr.19: 11554077-11554077 on Build GRCh38
rs1012823896 Chr.19: 11554165-11554165 on Build GRCh38
rs943613533 Chr.19: 11554007-11554007 on Build GRCh38
rs758363350 Chr.19: 11554293-11554293 on Build GRCh38
rs755727195 Chr.19: 11554341-11554341 on Build GRCh38
rs770288417 Chr.19: 11553954-11553970 on Build GRCh38
rs769288429 Chr.19: 11554224-11554224 on Build GRCh38
rs752856314 Chr.19: 11554032-11554032 on Build GRCh38
rs202130414 Chr.19: 11554324-11554324 on Build GRCh38
rs757437211 Chr.19: 11554287-11554287 on Build GRCh38
rs957104202 Chr.19: 11554246-11554246 on Build GRCh38
rs954375015 Chr.19: 11554127-11554127 on Build GRCh38
rs939326753 Chr.19: 11554273-11554273 on Build GRCh38
rs532574810 Chr.19: 11554042-11554042 on Build GRCh38
rs369142001 Chr.19: 11553995-11553995 on Build GRCh38
rs112322829 Chr.19: 11554215-11554215 on Build GRCh38
rs200004332 Chr.19: 11554107-11554107 on Build GRCh38
rs780117768 Chr.19: 11554305-11554305 on Build GRCh38
rs746822654 Chr.19: 11554198-11554198 on Build GRCh38
rs768454338 Chr.19: 11554071-11554071 on Build GRCh38
rs764718942 Chr.19: 11554096-11554096 on Build GRCh38
rs977062822 Chr.19: 11554102-11554102 on Build GRCh38
rs531343745 Chr.19: 11553900-11553900 on Build GRCh38
rs780216262 Chr.19: 11553892-11553892 on Build GRCh38
rs369144969 Chr.19: 11554119-11554119 on Build GRCh38
rs745870835 Chr.19: 11554062-11554062 on Build GRCh38
rs767264985 Chr.19: 11554179-11554179 on Build GRCh38
rs775976345 Chr.19: 11554340-11554340 on Build GRCh38
rs1031456862 Chr.19: 11554223-11554223 on Build GRCh38
rs754080376 Chr.19: 11554279-11554279 on Build GRCh38
rs1040804463 Chr.19: 11554009-11554009 on Build GRCh38
rs1047767287 Chr.19: 11554118-11554118 on Build GRCh38
rs7248573 Chr.19: 11553908-11553908 on Build GRCh38
rs776482035 Chr.19: 11553967-11553967 on Build GRCh38
rs750724225 Chr.19: 11554372-11554372 on Build GRCh38
rs764437047 Chr.19: 11554255-11554255 on Build GRCh38
rs764155783 Chr.19: 11554279-11554279 on Build GRCh38
rs973574124 Chr.19: 11554175-11554175 on Build GRCh38
rs752045865 Chr.19: 11554017-11554017 on Build GRCh38
rs773358169 Chr.19: 11553936-11553936 on Build GRCh38
rs747432383 Chr.19: 11553898-11553898 on Build GRCh38
rs756276387 Chr.19: 11554123-11554123 on Build GRCh38
rs752091371 Chr.19: 11554101-11554101 on Build GRCh38
rs943633264 Chr.19: 11554319-11554319 on Build GRCh38
rs566030761 Chr.19: 11554084-11554084 on Build GRCh38
rs761355214 Chr.19: 11554092-11554092 on Build GRCh38
rs745509079 Chr.19: 11554129-11554131 on Build GRCh38
rs749116401 Chr.19: 11553937-11553937 on Build GRCh38
rs958898033 Chr.19: 11554153-11554153 on Build GRCh38
rs571553586 Chr.19: 11554205-11554205 on Build GRCh38
rs775279247 Chr.19: 11553958-11553958 on Build GRCh38
rs755330142 Chr.19: 11554021-11554021 on Build GRCh38
rs868815582 Chr.19: 11554141-11554141 on Build GRCh38
rs906624502 Chr.19: 11553968-11553968 on Build GRCh38
rs759993365 Chr.19: 11554240-11554240 on Build GRCh38
rs538448238 Chr.19: 11554225-11554225 on Build GRCh38
rs770969300 Chr.19: 11554048-11554048 on Build GRCh38
rs778882533 Chr.19: 11554051-11554051 on Build GRCh38
rs748438124 Chr.19: 11553925-11553925 on Build GRCh38
rs537060999 Chr.19: 11554181-11554181 on Build GRCh38
rs756331430 Chr.19: 11554040-11554040 on Build GRCh38
rs765296221 Chr.19: 11554288-11554288 on Build GRCh38
rs140836187 Chr.19: 11554311-11554311 on Build GRCh38
rs768375595 Chr.19: 11554201-11554201 on Build GRCh38
rs146100806 Chr.19: 11554182-11554182 on Build GRCh38

View Full Product Details