Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16402027-16402547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15L1
Entrez Gene ID
58513
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases
EPS15R
UniGene
Hs.654639
Gene Chromosome Location
Chr.19: 16355244-16472012 on Build GRCh38
Transcripts
NM_001258374.1 NM_001258375.1 NM_001258376.1 NM_021235.2 NR_047665.1 NR_047666.1 XM_017027086.1 XM_017027087.1 XM_017027088.1 XM_017027089.1 XM_017027090.1 XM_017027091.1 XM_017027092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205549 c.1787C>T p.A596V Chr.19: 16402325-16402325 on Build GRCh38
6563397 c.1744C>T p.L582L Chr.19: 16402368-16402368 on Build GRCh38
7341272 c.1791+10G>A p.? Chr.19: 16402311-16402311 on Build GRCh38
6581318 c.1688A>C p.Q563P Chr.19: 16402424-16402424 on Build GRCh38
3284587 c.1683G>A p.L561L Chr.19: 16402429-16402429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560829867 Chr.19: 16402155-16402155 on Build GRCh38
rs189974243 Chr.19: 16402251-16402251 on Build GRCh38
rs557402959 Chr.19: 16402407-16402407 on Build GRCh38
rs971349872 Chr.19: 16402086-16402086 on Build GRCh38
rs150233580 Chr.19: 16402206-16402206 on Build GRCh38
rs1049692163 Chr.19: 16402056-16402056 on Build GRCh38
rs140189030 Chr.19: 16402357-16402357 on Build GRCh38
rs73021136 Chr.19: 16402502-16402502 on Build GRCh38
rs761516124 Chr.19: 16402377-16402377 on Build GRCh38
rs34999809 Chr.19: 16402093-16402094 on Build GRCh38
rs1020300831 Chr.19: 16402231-16402231 on Build GRCh38
rs202010883 Chr.19: 16402452-16402452 on Build GRCh38
rs927149705 Chr.19: 16402132-16402132 on Build GRCh38
rs759605386 Chr.19: 16402518-16402518 on Build GRCh38
rs772418516 Chr.19: 16402356-16402356 on Build GRCh38
rs751265283 Chr.19: 16402280-16402280 on Build GRCh38
rs756022019 Chr.19: 16402313-16402313 on Build GRCh38
rs548265747 Chr.19: 16402336-16402336 on Build GRCh38
rs186719562 Chr.19: 16402174-16402174 on Build GRCh38
rs753603960 Chr.19: 16402324-16402324 on Build GRCh38
rs370279365 Chr.19: 16402185-16402185 on Build GRCh38
rs537025989 Chr.19: 16402358-16402358 on Build GRCh38
rs765951004 Chr.19: 16402512-16402512 on Build GRCh38
rs775996027 Chr.19: 16402514-16402514 on Build GRCh38
rs543789262 Chr.19: 16402176-16402176 on Build GRCh38
rs765190445 Chr.19: 16402413-16402413 on Build GRCh38
rs891020928 Chr.19: 16402070-16402070 on Build GRCh38
rs771745548 Chr.19: 16402485-16402485 on Build GRCh38
rs760190116 Chr.19: 16402390-16402390 on Build GRCh38
rs776870350 Chr.19: 16402270-16402270 on Build GRCh38
rs376322635 Chr.19: 16402337-16402337 on Build GRCh38
rs779976858 Chr.19: 16402322-16402322 on Build GRCh38
rs893211234 Chr.19: 16402220-16402220 on Build GRCh38
rs984541875 Chr.19: 16402144-16402144 on Build GRCh38
rs932579777 Chr.19: 16402055-16402055 on Build GRCh38
rs887965022 Chr.19: 16402175-16402175 on Build GRCh38
rs140782885 Chr.19: 16402129-16402129 on Build GRCh38
rs757407541 Chr.19: 16402454-16402454 on Build GRCh38
rs1051746243 Chr.19: 16402188-16402189 on Build GRCh38
rs766247419 Chr.19: 16402397-16402397 on Build GRCh38
rs995251618 Chr.19: 16402161-16402161 on Build GRCh38
rs772605278 Chr.19: 16402381-16402381 on Build GRCh38
rs758079325 Chr.19: 16402484-16402484 on Build GRCh38
rs372940566 Chr.19: 16402304-16402304 on Build GRCh38
rs940387056 Chr.19: 16402148-16402148 on Build GRCh38
rs763762414 Chr.19: 16402278-16402278 on Build GRCh38
rs183328529 Chr.19: 16402510-16402510 on Build GRCh38
rs1020648748 Chr.19: 16402071-16402071 on Build GRCh38
rs528523953 Chr.19: 16402303-16402303 on Build GRCh38
rs918237894 Chr.19: 16402373-16402373 on Build GRCh38
rs889994307 Chr.19: 16402189-16402189 on Build GRCh38
rs137884743 Chr.19: 16402478-16402478 on Build GRCh38
rs770184097 Chr.19: 16402374-16402374 on Build GRCh38
rs746441140 Chr.19: 16402462-16402462 on Build GRCh38
rs138975721 Chr.19: 16402380-16402380 on Build GRCh38
rs182764320 Chr.19: 16402095-16402095 on Build GRCh38
rs937143889 Chr.19: 16402137-16402137 on Build GRCh38
rs576021536 Chr.19: 16402428-16402428 on Build GRCh38
rs770275949 Chr.19: 16402469-16402469 on Build GRCh38
rs781101518 Chr.19: 16402460-16402460 on Build GRCh38
rs925130258 Chr.19: 16402054-16402054 on Build GRCh38
rs777149194 Chr.19: 16402369-16402369 on Build GRCh38
rs992608584 Chr.19: 16402216-16402216 on Build GRCh38
rs544786872 Chr.19: 16402451-16402451 on Build GRCh38
rs958574370 Chr.19: 16402273-16402273 on Build GRCh38
rs758706169 Chr.19: 16402438-16402438 on Build GRCh38
rs896929888 Chr.19: 16402146-16402146 on Build GRCh38
rs1010295318 Chr.19: 16402228-16402228 on Build GRCh38
rs373743903 Chr.19: 16402422-16402422 on Build GRCh38
rs1033116658 Chr.19: 16402215-16402215 on Build GRCh38
rs1036696323 Chr.19: 16402117-16402117 on Build GRCh38
rs971762392 Chr.19: 16402234-16402234 on Build GRCh38
rs1013825112 Chr.19: 16402508-16402508 on Build GRCh38
rs1020947064 Chr.19: 16402250-16402250 on Build GRCh38
rs1054640715 Chr.19: 16402468-16402468 on Build GRCh38
rs1033900430 Chr.19: 16402198-16402198 on Build GRCh38
rs566042620 Chr.19: 16402284-16402284 on Build GRCh38
rs761427985 Chr.19: 16402296-16402296 on Build GRCh38
rs371006895 Chr.19: 16402342-16402342 on Build GRCh38
rs369310759 Chr.19: 16402272-16402272 on Build GRCh38
rs150310027 Chr.19: 16402408-16402408 on Build GRCh38
rs932488088 Chr.19: 16402285-16402285 on Build GRCh38
rs771374210 Chr.19: 16402363-16402363 on Build GRCh38

View Full Product Details