Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40348022-40348516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf47
Entrez Gene ID
126526
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
-
UniGene
Hs.631557
Gene Chromosome Location
Chr.19: 40292743-40348527 on Build GRCh38
Transcripts
NM_001256440.1 NM_001256441.1 NR_046202.1 XM_005258520.2 XM_011526460.2 XM_017026291.1 XM_017026293.1 XM_017026295.1 NM_001031696.3 NM_012268.3 XM_017026546.1 XM_017026549.1

Gene Symbol
PLD3
Entrez Gene ID
23646
Gene Name
phospholipase D family member 3
Gene Aliases
AD19, HU-K4, HUK4
UniGene
Hs.257008
Gene Chromosome Location
Chr.19: 40348395-40378490 on Build GRCh38
Transcripts
NM_001256440.1 NM_001256441.1 NR_046202.1 XM_005258520.2 XM_011526460.2 XM_017026291.1 XM_017026293.1 XM_017026295.1 NM_001031696.3 NM_012268.3 XM_017026546.1 XM_017026549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5762630 c.2T>C p.M1T Chr.19: 40348513-40348513 on Build GRCh38
6380270 c.60G>T p.E20D Chr.19: 40348455-40348455 on Build GRCh38
4416438 c.1A>C p.M1L Chr.19: 40348514-40348514 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1034712276 Chr.19: 40348399-40348399 on Build GRCh38
rs946422456 Chr.19: 40348352-40348352 on Build GRCh38
rs1004179274 Chr.19: 40348441-40348441 on Build GRCh38
rs899690498 Chr.19: 40348409-40348409 on Build GRCh38
rs370249381 Chr.19: 40348407-40348407 on Build GRCh38
rs544955999 Chr.19: 40348402-40348402 on Build GRCh38
rs920796759 Chr.19: 40348194-40348194 on Build GRCh38
rs899745042 Chr.19: 40348186-40348186 on Build GRCh38
rs1003222594 Chr.19: 40348393-40348393 on Build GRCh38
rs924375010 Chr.19: 40348384-40348384 on Build GRCh38
rs1035288588 Chr.19: 40348162-40348162 on Build GRCh38
rs996650283 Chr.19: 40348072-40348072 on Build GRCh38
rs959498409 Chr.19: 40348163-40348163 on Build GRCh38
rs760010676 Chr.19: 40348425-40348425 on Build GRCh38
rs375524715 Chr.19: 40348090-40348090 on Build GRCh38
rs187325420 Chr.19: 40348136-40348136 on Build GRCh38
rs78044607 Chr.19: 40348285-40348285 on Build GRCh38
rs578237018 Chr.19: 40348484-40348484 on Build GRCh38
rs959825141 Chr.19: 40348482-40348482 on Build GRCh38
rs952219662 Chr.19: 40348218-40348219 on Build GRCh38
rs1048319844 Chr.19: 40348432-40348432 on Build GRCh38
rs1019899033 Chr.19: 40348178-40348178 on Build GRCh38
rs749592252 Chr.19: 40348416-40348416 on Build GRCh38
rs549528292 Chr.19: 40348137-40348137 on Build GRCh38
rs1003756439 Chr.19: 40348150-40348150 on Build GRCh38
rs983563406 Chr.19: 40348237-40348237 on Build GRCh38
rs893672238 Chr.19: 40348331-40348331 on Build GRCh38
rs952249470 Chr.19: 40348462-40348462 on Build GRCh38
rs938298059 Chr.19: 40348056-40348056 on Build GRCh38
rs192815715 Chr.19: 40348252-40348252 on Build GRCh38
rs539898169 Chr.19: 40348222-40348222 on Build GRCh38
rs914217668 Chr.19: 40348317-40348317 on Build GRCh38
rs888410489 Chr.19: 40348438-40348438 on Build GRCh38
rs915503217 Chr.19: 40348495-40348495 on Build GRCh38
rs764866409 Chr.19: 40348468-40348468 on Build GRCh38
rs943822464 Chr.19: 40348403-40348403 on Build GRCh38
rs1040863047 Chr.19: 40348408-40348408 on Build GRCh38
rs558011897 Chr.19: 40348241-40348241 on Build GRCh38
rs893531107 Chr.19: 40348092-40348092 on Build GRCh38
rs769966141 Chr.19: 40348406-40348406 on Build GRCh38
rs571141092 Chr.19: 40348200-40348200 on Build GRCh38
rs1041452799 Chr.19: 40348338-40348338 on Build GRCh38
rs773841513 Chr.19: 40348188-40348188 on Build GRCh38
rs892830682 Chr.19: 40348087-40348087 on Build GRCh38
rs1041505661 Chr.19: 40348134-40348134 on Build GRCh38
rs768088100 Chr.19: 40348448-40348448 on Build GRCh38
rs1008068509 Chr.19: 40348173-40348173 on Build GRCh38
rs947037829 Chr.19: 40348327-40348327 on Build GRCh38
rs771069658 Chr.19: 40348418-40348418 on Build GRCh38
rs926824991 Chr.19: 40348264-40348264 on Build GRCh38
rs1049550069 Chr.19: 40348302-40348302 on Build GRCh38
rs761194118 Chr.19: 40348464-40348464 on Build GRCh38
rs952730105 Chr.19: 40348242-40348242 on Build GRCh38
rs10415745 Chr.19: 40348253-40348253 on Build GRCh38
rs761276738 Chr.19: 40348284-40348284 on Build GRCh38
rs557727596 Chr.19: 40348387-40348387 on Build GRCh38
rs982296137 Chr.19: 40348254-40348254 on Build GRCh38
rs939898905 Chr.19: 40348051-40348056 on Build GRCh38
rs1026927786 Chr.19: 40348195-40348195 on Build GRCh38
rs138513107 Chr.19: 40348245-40348245 on Build GRCh38
rs767990589 Chr.19: 40348071-40348071 on Build GRCh38
rs1004060208 Chr.19: 40348461-40348461 on Build GRCh38
rs1010697887 Chr.19: 40348117-40348117 on Build GRCh38
rs1042139906 Chr.19: 40348370-40348370 on Build GRCh38
rs776184936 Chr.19: 40348459-40348459 on Build GRCh38
rs10414431 Chr.19: 40348424-40348424 on Build GRCh38

View Full Product Details