Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40704908-40705398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ8B
Entrez Gene ID
79934
Gene Name
coenzyme Q8B
Gene Aliases
ADCK4, NPHS9
UniGene
Hs.130712
Gene Chromosome Location
Chr.19: 40691529-40716885 on Build GRCh38
Transcripts
NM_001142555.2 NM_024876.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5938415 c.422G>A p.R141Q Chr.19: 40705393-40705393 on Build GRCh38
5577969 c.490G>A p.D164N Chr.19: 40705325-40705325 on Build GRCh38
148633 c.501C>A p.F167L Chr.19: 40705171-40705171 on Build GRCh38
5564773 c.532C>T p.R178W Chr.19: 40705140-40705140 on Build GRCh38
148631 c.521A>G p.H174R Chr.19: 40705151-40705151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3865452 Chr.19: 40705151-40705151 on Build GRCh38
rs887025090 Chr.19: 40705230-40705230 on Build GRCh38
rs745401575 Chr.19: 40705055-40705055 on Build GRCh38
rs376835091 Chr.19: 40705352-40705352 on Build GRCh38
rs757869483 Chr.19: 40705234-40705234 on Build GRCh38
rs758303807 Chr.19: 40705110-40705110 on Build GRCh38
rs776487329 Chr.19: 40705074-40705074 on Build GRCh38
rs754329107 Chr.19: 40705109-40705109 on Build GRCh38
rs778169057 Chr.19: 40705133-40705133 on Build GRCh38
rs147603227 Chr.19: 40705152-40705152 on Build GRCh38
rs760103716 Chr.19: 40705125-40705125 on Build GRCh38
rs771548569 Chr.19: 40705139-40705139 on Build GRCh38
rs190206283 Chr.19: 40705215-40705215 on Build GRCh38
rs141658497 Chr.19: 40705359-40705359 on Build GRCh38
rs987870810 Chr.19: 40705296-40705296 on Build GRCh38
rs59062602 Chr.19: 40705227-40705227 on Build GRCh38
rs142234074 Chr.19: 40705117-40705117 on Build GRCh38
rs547302540 Chr.19: 40705321-40705321 on Build GRCh38
rs763831232 Chr.19: 40705073-40705073 on Build GRCh38
rs374201690 Chr.19: 40705082-40705082 on Build GRCh38
rs753361236 Chr.19: 40705301-40705301 on Build GRCh38
rs765245063 Chr.19: 40705095-40705095 on Build GRCh38
rs781206157 Chr.19: 40705123-40705123 on Build GRCh38
rs780725788 Chr.19: 40705278-40705278 on Build GRCh38
rs954952320 Chr.19: 40705357-40705357 on Build GRCh38
rs545319604 Chr.19: 40705189-40705189 on Build GRCh38
rs755262527 Chr.19: 40705119-40705119 on Build GRCh38
rs758921688 Chr.19: 40705374-40705374 on Build GRCh38
rs979101848 Chr.19: 40705068-40705068 on Build GRCh38
rs1002678520 Chr.19: 40705242-40705242 on Build GRCh38
rs200697107 Chr.19: 40705216-40705216 on Build GRCh38
rs371816247 Chr.19: 40705229-40705229 on Build GRCh38
rs145416274 Chr.19: 40705372-40705372 on Build GRCh38
rs766495150 Chr.19: 40705113-40705113 on Build GRCh38
rs754265164 Chr.19: 40705364-40705364 on Build GRCh38
rs970596859 Chr.19: 40705327-40705327 on Build GRCh38
rs995195939 Chr.19: 40705222-40705222 on Build GRCh38
rs757644020 Chr.19: 40705366-40705366 on Build GRCh38
rs775738775 Chr.19: 40705017-40705017 on Build GRCh38
rs755774439 Chr.19: 40705201-40705201 on Build GRCh38
rs534263552 Chr.19: 40704929-40704929 on Build GRCh38
rs774230786 Chr.19: 40705320-40705320 on Build GRCh38
rs760868849 Chr.19: 40705358-40705358 on Build GRCh38
rs201047027 Chr.19: 40705205-40705205 on Build GRCh38
rs776100391 Chr.19: 40705158-40705158 on Build GRCh38
rs113398730 Chr.19: 40705024-40705024 on Build GRCh38
rs750292169 Chr.19: 40705102-40705102 on Build GRCh38
rs372213778 Chr.19: 40705199-40705199 on Build GRCh38
rs1052589088 Chr.19: 40704948-40704948 on Build GRCh38
rs759562049 Chr.19: 40705202-40705202 on Build GRCh38
rs1006472560 Chr.19: 40705054-40705054 on Build GRCh38
rs1032443173 Chr.19: 40705246-40705246 on Build GRCh38
rs56096945 Chr.19: 40705183-40705183 on Build GRCh38
rs1051877729 Chr.19: 40705166-40705166 on Build GRCh38
rs372379297 Chr.19: 40705066-40705066 on Build GRCh38
rs747808667 Chr.19: 40705280-40705280 on Build GRCh38
rs770870600 Chr.19: 40705311-40705311 on Build GRCh38
rs988771793 Chr.19: 40705145-40705145 on Build GRCh38
rs866812916 Chr.19: 40705134-40705134 on Build GRCh38
rs151217582 Chr.19: 40705126-40705126 on Build GRCh38
rs369387219 Chr.19: 40705174-40705174 on Build GRCh38
rs773038734 Chr.19: 40705180-40705180 on Build GRCh38
rs71358927 Chr.19: 40705001-40705001 on Build GRCh38
rs139167042 Chr.19: 40705138-40705138 on Build GRCh38
rs766156953 Chr.19: 40705277-40705278 on Build GRCh38
rs79156288 Chr.19: 40705237-40705237 on Build GRCh38
rs913944281 Chr.19: 40705334-40705334 on Build GRCh38
rs762936896 Chr.19: 40705186-40705186 on Build GRCh38
rs769474622 Chr.19: 40705307-40705307 on Build GRCh38
rs867772820 Chr.19: 40705135-40705135 on Build GRCh38
rs958246266 Chr.19: 40705268-40705268 on Build GRCh38
rs55793080 Chr.19: 40705171-40705171 on Build GRCh38
rs771747054 Chr.19: 40705058-40705058 on Build GRCh38
rs773336989 Chr.19: 40704952-40704952 on Build GRCh38
rs751645641 Chr.19: 40705200-40705200 on Build GRCh38
rs754578125 Chr.19: 40705324-40705324 on Build GRCh38
rs377527612 Chr.19: 40705232-40705232 on Build GRCh38
rs751178358 Chr.19: 40705308-40705308 on Build GRCh38
rs549359403 Chr.19: 40705236-40705236 on Build GRCh38
rs751011521 Chr.19: 40705373-40705373 on Build GRCh38
rs147251838 Chr.19: 40705062-40705062 on Build GRCh38
rs767810360 Chr.19: 40705203-40705203 on Build GRCh38
rs751876051 Chr.19: 40705094-40705094 on Build GRCh38
rs555867503 Chr.19: 40705016-40705016 on Build GRCh38
rs398122978 Chr.19: 40705140-40705140 on Build GRCh38
rs767542746 Chr.19: 40705325-40705325 on Build GRCh38
rs768013882 Chr.19: 40705266-40705266 on Build GRCh38
rs532418960 Chr.19: 40705309-40705309 on Build GRCh38

View Full Product Details