Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26181650-26182193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
NM_001168241.1 NM_001191033.1 XM_006711951.3 XM_011532564.2 XM_011532565.2 XM_011532566.2 XM_011532567.2

SNP Detail

SNP ID Chromosome Location
rs542076774 Chr.2: 26181823-26181823 on Build GRCh38
rs1046993793 Chr.2: 26181690-26181690 on Build GRCh38
rs768388268 Chr.2: 26182125-26182125 on Build GRCh38
rs989470012 Chr.2: 26181747-26181747 on Build GRCh38
rs571451697 Chr.2: 26181922-26181922 on Build GRCh38
rs190800549 Chr.2: 26181889-26181889 on Build GRCh38
rs990319445 Chr.2: 26181839-26181839 on Build GRCh38
rs145825451 Chr.2: 26181792-26181792 on Build GRCh38
rs918423819 Chr.2: 26181824-26181824 on Build GRCh38
rs185989656 Chr.2: 26181763-26181763 on Build GRCh38
rs183942478 Chr.2: 26182099-26182099 on Build GRCh38
rs565781368 Chr.2: 26182020-26182020 on Build GRCh38
rs1030102581 Chr.2: 26181758-26181758 on Build GRCh38
rs2384360 Chr.2: 26182147-26182147 on Build GRCh38
rs957497334 Chr.2: 26181831-26181831 on Build GRCh38
rs150233700 Chr.2: 26181749-26181749 on Build GRCh38
rs35042519 Chr.2: 26181953-26181954 on Build GRCh38
rs968985184 Chr.2: 26182000-26182000 on Build GRCh38
rs998873492 Chr.2: 26182050-26182050 on Build GRCh38
rs572134764 Chr.2: 26181692-26181692 on Build GRCh38
rs1034785927 Chr.2: 26182165-26182165 on Build GRCh38
rs532582887 Chr.2: 26181794-26181794 on Build GRCh38
rs1005394968 Chr.2: 26181707-26181707 on Build GRCh38
rs893424507 Chr.2: 26182098-26182098 on Build GRCh38
rs980445082 Chr.2: 26181869-26181869 on Build GRCh38
rs908461359 Chr.2: 26181913-26181913 on Build GRCh38
rs181345433 Chr.2: 26181861-26181861 on Build GRCh38
rs932749581 Chr.2: 26181996-26181996 on Build GRCh38
rs559614794 Chr.2: 26181813-26181813 on Build GRCh38
rs1022318489 Chr.2: 26181977-26181977 on Build GRCh38
rs368791016 Chr.2: 26181919-26181919 on Build GRCh38
rs1007069455 Chr.2: 26182010-26182010 on Build GRCh38
rs542663790 Chr.2: 26181705-26181705 on Build GRCh38
rs554567133 Chr.2: 26181710-26181710 on Build GRCh38
rs541897549 Chr.2: 26181806-26181806 on Build GRCh38
rs538468577 Chr.2: 26181957-26181957 on Build GRCh38
rs1035737715 Chr.2: 26181910-26181910 on Build GRCh38
rs899956424 Chr.2: 26181746-26181746 on Build GRCh38
rs749047704 Chr.2: 26182022-26182022 on Build GRCh38
rs968829847 Chr.2: 26181770-26181770 on Build GRCh38
rs554653784 Chr.2: 26182106-26182106 on Build GRCh38
rs1038243931 Chr.2: 26181958-26181958 on Build GRCh38
rs976480665 Chr.2: 26182030-26182030 on Build GRCh38
rs1023115359 Chr.2: 26182111-26182111 on Build GRCh38
rs1038422735 Chr.2: 26181732-26181732 on Build GRCh38
rs186041737 Chr.2: 26181867-26181867 on Build GRCh38
rs1031883090 Chr.2: 26182055-26182055 on Build GRCh38
rs1047389059 Chr.2: 26181909-26181909 on Build GRCh38
rs4665832 Chr.2: 26181906-26181906 on Build GRCh38
rs148946831 Chr.2: 26181965-26181965 on Build GRCh38
rs541580505 Chr.2: 26181805-26181805 on Build GRCh38
rs1048823380 Chr.2: 26181846-26181846 on Build GRCh38
rs373245533 Chr.2: 26181998-26181998 on Build GRCh38
rs190011995 Chr.2: 26181853-26181853 on Build GRCh38
rs751644470 Chr.2: 26181706-26181706 on Build GRCh38
rs1031934793 Chr.2: 26181821-26181821 on Build GRCh38

View Full Product Details