Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26186083-26186578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
NM_001168241.1 NM_001191033.1 XM_006711951.3 XM_011532564.2 XM_011532565.2 XM_011532566.2 XM_011532567.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407176 c.1576T>A p.Y526N Chr.2: 26186336-26186336 on Build GRCh38
1407174 c.1525C>T p.R509C Chr.2: 26186285-26186285 on Build GRCh38
1407175 c.1567C>A p.L523I Chr.2: 26186327-26186327 on Build GRCh38
5745877 c.1530delC p.K512fs*80 Chr.2: 26186290-26186290 on Build GRCh38
5708167 c.1526G>A p.R509H Chr.2: 26186286-26186286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750870583 Chr.2: 26186270-26186270 on Build GRCh38
rs935543557 Chr.2: 26186287-26186287 on Build GRCh38
rs533418270 Chr.2: 26186532-26186532 on Build GRCh38
rs1014698987 Chr.2: 26186469-26186469 on Build GRCh38
rs961941883 Chr.2: 26186170-26186170 on Build GRCh38
rs773159472 Chr.2: 26186193-26186195 on Build GRCh38
rs776701567 Chr.2: 26186391-26186391 on Build GRCh38
rs757117763 Chr.2: 26186209-26186209 on Build GRCh38
rs973213505 Chr.2: 26186251-26186251 on Build GRCh38
rs754300310 Chr.2: 26186252-26186252 on Build GRCh38
rs984674206 Chr.2: 26186484-26186484 on Build GRCh38
rs991439161 Chr.2: 26186552-26186552 on Build GRCh38
rs749166184 Chr.2: 26186370-26186370 on Build GRCh38
rs958488578 Chr.2: 26186496-26186496 on Build GRCh38
rs780906649 Chr.2: 26186329-26186329 on Build GRCh38
rs760790718 Chr.2: 26186234-26186234 on Build GRCh38
rs1015078574 Chr.2: 26186148-26186148 on Build GRCh38
rs930469363 Chr.2: 26186238-26186238 on Build GRCh38
rs372552496 Chr.2: 26186256-26186256 on Build GRCh38
rs762156948 Chr.2: 26186405-26186405 on Build GRCh38
rs757977775 Chr.2: 26186274-26186274 on Build GRCh38
rs1023952429 Chr.2: 26186404-26186404 on Build GRCh38
rs890160549 Chr.2: 26186451-26186451 on Build GRCh38
rs550281953 Chr.2: 26186350-26186350 on Build GRCh38
rs971939262 Chr.2: 26186442-26186442 on Build GRCh38
rs971062966 Chr.2: 26186424-26186424 on Build GRCh38
rs889244412 Chr.2: 26186255-26186255 on Build GRCh38
rs748040879 Chr.2: 26186346-26186346 on Build GRCh38
rs557237676 Chr.2: 26186109-26186109 on Build GRCh38
rs1008975599 Chr.2: 26186466-26186466 on Build GRCh38
rs1053896250 Chr.2: 26186313-26186313 on Build GRCh38
rs549745957 Chr.2: 26186492-26186492 on Build GRCh38
rs764552146 Chr.2: 26186236-26186236 on Build GRCh38
rs754500051 Chr.2: 26186310-26186310 on Build GRCh38
rs982258521 Chr.2: 26186276-26186276 on Build GRCh38
rs760915270 Chr.2: 26186379-26186379 on Build GRCh38
rs745366691 Chr.2: 26186375-26186375 on Build GRCh38
rs780873920 Chr.2: 26186323-26186323 on Build GRCh38
rs769776103 Chr.2: 26186353-26186353 on Build GRCh38
rs759610810 Chr.2: 26186191-26186191 on Build GRCh38
rs767845675 Chr.2: 26186214-26186214 on Build GRCh38
rs561382152 Chr.2: 26186286-26186286 on Build GRCh38
rs528876581 Chr.2: 26186302-26186302 on Build GRCh38
rs148790250 Chr.2: 26186105-26186105 on Build GRCh38
rs775728982 Chr.2: 26186376-26186376 on Build GRCh38
rs545979295 Chr.2: 26186174-26186174 on Build GRCh38
rs570859185 Chr.2: 26186532-26186533 on Build GRCh38
rs564083150 Chr.2: 26186222-26186222 on Build GRCh38
rs917143931 Chr.2: 26186144-26186144 on Build GRCh38
rs1050600561 Chr.2: 26186421-26186422 on Build GRCh38
rs768670941 Chr.2: 26186380-26186380 on Build GRCh38
rs951855424 Chr.2: 26186476-26186476 on Build GRCh38
rs769570232 Chr.2: 26186473-26186473 on Build GRCh38
rs777424902 Chr.2: 26186365-26186365 on Build GRCh38
rs893954675 Chr.2: 26186338-26186340 on Build GRCh38
rs142467198 Chr.2: 26186113-26186113 on Build GRCh38
rs1021171437 Chr.2: 26186281-26186281 on Build GRCh38
rs1040404678 Chr.2: 26186266-26186266 on Build GRCh38

View Full Product Details