Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31260481-31260986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD3
Entrez Gene ID
30845
Gene Name
EH domain containing 3
Gene Aliases
PAST3
UniGene
Hs.368808
Gene Chromosome Location
Chr.2: 31234014-31269451 on Build GRCh38
Transcripts
NM_014600.2 XM_011532806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6509235 c.692G>A p.R231Q Chr.2: 31260699-31260699 on Build GRCh38
3581080 c.579A>T p.K193N Chr.2: 31260586-31260586 on Build GRCh38
1205077 c.860G>A p.R287Q Chr.2: 31260867-31260867 on Build GRCh38
4606804 c.802C>T p.R268W Chr.2: 31260809-31260809 on Build GRCh38
3581082 c.673G>A p.E225K Chr.2: 31260680-31260680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757561609 Chr.2: 31260679-31260679 on Build GRCh38
rs765621773 Chr.2: 31260716-31260716 on Build GRCh38
rs931682708 Chr.2: 31260935-31260935 on Build GRCh38
rs976025920 Chr.2: 31260872-31260872 on Build GRCh38
rs754381951 Chr.2: 31260558-31260558 on Build GRCh38
rs967872901 Chr.2: 31260823-31260823 on Build GRCh38
rs372378751 Chr.2: 31260958-31260958 on Build GRCh38
rs900182342 Chr.2: 31260580-31260580 on Build GRCh38
rs147844646 Chr.2: 31260706-31260706 on Build GRCh38
rs755285979 Chr.2: 31260693-31260693 on Build GRCh38
rs767114443 Chr.2: 31260661-31260661 on Build GRCh38
rs746464253 Chr.2: 31260965-31260965 on Build GRCh38
rs371507212 Chr.2: 31260587-31260587 on Build GRCh38
rs550566065 Chr.2: 31260738-31260738 on Build GRCh38
rs199750636 Chr.2: 31260892-31260892 on Build GRCh38
rs768625383 Chr.2: 31260631-31260631 on Build GRCh38
rs565201197 Chr.2: 31260853-31260853 on Build GRCh38
rs781388986 Chr.2: 31260826-31260827 on Build GRCh38
rs143059948 Chr.2: 31260866-31260866 on Build GRCh38
rs111364798 Chr.2: 31260692-31260692 on Build GRCh38
rs773077288 Chr.2: 31260700-31260700 on Build GRCh38
rs1048803980 Chr.2: 31260651-31260651 on Build GRCh38
rs776537856 Chr.2: 31260636-31260636 on Build GRCh38
rs879115769 Chr.2: 31260783-31260783 on Build GRCh38
rs774214784 Chr.2: 31260788-31260788 on Build GRCh38
rs200795713 Chr.2: 31260685-31260685 on Build GRCh38
rs766994253 Chr.2: 31260882-31260882 on Build GRCh38
rs758911736 Chr.2: 31260736-31260736 on Build GRCh38
rs765669139 Chr.2: 31260680-31260680 on Build GRCh38
rs200458492 Chr.2: 31260936-31260936 on Build GRCh38
rs977945348 Chr.2: 31260833-31260833 on Build GRCh38
rs567062495 Chr.2: 31260504-31260504 on Build GRCh38
rs376729413 Chr.2: 31260787-31260787 on Build GRCh38
rs763181531 Chr.2: 31260867-31260867 on Build GRCh38
rs747225097 Chr.2: 31260623-31260623 on Build GRCh38
rs777592016 Chr.2: 31260761-31260761 on Build GRCh38
rs921560139 Chr.2: 31260899-31260899 on Build GRCh38
rs557753544 Chr.2: 31260637-31260637 on Build GRCh38
rs575875306 Chr.2: 31260549-31260549 on Build GRCh38
rs138007410 Chr.2: 31260770-31260770 on Build GRCh38
rs770386941 Chr.2: 31260707-31260707 on Build GRCh38
rs751219079 Chr.2: 31260650-31260650 on Build GRCh38
rs540341129 Chr.2: 31260676-31260676 on Build GRCh38
rs776880592 Chr.2: 31260649-31260649 on Build GRCh38
rs753006287 Chr.2: 31260922-31260922 on Build GRCh38
rs200027155 Chr.2: 31260505-31260505 on Build GRCh38
rs112609329 Chr.2: 31260652-31260652 on Build GRCh38
rs771567803 Chr.2: 31260809-31260809 on Build GRCh38
rs146929365 Chr.2: 31260715-31260715 on Build GRCh38
rs201814896 Chr.2: 31260684-31260684 on Build GRCh38
rs753679668 Chr.2: 31260943-31260943 on Build GRCh38
rs959364349 Chr.2: 31260748-31260748 on Build GRCh38
rs745950962 Chr.2: 31260589-31260589 on Build GRCh38
rs778088572 Chr.2: 31260557-31260557 on Build GRCh38
rs781636692 Chr.2: 31260713-31260713 on Build GRCh38
rs369917897 Chr.2: 31260950-31260950 on Build GRCh38
rs562001448 Chr.2: 31260697-31260697 on Build GRCh38
rs369424115 Chr.2: 31260658-31260658 on Build GRCh38
rs750945058 Chr.2: 31260734-31260734 on Build GRCh38
rs767679000 Chr.2: 31260909-31260909 on Build GRCh38
rs745793875 Chr.2: 31260953-31260953 on Build GRCh38
rs146103643 Chr.2: 31260610-31260610 on Build GRCh38
rs375784165 Chr.2: 31260600-31260600 on Build GRCh38
rs756935908 Chr.2: 31260948-31260948 on Build GRCh38
rs772124528 Chr.2: 31260599-31260599 on Build GRCh38
rs766758949 Chr.2: 31260737-31260737 on Build GRCh38
rs758996076 Chr.2: 31260712-31260712 on Build GRCh38
rs180893282 Chr.2: 31260698-31260698 on Build GRCh38
rs764883628 Chr.2: 31260526-31260526 on Build GRCh38
rs753016337 Chr.2: 31260830-31260831 on Build GRCh38
rs745387793 Chr.2: 31260793-31260793 on Build GRCh38
rs760353877 Chr.2: 31260846-31260846 on Build GRCh38
rs139061856 Chr.2: 31260865-31260865 on Build GRCh38
rs755907641 Chr.2: 31260759-31260759 on Build GRCh38
rs779511192 Chr.2: 31260961-31260961 on Build GRCh38
rs113087929 Chr.2: 31260699-31260699 on Build GRCh38
rs139542048 Chr.2: 31260522-31260522 on Build GRCh38
rs775181095 Chr.2: 31260810-31260810 on Build GRCh38
rs373688842 Chr.2: 31260758-31260758 on Build GRCh38
rs962040379 Chr.2: 31260677-31260677 on Build GRCh38
rs777809819 Chr.2: 31260938-31260938 on Build GRCh38
rs765564503 Chr.2: 31260856-31260856 on Build GRCh38
rs773132739 Chr.2: 31260640-31260640 on Build GRCh38
rs769628934 Chr.2: 31260638-31260638 on Build GRCh38
rs556019532 Chr.2: 31260544-31260544 on Build GRCh38
rs369599160 Chr.2: 31260799-31260799 on Build GRCh38
rs749920764 Chr.2: 31260527-31260527 on Build GRCh38
rs200471807 Chr.2: 31260757-31260757 on Build GRCh38
rs773880032 Chr.2: 31260710-31260710 on Build GRCh38
rs1057418700 Chr.2: 31260532-31260532 on Build GRCh38
rs780163743 Chr.2: 31260691-31260691 on Build GRCh38
rs537831838 Chr.2: 31260574-31260574 on Build GRCh38
rs762432989 Chr.2: 31260648-31260648 on Build GRCh38
rs146384685 Chr.2: 31260511-31260511 on Build GRCh38

View Full Product Details