Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50586801-50587304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528585.2 XM_017027681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4137042 c.2776A>G p.K926E Chr.20: 50587297-50587297 on Build GRCh38
5434692 c.2786C>T p.S929L Chr.20: 50587287-50587287 on Build GRCh38
5031250 c.2810A>T p.Q937L Chr.20: 50587263-50587263 on Build GRCh38
1206411 c.2811G>T p.Q937H Chr.20: 50587262-50587262 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005924655 Chr.20: 50586837-50586837 on Build GRCh38
rs948787625 Chr.20: 50586994-50586994 on Build GRCh38
rs988071536 Chr.20: 50586876-50586876 on Build GRCh38
rs761339265 Chr.20: 50587240-50587240 on Build GRCh38
rs190116407 Chr.20: 50586990-50586990 on Build GRCh38
rs987010645 Chr.20: 50586954-50586954 on Build GRCh38
rs914526336 Chr.20: 50587256-50587256 on Build GRCh38
rs762198709 Chr.20: 50587185-50587185 on Build GRCh38
rs1022545846 Chr.20: 50587132-50587132 on Build GRCh38
rs948753664 Chr.20: 50587281-50587281 on Build GRCh38
rs888740746 Chr.20: 50587022-50587022 on Build GRCh38
rs143373465 Chr.20: 50587206-50587206 on Build GRCh38
rs144300885 Chr.20: 50587252-50587252 on Build GRCh38
rs559319731 Chr.20: 50587216-50587216 on Build GRCh38
rs536016005 Chr.20: 50586836-50586836 on Build GRCh38
rs1047197657 Chr.20: 50586997-50586997 on Build GRCh38
rs751702492 Chr.20: 50587188-50587188 on Build GRCh38
rs554095693 Chr.20: 50586839-50586839 on Build GRCh38
rs778650387 Chr.20: 50587199-50587199 on Build GRCh38
rs3746564 Chr.20: 50586843-50586843 on Build GRCh38
rs576071435 Chr.20: 50587095-50587095 on Build GRCh38
rs772613254 Chr.20: 50587142-50587142 on Build GRCh38
rs766487640 Chr.20: 50587228-50587228 on Build GRCh38
rs773696459 Chr.20: 50587250-50587250 on Build GRCh38
rs369257375 Chr.20: 50587180-50587180 on Build GRCh38
rs955569587 Chr.20: 50586921-50586921 on Build GRCh38
rs927280415 Chr.20: 50586945-50586945 on Build GRCh38
rs1047671120 Chr.20: 50587123-50587123 on Build GRCh38
rs761083911 Chr.20: 50587227-50587228 on Build GRCh38
rs997951964 Chr.20: 50587068-50587068 on Build GRCh38
rs554387478 Chr.20: 50587198-50587198 on Build GRCh38
rs770347051 Chr.20: 50587244-50587244 on Build GRCh38
rs767741112 Chr.20: 50587189-50587190 on Build GRCh38
rs886385628 Chr.20: 50587138-50587138 on Build GRCh38
rs80119536 Chr.20: 50587104-50587104 on Build GRCh38
rs941598085 Chr.20: 50587026-50587026 on Build GRCh38
rs924897575 Chr.20: 50586965-50586965 on Build GRCh38
rs546120876 Chr.20: 50587079-50587079 on Build GRCh38
rs921398313 Chr.20: 50586894-50586894 on Build GRCh38
rs745875659 Chr.20: 50587202-50587202 on Build GRCh38
rs1009427774 Chr.20: 50587098-50587098 on Build GRCh38
rs759685628 Chr.20: 50587276-50587276 on Build GRCh38
rs1022241268 Chr.20: 50586878-50586881 on Build GRCh38
rs139941208 Chr.20: 50587088-50587088 on Build GRCh38
rs567262497 Chr.20: 50586824-50586824 on Build GRCh38
rs752073838 Chr.20: 50587259-50587259 on Build GRCh38
rs776391362 Chr.20: 50587236-50587236 on Build GRCh38
rs34633644 Chr.20: 50587014-50587015 on Build GRCh38
rs767999200 Chr.20: 50587234-50587234 on Build GRCh38
rs956501971 Chr.20: 50586865-50586865 on Build GRCh38
rs746536957 Chr.20: 50587218-50587218 on Build GRCh38
rs77193043 Chr.20: 50587242-50587242 on Build GRCh38
rs1012925088 Chr.20: 50586994-50587009 on Build GRCh38
rs950361002 Chr.20: 50586825-50586825 on Build GRCh38
rs750986720 Chr.20: 50587190-50587194 on Build GRCh38
rs906993778 Chr.20: 50587069-50587069 on Build GRCh38
rs779914064 Chr.20: 50587211-50587211 on Build GRCh38
rs899456628 Chr.20: 50587052-50587052 on Build GRCh38
rs1039379039 Chr.20: 50587031-50587031 on Build GRCh38
rs777912939 Chr.20: 50586969-50586969 on Build GRCh38
rs781508356 Chr.20: 50587193-50587193 on Build GRCh38
rs964415608 Chr.20: 50587151-50587151 on Build GRCh38
rs937452261 Chr.20: 50586967-50586967 on Build GRCh38
rs1002498739 Chr.20: 50587155-50587155 on Build GRCh38
rs551747121 Chr.20: 50587254-50587254 on Build GRCh38
rs770930097 Chr.20: 50586991-50586991 on Build GRCh38
rs75292178 Chr.20: 50587148-50587148 on Build GRCh38
rs974531577 Chr.20: 50586851-50586851 on Build GRCh38
rs774441526 Chr.20: 50587005-50587005 on Build GRCh38
rs759983461 Chr.20: 50587077-50587077 on Build GRCh38
rs964842818 Chr.20: 50586842-50586842 on Build GRCh38
rs267605990 Chr.20: 50587278-50587278 on Build GRCh38
rs754946187 Chr.20: 50587192-50587192 on Build GRCh38
rs992887300 Chr.20: 50587194-50587195 on Build GRCh38

View Full Product Details