Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50587700-50588198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528585.2 XM_017027681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3547625 c.2708G>A p.R903Q Chr.20: 50587834-50587834 on Build GRCh38
3770843 c.2660G>C p.S887T Chr.20: 50587882-50587882 on Build GRCh38
5858104 c.2722G>A p.E908K Chr.20: 50587820-50587820 on Build GRCh38
6895221 c.2665G>C p.D889H Chr.20: 50587877-50587877 on Build GRCh38
5546752 c.2713_2716delGCAG p.A905fs*14 Chr.20: 50587826-50587829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778465066 Chr.20: 50587944-50587944 on Build GRCh38
rs953336317 Chr.20: 50588101-50588101 on Build GRCh38
rs866388755 Chr.20: 50587744-50587744 on Build GRCh38
rs1001761549 Chr.20: 50587949-50587949 on Build GRCh38
rs373645112 Chr.20: 50587822-50587822 on Build GRCh38
rs368478054 Chr.20: 50587858-50587858 on Build GRCh38
rs368176656 Chr.20: 50587878-50587878 on Build GRCh38
rs979540302 Chr.20: 50588099-50588100 on Build GRCh38
rs147744114 Chr.20: 50588080-50588080 on Build GRCh38
rs751353711 Chr.20: 50587915-50587915 on Build GRCh38
rs35841086 Chr.20: 50587941-50587942 on Build GRCh38
rs1028499526 Chr.20: 50588081-50588081 on Build GRCh38
rs921751589 Chr.20: 50588085-50588085 on Build GRCh38
rs761820920 Chr.20: 50587879-50587879 on Build GRCh38
rs200228661 Chr.20: 50587830-50587830 on Build GRCh38
rs748927760 Chr.20: 50587782-50587782 on Build GRCh38
rs753204761 Chr.20: 50588079-50588079 on Build GRCh38
rs1004645360 Chr.20: 50588059-50588059 on Build GRCh38
rs754024328 Chr.20: 50587764-50587764 on Build GRCh38
rs755735641 Chr.20: 50587842-50587842 on Build GRCh38
rs752708232 Chr.20: 50587756-50587756 on Build GRCh38
rs755215697 Chr.20: 50587811-50587812 on Build GRCh38
rs767921187 Chr.20: 50587950-50587950 on Build GRCh38
rs758234166 Chr.20: 50587767-50587767 on Build GRCh38
rs200645158 Chr.20: 50587835-50587835 on Build GRCh38
rs376014995 Chr.20: 50587839-50587839 on Build GRCh38
rs759271502 Chr.20: 50587899-50587899 on Build GRCh38
rs1011783525 Chr.20: 50587813-50587813 on Build GRCh38
rs200853240 Chr.20: 50587851-50587851 on Build GRCh38
rs752889112 Chr.20: 50587912-50587915 on Build GRCh38
rs949321474 Chr.20: 50587971-50587971 on Build GRCh38
rs766252665 Chr.20: 50587832-50587832 on Build GRCh38
rs370425552 Chr.20: 50587875-50587875 on Build GRCh38
rs772445641 Chr.20: 50587867-50587867 on Build GRCh38
rs1047535360 Chr.20: 50588131-50588131 on Build GRCh38
rs779071415 Chr.20: 50587909-50587910 on Build GRCh38
rs548944540 Chr.20: 50587838-50587838 on Build GRCh38
rs911521037 Chr.20: 50587891-50587891 on Build GRCh38
rs534364328 Chr.20: 50587743-50587743 on Build GRCh38
rs774576887 Chr.20: 50587788-50587788 on Build GRCh38
rs1022281012 Chr.20: 50587825-50587825 on Build GRCh38
rs949248693 Chr.20: 50588115-50588115 on Build GRCh38
rs569984879 Chr.20: 50588063-50588063 on Build GRCh38
rs762015153 Chr.20: 50587831-50587831 on Build GRCh38
rs979109550 Chr.20: 50587870-50587870 on Build GRCh38
rs756722979 Chr.20: 50587943-50587943 on Build GRCh38
rs1013419150 Chr.20: 50587745-50587745 on Build GRCh38
rs553152340 Chr.20: 50588135-50588135 on Build GRCh38
rs573998664 Chr.20: 50588105-50588105 on Build GRCh38
rs764308750 Chr.20: 50587757-50587757 on Build GRCh38
rs934985749 Chr.20: 50588176-50588176 on Build GRCh38
rs879371870 Chr.20: 50587942-50587942 on Build GRCh38
rs755645820 Chr.20: 50587776-50587776 on Build GRCh38
rs559497808 Chr.20: 50587775-50587775 on Build GRCh38
rs145620777 Chr.20: 50587778-50587778 on Build GRCh38
rs898458020 Chr.20: 50588164-50588169 on Build GRCh38
rs906040452 Chr.20: 50587929-50587929 on Build GRCh38
rs765379666 Chr.20: 50587895-50587895 on Build GRCh38
rs960019135 Chr.20: 50588005-50588005 on Build GRCh38
rs563542623 Chr.20: 50587823-50587823 on Build GRCh38
rs1038785710 Chr.20: 50588157-50588157 on Build GRCh38
rs753414409 Chr.20: 50587938-50587938 on Build GRCh38
rs746226946 Chr.20: 50587866-50587866 on Build GRCh38
rs976027068 Chr.20: 50587787-50587787 on Build GRCh38
rs772472811 Chr.20: 50587802-50587802 on Build GRCh38
rs962809148 Chr.20: 50587783-50587783 on Build GRCh38
rs900445699 Chr.20: 50587739-50587739 on Build GRCh38
rs549504616 Chr.20: 50587780-50587781 on Build GRCh38
rs779789648 Chr.20: 50587770-50587770 on Build GRCh38
rs1004914076 Chr.20: 50588052-50588052 on Build GRCh38
rs201626072 Chr.20: 50587877-50587877 on Build GRCh38
rs756959501 Chr.20: 50587854-50587854 on Build GRCh38
rs754855419 Chr.20: 50587771-50587771 on Build GRCh38
rs776001816 Chr.20: 50587806-50587806 on Build GRCh38
rs781044685 Chr.20: 50587981-50587981 on Build GRCh38
rs751323373 Chr.20: 50587834-50587834 on Build GRCh38
rs917762442 Chr.20: 50588106-50588106 on Build GRCh38
rs560759358 Chr.20: 50587848-50587848 on Build GRCh38
rs375357170 Chr.20: 50587898-50587898 on Build GRCh38
rs1014995698 Chr.20: 50588073-50588073 on Build GRCh38
rs537946504 Chr.20: 50588002-50588002 on Build GRCh38
rs199517150 Chr.20: 50587919-50587919 on Build GRCh38
rs756117606 Chr.20: 50587918-50587918 on Build GRCh38
rs1054961739 Chr.20: 50587732-50587732 on Build GRCh38
rs767705794 Chr.20: 50587753-50587753 on Build GRCh38
rs966970641 Chr.20: 50588096-50588096 on Build GRCh38
rs186625375 Chr.20: 50588084-50588084 on Build GRCh38
rs182338293 Chr.20: 50587910-50587910 on Build GRCh38
rs1047608603 Chr.20: 50587972-50587972 on Build GRCh38
rs545233123 Chr.20: 50587784-50587784 on Build GRCh38
rs142563909 Chr.20: 50588134-50588134 on Build GRCh38
rs771160794 Chr.20: 50588065-50588065 on Build GRCh38
rs767321802 Chr.20: 50587904-50587904 on Build GRCh38
rs886343949 Chr.20: 50587972-50587973 on Build GRCh38

View Full Product Details