Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 6452041-6452542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBS
Entrez Gene ID
875
Gene Name
cystathionine-beta-synthase
Gene Aliases
HIP4
UniGene
Hs.533013
Gene Chromosome Location
Chr.21: 43053190-43076868 on Build GRCh38
Transcripts
NM_000071.2 NM_001178008.2 NM_001178009.2 NM_001320298.1 NM_001321072.1 XM_011529773.2 XM_011529774.2 XM_011529775.2 XM_011529776.2 XM_011529777.1 XM_011529781.1 XM_011529782.1 XM_011529783.2 XM_017028489.1 XM_017028490.1 XM_017028491.1 XM_017028492.1 XM_017028493.1 NM_001321073.1 XM_011546094.1 XM_011546095.2 XM_011546097.2 XM_011546098.1 XM_011546099.1 XM_017028209.1 XM_017028210.1 XM_017028211.1 XM_017028212.1 XM_017028213.1 XM_017028214.1 XM_017028215.1 XM_017028216.1 XM_017028217.1 XM_017028218.1 XM_017028222.1 XM_017028224.1

Gene Symbol
CBSL
Entrez Gene ID
102724560
Gene Name
cystathionine-beta-synthase like
Gene Aliases
-
UniGene
Hs.533013
Gene Chromosome Location
Chr.21: 6444868-6468606 on Build GRCh38
Transcripts
NM_000071.2 NM_001178008.2 NM_001178009.2 NM_001320298.1 NM_001321072.1 XM_011529773.2 XM_011529774.2 XM_011529775.2 XM_011529776.2 XM_011529777.1 XM_011529781.1 XM_011529782.1 XM_011529783.2 XM_017028489.1 XM_017028490.1 XM_017028491.1 XM_017028492.1 XM_017028493.1 NM_001321073.1 XM_011546094.1 XM_011546095.2 XM_011546097.2 XM_011546098.1 XM_011546099.1 XM_017028209.1 XM_017028210.1 XM_017028211.1 XM_017028212.1 XM_017028213.1 XM_017028214.1 XM_017028215.1 XM_017028216.1 XM_017028217.1 XM_017028218.1 XM_017028222.1 XM_017028224.1

Gene Symbol
LOC102724843
Entrez Gene ID
102724843
Gene Name
uncharacterized LOC102724843
Gene Aliases
-
UniGene
Hs.743984
Gene Chromosome Location
Chr.21: 6318410-7087230 on Build GRCh38
Transcripts
NM_000071.2 NM_001178008.2 NM_001178009.2 NM_001320298.1 NM_001321072.1 XM_011529773.2 XM_011529774.2 XM_011529775.2 XM_011529776.2 XM_011529777.1 XM_011529781.1 XM_011529782.1 XM_011529783.2 XM_017028489.1 XM_017028490.1 XM_017028491.1 XM_017028492.1 XM_017028493.1 NM_001321073.1 XM_011546094.1 XM_011546095.2 XM_011546097.2 XM_011546098.1 XM_011546099.1 XM_017028209.1 XM_017028210.1 XM_017028211.1 XM_017028212.1 XM_017028213.1 XM_017028214.1 XM_017028215.1 XM_017028216.1 XM_017028217.1 XM_017028218.1 XM_017028222.1 XM_017028224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4521936 c.1137G>A p.R379R Chr.21: 43060449-43060449 on Build GRCh38
4101839 c.1071C>T p.A357A Chr.21: 43060515-43060515 on Build GRCh38
4881408 c.1048G>T p.A350S Chr.21: 43060538-43060538 on Build GRCh38
2820065 c.1069G>A p.A357T Chr.21: 43060517-43060517 on Build GRCh38
3758920 c.1080C>T p.A360A Chr.21: 43060506-43060506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773495488 Chr.21: 43060585-43060585 on Build GRCh38
rs938002701 Chr.21: 43060425-43060425 on Build GRCh38
rs750399521 Chr.21: 43060553-43060553 on Build GRCh38
rs778666171 Chr.21: 43060406-43060406 on Build GRCh38
rs567744395 Chr.21: 43060703-43060703 on Build GRCh38
rs775813544 Chr.21: 43060438-43060438 on Build GRCh38
rs777371070 Chr.21: 43060422-43060422 on Build GRCh38
rs748552493 Chr.21: 43060521-43060521 on Build GRCh38
rs868467561 Chr.21: 43060463-43060463 on Build GRCh38
rs994885058 Chr.21: 43060704-43060704 on Build GRCh38
rs906809517 Chr.21: 43060673-43060673 on Build GRCh38
rs547713475 Chr.21: 43060454-43060454 on Build GRCh38
rs771062114 Chr.21: 43060424-43060424 on Build GRCh38
rs747391904 Chr.21: 43060586-43060586 on Build GRCh38
rs769080151 Chr.21: 43060451-43060451 on Build GRCh38
rs745764562 Chr.21: 43060505-43060505 on Build GRCh38
rs368206976 Chr.21: 43060515-43060515 on Build GRCh38
rs750970970 Chr.21: 43060794-43060794 on Build GRCh38
rs121964972 Chr.21: 43060528-43060528 on Build GRCh38
rs768059282 Chr.21: 43060470-43060470 on Build GRCh38
rs766784001 Chr.21: 43060839-43060839 on Build GRCh38
rs762412770 Chr.21: 43060590-43060590 on Build GRCh38
rs773726723 Chr.21: 43060525-43060525 on Build GRCh38
rs763368479 Chr.21: 43060530-43060536 on Build GRCh38
rs142991416 Chr.21: 43060680-43060680 on Build GRCh38
rs370851632 Chr.21: 43060504-43060504 on Build GRCh38
rs751709562 Chr.21: 43060653-43060653 on Build GRCh38
rs61735859 Chr.21: 43060527-43060527 on Build GRCh38
rs779940364 Chr.21: 43060510-43060510 on Build GRCh38
rs762513900 Chr.21: 43060538-43060538 on Build GRCh38
rs146146585 Chr.21: 43060769-43060769 on Build GRCh38
rs1027589416 Chr.21: 43060721-43060721 on Build GRCh38
rs781323559 Chr.21: 43060503-43060503 on Build GRCh38
rs891484295 Chr.21: 43060630-43060630 on Build GRCh38
rs1054082851 Chr.21: 43060627-43060627 on Build GRCh38
rs764604865 Chr.21: 43060771-43060771 on Build GRCh38
rs973369208 Chr.21: 43060540-43060540 on Build GRCh38
rs757098275 Chr.21: 43060490-43060490 on Build GRCh38
rs148589243 Chr.21: 43060514-43060514 on Build GRCh38
rs139145075 Chr.21: 43060473-43060473 on Build GRCh38
rs766290120 Chr.21: 43060594-43060594 on Build GRCh38
rs1033845015 Chr.21: 43060679-43060679 on Build GRCh38
rs11700812 Chr.21: 43060480-43060480 on Build GRCh38
rs780038004 Chr.21: 43060571-43060571 on Build GRCh38
rs115880859 Chr.21: 43060821-43060821 on Build GRCh38
rs863223436 Chr.21: 43060517-43060525 on Build GRCh38
rs752404089 Chr.21: 43060476-43060476 on Build GRCh38
rs201158177 Chr.21: 43060434-43060434 on Build GRCh38
rs774926464 Chr.21: 43060535-43060535 on Build GRCh38
rs767372401 Chr.21: 43060552-43060552 on Build GRCh38
rs376602117 Chr.21: 43060589-43060589 on Build GRCh38
rs921763258 Chr.21: 43060801-43060801 on Build GRCh38
rs920511437 Chr.21: 43060457-43060457 on Build GRCh38
rs267606145 Chr.21: 43060445-43060445 on Build GRCh38
rs117687681 Chr.21: 43060481-43060481 on Build GRCh38
rs863223437 Chr.21: 43060516-43060516 on Build GRCh38
rs757153278 Chr.21: 43060549-43060549 on Build GRCh38
rs749375746 Chr.21: 43060573-43060573 on Build GRCh38
rs187967192 Chr.21: 43060661-43060661 on Build GRCh38
rs779137466 Chr.21: 43060578-43060578 on Build GRCh38
rs74677118 Chr.21: 43060662-43060662 on Build GRCh38
rs267606146 Chr.21: 43060526-43060526 on Build GRCh38
rs772384826 Chr.21: 43060522-43060522 on Build GRCh38
rs553241579 Chr.21: 43060620-43060620 on Build GRCh38
rs755051842 Chr.21: 43060574-43060574 on Build GRCh38
rs754461948 Chr.21: 43060404-43060404 on Build GRCh38
rs966022443 Chr.21: 43060774-43060774 on Build GRCh38
rs763890444 Chr.21: 43060541-43060541 on Build GRCh38
rs547451850 Chr.21: 43060822-43060822 on Build GRCh38
rs745997797 Chr.21: 43060432-43060432 on Build GRCh38
rs921671074 Chr.21: 43060621-43060621 on Build GRCh38
rs370163789 Chr.21: 43060519-43060519 on Build GRCh38
rs372010465 Chr.21: 43060475-43060475 on Build GRCh38
rs950790922 Chr.21: 43060724-43060724 on Build GRCh38
rs112893409 Chr.21: 43060626-43060626 on Build GRCh38
rs150863939 Chr.21: 43060431-43060431 on Build GRCh38
rs372074099 Chr.21: 43060579-43060579 on Build GRCh38
rs775354680 Chr.21: 43060472-43060472 on Build GRCh38
rs758231468 Chr.21: 43060410-43060410 on Build GRCh38
rs944518203 Chr.21: 43060606-43060606 on Build GRCh38
rs752248248 Chr.21: 43060407-43060407 on Build GRCh38
rs863223429 Chr.21: 43060560-43060560 on Build GRCh38
rs543159513 Chr.21: 43060749-43060749 on Build GRCh38
rs570135008 Chr.21: 43060512-43060512 on Build GRCh38
rs573954005 Chr.21: 43060545-43060545 on Build GRCh38
rs757920190 Chr.21: 43060477-43060477 on Build GRCh38
rs146180894 Chr.21: 43060461-43060461 on Build GRCh38
rs763036586 Chr.21: 43060450-43060450 on Build GRCh38
rs755940179 Chr.21: 43060562-43060562 on Build GRCh38
rs753402547 Chr.21: 43060398-43060398 on Build GRCh38
rs1801181 Chr.21: 43060506-43060506 on Build GRCh38

View Full Product Details