Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 6486244-6486758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724843
Entrez Gene ID
102724843
Gene Name
uncharacterized LOC102724843
Gene Aliases
-
UniGene
Hs.743984
Gene Chromosome Location
Chr.21: 6318410-7087230 on Build GRCh38
Transcripts
XM_017028222.1 XM_017028224.1 NM_001025203.1 NM_001025204.1 NM_006758.2 XM_011529743.2 XM_017028468.1 NM_001320646.1 NM_001320648.1 NM_001320650.1 NM_001320651.1 XM_017028219.1 XM_017028220.1

Gene Symbol
U2AF1
Entrez Gene ID
7307
Gene Name
U2 small nuclear RNA auxiliary factor 1
Gene Aliases
FP793, RN, RNU2AF1, U2AF35, U2AFBP
UniGene
Hs.365116
Gene Chromosome Location
Chr.21: 43092956-43108291 on Build GRCh38
Transcripts
XM_017028222.1 XM_017028224.1 NM_001025203.1 NM_001025204.1 NM_006758.2 XM_011529743.2 XM_017028468.1 NM_001320646.1 NM_001320648.1 NM_001320650.1 NM_001320651.1 XM_017028219.1 XM_017028220.1

Gene Symbol
U2AF1L5
Entrez Gene ID
102724594
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 5
Gene Aliases
-
UniGene
Hs.365116
Gene Chromosome Location
Chr.21: 6484623-6499284 on Build GRCh38
Transcripts
XM_017028222.1 XM_017028224.1 NM_001025203.1 NM_001025204.1 NM_006758.2 XM_011529743.2 XM_017028468.1 NM_001320646.1 NM_001320648.1 NM_001320650.1 NM_001320651.1 XM_017028219.1 XM_017028220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481635 c.355C>T p.R119C Chr.21: 43094782-43094782 on Build GRCh38
5046554 c.443C>T p.T148M Chr.21: 43094694-43094694 on Build GRCh38
5552539 c.447C>T p.D149D Chr.21: 43094690-43094690 on Build GRCh38
7413186 c.471G>C p.Q157H Chr.21: 43094666-43094666 on Build GRCh38
4424185 c.409G>A p.G137R Chr.21: 43094728-43094728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562921350 Chr.21: 43094969-43094969 on Build GRCh38
rs983977689 Chr.21: 43094617-43094617 on Build GRCh38
rs750486161 Chr.21: 43094750-43094750 on Build GRCh38
rs79472640 Chr.21: 43094913-43094913 on Build GRCh38
rs1036827276 Chr.21: 43094645-43094645 on Build GRCh38
rs771982552 Chr.21: 43094880-43094880 on Build GRCh38
rs758962603 Chr.21: 43094811-43094811 on Build GRCh38
rs369161474 Chr.21: 43094607-43094607 on Build GRCh38
rs1014361823 Chr.21: 43094849-43094849 on Build GRCh38
rs754274409 Chr.21: 43094784-43094784 on Build GRCh38
rs755386140 Chr.21: 43094864-43094865 on Build GRCh38
rs554981578 Chr.21: 43094803-43094803 on Build GRCh38
rs201013690 Chr.21: 43094823-43094823 on Build GRCh38
rs574927868 Chr.21: 43094999-43094999 on Build GRCh38
rs201257428 Chr.21: 43094726-43094726 on Build GRCh38
rs375393848 Chr.21: 43094782-43094782 on Build GRCh38
rs761884349 Chr.21: 43094714-43094714 on Build GRCh38
rs764821406 Chr.21: 43094626-43094627 on Build GRCh38
rs755261252 Chr.21: 43094792-43094792 on Build GRCh38
rs1011002013 Chr.21: 43094759-43094759 on Build GRCh38
rs769567889 Chr.21: 43094670-43094670 on Build GRCh38
rs116363214 Chr.21: 43094873-43094873 on Build GRCh38
rs76116823 Chr.21: 43094859-43094859 on Build GRCh38
rs532889582 Chr.21: 43095020-43095020 on Build GRCh38
rs760951989 Chr.21: 43094768-43094768 on Build GRCh38
rs369666226 Chr.21: 43094972-43094972 on Build GRCh38
rs113762595 Chr.21: 43094791-43094791 on Build GRCh38
rs143181915 Chr.21: 43094711-43094711 on Build GRCh38
rs534135161 Chr.21: 43094883-43094883 on Build GRCh38
rs749139720 Chr.21: 43094837-43094837 on Build GRCh38
rs910971174 Chr.21: 43094907-43094908 on Build GRCh38
rs1049463459 Chr.21: 43094905-43094905 on Build GRCh38
rs199839633 Chr.21: 43094861-43094861 on Build GRCh38
rs745340227 Chr.21: 43094818-43094818 on Build GRCh38
rs767525261 Chr.21: 43094741-43094741 on Build GRCh38
rs895959470 Chr.21: 43095007-43095008 on Build GRCh38
rs749608770 Chr.21: 43094628-43094628 on Build GRCh38
rs762634673 Chr.21: 43094693-43094693 on Build GRCh38
rs972532324 Chr.21: 43094900-43094900 on Build GRCh38
rs770502189 Chr.21: 43094647-43094647 on Build GRCh38
rs767756140 Chr.21: 43094609-43094609 on Build GRCh38
rs150816679 Chr.21: 43095025-43095026 on Build GRCh38
rs189807399 Chr.21: 43094615-43094615 on Build GRCh38
rs542479458 Chr.21: 43095038-43095038 on Build GRCh38
rs372625984 Chr.21: 43094608-43094608 on Build GRCh38
rs746563559 Chr.21: 43094638-43094638 on Build GRCh38
rs750576052 Chr.21: 43094611-43094611 on Build GRCh38
rs1043452452 Chr.21: 43094934-43094934 on Build GRCh38
rs755545126 Chr.21: 43094631-43094631 on Build GRCh38
rs201658508 Chr.21: 43094678-43094678 on Build GRCh38
rs780918853 Chr.21: 43094813-43094813 on Build GRCh38
rs376810756 Chr.21: 43094614-43094614 on Build GRCh38
rs778826270 Chr.21: 43094824-43094824 on Build GRCh38
rs1014848210 Chr.21: 43095051-43095051 on Build GRCh38
rs761265676 Chr.21: 43094964-43094964 on Build GRCh38
rs779244170 Chr.21: 43094632-43094632 on Build GRCh38
rs745746725 Chr.21: 43094830-43094830 on Build GRCh38
rs138469042 Chr.21: 43094724-43094725 on Build GRCh38
rs939822449 Chr.21: 43094624-43094624 on Build GRCh38
rs61737061 Chr.21: 43094699-43094699 on Build GRCh38
rs11909645 Chr.21: 43094902-43094902 on Build GRCh38
rs200446004 Chr.21: 43094802-43094802 on Build GRCh38
rs34227794 Chr.21: 43094860-43094861 on Build GRCh38
rs933837032 Chr.21: 43094904-43094904 on Build GRCh38
rs150833884 Chr.21: 43094859-43094860 on Build GRCh38
rs540735407 Chr.21: 43094891-43094891 on Build GRCh38
rs371246226 Chr.21: 43094667-43094667 on Build GRCh38
rs182331226 Chr.21: 43094887-43094887 on Build GRCh38

View Full Product Details