Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30371323-30371833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC157
Entrez Gene ID
550631
Gene Name
coiled-coil domain containing 157
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30356635-30378655 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530269.2 XM_011530270.2 NR_046312.1

Gene Symbol
KIAA1656
Entrez Gene ID
85371
Gene Name
KIAA1656 protein
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30368811-30377905 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530269.2 XM_011530270.2 NR_046312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
141623 c.1081G>A p.E361K Chr.22: 30371685-30371685 on Build GRCh38
5366357 c.893-10_893-8delGCT p.? Chr.22: 30371640-30371642 on Build GRCh38
233898 c.893-5C>A p.? Chr.22: 30371645-30371645 on Build GRCh38
6423598 c.1108T>C p.S370P Chr.22: 30371712-30371712 on Build GRCh38
5366356 c.1046-10_1046-8delGCT p.? Chr.22: 30371640-30371642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376947614 Chr.22: 30371753-30371753 on Build GRCh38
rs770132371 Chr.22: 30371612-30371612 on Build GRCh38
rs771992873 Chr.22: 30371743-30371743 on Build GRCh38
rs974756025 Chr.22: 30371361-30371361 on Build GRCh38
rs914083669 Chr.22: 30371489-30371489 on Build GRCh38
rs764298099 Chr.22: 30371770-30371770 on Build GRCh38
rs768112594 Chr.22: 30371777-30371777 on Build GRCh38
rs192417577 Chr.22: 30371390-30371390 on Build GRCh38
rs982984613 Chr.22: 30371564-30371564 on Build GRCh38
rs756307863 Chr.22: 30371727-30371727 on Build GRCh38
rs941479230 Chr.22: 30371393-30371393 on Build GRCh38
rs376722350 Chr.22: 30371757-30371757 on Build GRCh38
rs751800987 Chr.22: 30371539-30371539 on Build GRCh38
rs757844117 Chr.22: 30371723-30371723 on Build GRCh38
rs1011178554 Chr.22: 30371538-30371538 on Build GRCh38
rs1022964145 Chr.22: 30371793-30371793 on Build GRCh38
rs1018984806 Chr.22: 30371554-30371554 on Build GRCh38
rs760246727 Chr.22: 30371673-30371673 on Build GRCh38
rs780815370 Chr.22: 30371574-30371574 on Build GRCh38
rs966135353 Chr.22: 30371559-30371559 on Build GRCh38
rs368800402 Chr.22: 30371575-30371575 on Build GRCh38
rs375013755 Chr.22: 30371611-30371611 on Build GRCh38
rs575107732 Chr.22: 30371507-30371507 on Build GRCh38
rs368568524 Chr.22: 30371418-30371418 on Build GRCh38
rs921838953 Chr.22: 30371363-30371363 on Build GRCh38
rs1076299 Chr.22: 30371792-30371792 on Build GRCh38
rs540000220 Chr.22: 30371503-30371503 on Build GRCh38
rs915763891 Chr.22: 30371424-30371424 on Build GRCh38
rs780105129 Chr.22: 30371728-30371728 on Build GRCh38
rs757233947 Chr.22: 30371605-30371605 on Build GRCh38
rs201501105 Chr.22: 30371601-30371601 on Build GRCh38
rs149676692 Chr.22: 30371719-30371719 on Build GRCh38
rs372836665 Chr.22: 30371483-30371483 on Build GRCh38
rs528916999 Chr.22: 30371527-30371527 on Build GRCh38
rs1076297 Chr.22: 30371350-30371350 on Build GRCh38
rs947000259 Chr.22: 30371647-30371647 on Build GRCh38
rs763198034 Chr.22: 30371707-30371707 on Build GRCh38
rs1076298 Chr.22: 30371740-30371740 on Build GRCh38
rs1004441607 Chr.22: 30371746-30371746 on Build GRCh38
rs907677838 Chr.22: 30371412-30371412 on Build GRCh38
rs985011947 Chr.22: 30371403-30371403 on Build GRCh38
rs970576158 Chr.22: 30371805-30371805 on Build GRCh38
rs762321290 Chr.22: 30371776-30371776 on Build GRCh38
rs41296241 Chr.22: 30371655-30371655 on Build GRCh38
rs934576699 Chr.22: 30371630-30371631 on Build GRCh38
rs781434236 Chr.22: 30371610-30371610 on Build GRCh38
rs746502360 Chr.22: 30371750-30371750 on Build GRCh38
rs375452830 Chr.22: 30371358-30371358 on Build GRCh38
rs147173226 Chr.22: 30371667-30371667 on Build GRCh38
rs573063942 Chr.22: 30371502-30371502 on Build GRCh38
rs546070583 Chr.22: 30371428-30371428 on Build GRCh38
rs892776734 Chr.22: 30371509-30371509 on Build GRCh38
rs140381193 Chr.22: 30371706-30371706 on Build GRCh38
rs748753913 Chr.22: 30371739-30371739 on Build GRCh38
rs368764882 Chr.22: 30371600-30371600 on Build GRCh38
rs907149186 Chr.22: 30371439-30371439 on Build GRCh38
rs552415037 Chr.22: 30371598-30371598 on Build GRCh38
rs764540776 Chr.22: 30371710-30371710 on Build GRCh38
rs557992150 Chr.22: 30371497-30371497 on Build GRCh38
rs768202447 Chr.22: 30371724-30371724 on Build GRCh38
rs755987405 Chr.22: 30371656-30371656 on Build GRCh38
rs774119880 Chr.22: 30371646-30371646 on Build GRCh38
rs940518972 Chr.22: 30371433-30371433 on Build GRCh38
rs201950542 Chr.22: 30371735-30371735 on Build GRCh38
rs377739355 Chr.22: 30371597-30371597 on Build GRCh38
rs773192625 Chr.22: 30371748-30371748 on Build GRCh38
rs774772794 Chr.22: 30371774-30371774 on Build GRCh38
rs759238324 Chr.22: 30371759-30371759 on Build GRCh38
rs866471861 Chr.22: 30371654-30371654 on Build GRCh38
rs750514056 Chr.22: 30371726-30371726 on Build GRCh38
rs370958308 Chr.22: 30371464-30371464 on Build GRCh38
rs367593529 Chr.22: 30371615-30371615 on Build GRCh38
rs749559164 Chr.22: 30371732-30371732 on Build GRCh38
rs997259459 Chr.22: 30371714-30371714 on Build GRCh38
rs768534098 Chr.22: 30371630-30371630 on Build GRCh38
rs780050086 Chr.22: 30371701-30371701 on Build GRCh38
rs1045658206 Chr.22: 30371434-30371434 on Build GRCh38
rs755071959 Chr.22: 30371637-30371638 on Build GRCh38
rs1045473570 Chr.22: 30371471-30371471 on Build GRCh38
rs140805007 Chr.22: 30371489-30371490 on Build GRCh38
rs948520638 Chr.22: 30371429-30371429 on Build GRCh38
rs749011319 Chr.22: 30371620-30371620 on Build GRCh38
rs867564668 Chr.22: 30371496-30371496 on Build GRCh38
rs1015760137 Chr.22: 30371748-30371748 on Build GRCh38
rs570053602 Chr.22: 30371540-30371540 on Build GRCh38
rs374393427 Chr.22: 30371377-30371377 on Build GRCh38
rs11705092 Chr.22: 30371547-30371547 on Build GRCh38
rs974702033 Chr.22: 30371572-30371572 on Build GRCh38
rs906953663 Chr.22: 30371488-30371488 on Build GRCh38

View Full Product Details