Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30371711-30372202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC157
Entrez Gene ID
550631
Gene Name
coiled-coil domain containing 157
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30356635-30378655 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530269.2 XM_011530270.2 NR_046312.1

Gene Symbol
KIAA1656
Entrez Gene ID
85371
Gene Name
KIAA1656 protein
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30368811-30377905 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530269.2 XM_011530270.2 NR_046312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3708205 c.1048C>T p.Q350* Chr.22: 30372152-30372152 on Build GRCh38
5652870 c.1138C>A p.Q380K Chr.22: 30372089-30372089 on Build GRCh38
5652871 c.985C>A p.Q329K Chr.22: 30372089-30372089 on Build GRCh38
3708204 c.1201C>T p.Q401* Chr.22: 30372152-30372152 on Build GRCh38
5881100 c.1179_1199del21 p.E400_E406delEQVQQLE Chr.22: 30372130-30372150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550787198 Chr.22: 30372143-30372143 on Build GRCh38
rs1023400624 Chr.22: 30372120-30372120 on Build GRCh38
rs376947614 Chr.22: 30371753-30371753 on Build GRCh38
rs771992873 Chr.22: 30371743-30371743 on Build GRCh38
rs769470471 Chr.22: 30372117-30372117 on Build GRCh38
rs764298099 Chr.22: 30371770-30371770 on Build GRCh38
rs781297258 Chr.22: 30372137-30372177 on Build GRCh38
rs1006213402 Chr.22: 30372123-30372123 on Build GRCh38
rs981520656 Chr.22: 30371877-30371877 on Build GRCh38
rs768112594 Chr.22: 30371777-30371777 on Build GRCh38
rs376722350 Chr.22: 30371757-30371757 on Build GRCh38
rs996880187 Chr.22: 30371836-30371836 on Build GRCh38
rs769519943 Chr.22: 30372151-30372171 on Build GRCh38
rs144439139 Chr.22: 30372083-30372083 on Build GRCh38
rs866396058 Chr.22: 30372079-30372080 on Build GRCh38
rs759592497 Chr.22: 30372184-30372184 on Build GRCh38
rs543927069 Chr.22: 30371943-30371943 on Build GRCh38
rs746982307 Chr.22: 30372104-30372104 on Build GRCh38
rs745615684 Chr.22: 30372143-30372144 on Build GRCh38
rs1022964145 Chr.22: 30371793-30371793 on Build GRCh38
rs777658110 Chr.22: 30372095-30372095 on Build GRCh38
rs151180751 Chr.22: 30372059-30372059 on Build GRCh38
rs184604773 Chr.22: 30371884-30371884 on Build GRCh38
rs573704571 Chr.22: 30371916-30371916 on Build GRCh38
rs758238452 Chr.22: 30372085-30372085 on Build GRCh38
rs199978521 Chr.22: 30372135-30372135 on Build GRCh38
rs531291137 Chr.22: 30371964-30371965 on Build GRCh38
rs760012591 Chr.22: 30372101-30372101 on Build GRCh38
rs558080511 Chr.22: 30371838-30371838 on Build GRCh38
rs1003371738 Chr.22: 30372163-30372163 on Build GRCh38
rs933081144 Chr.22: 30372030-30372030 on Build GRCh38
rs866469501 Chr.22: 30372128-30372128 on Build GRCh38
rs189397329 Chr.22: 30372031-30372031 on Build GRCh38
rs747774293 Chr.22: 30371878-30371878 on Build GRCh38
rs1076299 Chr.22: 30371792-30371792 on Build GRCh38
rs991224475 Chr.22: 30371976-30371976 on Build GRCh38
rs568512072 Chr.22: 30372106-30372106 on Build GRCh38
rs1041811297 Chr.22: 30372022-30372022 on Build GRCh38
rs773808964 Chr.22: 30372140-30372140 on Build GRCh38
rs924184402 Chr.22: 30372026-30372026 on Build GRCh38
rs1076298 Chr.22: 30371740-30371740 on Build GRCh38
rs751003621 Chr.22: 30372025-30372025 on Build GRCh38
rs1004441607 Chr.22: 30371746-30371746 on Build GRCh38
rs914312372 Chr.22: 30372002-30372002 on Build GRCh38
rs970576158 Chr.22: 30371805-30371805 on Build GRCh38
rs762321290 Chr.22: 30371776-30371776 on Build GRCh38
rs775050973 Chr.22: 30372121-30372121 on Build GRCh38
rs757200175 Chr.22: 30372135-30372135 on Build GRCh38
rs532758341 Chr.22: 30371979-30371979 on Build GRCh38
rs952046965 Chr.22: 30371850-30371850 on Build GRCh38
rs746502360 Chr.22: 30371750-30371750 on Build GRCh38
rs368707665 Chr.22: 30372118-30372118 on Build GRCh38
rs766504356 Chr.22: 30372032-30372032 on Build GRCh38
rs745435347 Chr.22: 30372111-30372111 on Build GRCh38
rs562156181 Chr.22: 30371956-30371956 on Build GRCh38
rs748753913 Chr.22: 30371739-30371739 on Build GRCh38
rs569319348 Chr.22: 30372164-30372164 on Build GRCh38
rs779076098 Chr.22: 30372071-30372071 on Build GRCh38
rs539784385 Chr.22: 30371812-30371812 on Build GRCh38
rs555237613 Chr.22: 30371915-30371915 on Build GRCh38
rs761280727 Chr.22: 30372160-30372160 on Build GRCh38
rs371254570 Chr.22: 30372171-30372171 on Build GRCh38
rs778926988 Chr.22: 30372119-30372138 on Build GRCh38
rs201950542 Chr.22: 30371735-30371735 on Build GRCh38
rs540407405 Chr.22: 30371907-30371907 on Build GRCh38
rs773192625 Chr.22: 30371748-30371748 on Build GRCh38
rs774772794 Chr.22: 30371774-30371774 on Build GRCh38
rs759238324 Chr.22: 30371759-30371759 on Build GRCh38
rs755237373 Chr.22: 30372070-30372070 on Build GRCh38
rs148797876 Chr.22: 30372155-30372155 on Build GRCh38
rs749559164 Chr.22: 30371732-30371732 on Build GRCh38
rs928474591 Chr.22: 30371928-30371928 on Build GRCh38
rs935609931 Chr.22: 30372035-30372035 on Build GRCh38
rs958544252 Chr.22: 30371932-30371932 on Build GRCh38
rs1040623214 Chr.22: 30372056-30372056 on Build GRCh38
rs752824336 Chr.22: 30372130-30372150 on Build GRCh38
rs1015366870 Chr.22: 30372124-30372124 on Build GRCh38
rs772593756 Chr.22: 30372137-30372137 on Build GRCh38
rs770367866 Chr.22: 30372105-30372105 on Build GRCh38
rs1014839548 Chr.22: 30371892-30371892 on Build GRCh38
rs1015760137 Chr.22: 30371748-30371748 on Build GRCh38
rs41308741 Chr.22: 30372008-30372008 on Build GRCh38
rs373226405 Chr.22: 30371822-30371822 on Build GRCh38
rs1037154269 Chr.22: 30372110-30372110 on Build GRCh38

View Full Product Details