Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37127315-37127855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RB
Entrez Gene ID
3560
Gene Name
interleukin 2 receptor subunit beta
Gene Aliases
CD122, IL15RB, P70-75
UniGene
Hs.474787
Gene Chromosome Location
Chr.22: 37125838-37149922 on Build GRCh38
Transcripts
NM_000878.3

SNP Detail

SNP ID Chromosome Location
rs750963931 Chr.22: 37127793-37127793 on Build GRCh38
rs76794334 Chr.22: 37127796-37127796 on Build GRCh38
rs540668132 Chr.22: 37127734-37127734 on Build GRCh38
rs1037035913 Chr.22: 37127357-37127357 on Build GRCh38
rs3218365 Chr.22: 37127450-37127450 on Build GRCh38
rs942344897 Chr.22: 37127662-37127662 on Build GRCh38
rs1046507835 Chr.22: 37127442-37127442 on Build GRCh38
rs1005805746 Chr.22: 37127517-37127517 on Build GRCh38
rs935163239 Chr.22: 37127694-37127694 on Build GRCh38
rs954505416 Chr.22: 37127593-37127593 on Build GRCh38
rs771615675 Chr.22: 37127607-37127607 on Build GRCh38
rs569105857 Chr.22: 37127696-37127698 on Build GRCh38
rs868228198 Chr.22: 37127623-37127623 on Build GRCh38
rs774409608 Chr.22: 37127473-37127473 on Build GRCh38
rs888667291 Chr.22: 37127516-37127516 on Build GRCh38
rs774803579 Chr.22: 37127622-37127622 on Build GRCh38
rs962151750 Chr.22: 37127791-37127791 on Build GRCh38
rs999283527 Chr.22: 37127786-37127786 on Build GRCh38
rs923832112 Chr.22: 37127371-37127371 on Build GRCh38
rs753430154 Chr.22: 37127487-37127487 on Build GRCh38
rs954579491 Chr.22: 37127758-37127758 on Build GRCh38
rs905207206 Chr.22: 37127746-37127746 on Build GRCh38
rs767890904 Chr.22: 37127481-37127481 on Build GRCh38
rs551022537 Chr.22: 37127610-37127610 on Build GRCh38
rs773201526 Chr.22: 37127342-37127342 on Build GRCh38
rs1047471213 Chr.22: 37127693-37127693 on Build GRCh38
rs559022107 Chr.22: 37127739-37127739 on Build GRCh38
rs3218331 Chr.22: 37127561-37127561 on Build GRCh38
rs3218330 Chr.22: 37127624-37127624 on Build GRCh38
rs1029799971 Chr.22: 37127757-37127757 on Build GRCh38
rs41309804 Chr.22: 37127445-37127445 on Build GRCh38
rs528844032 Chr.22: 37127729-37127729 on Build GRCh38
rs1031343330 Chr.22: 37127463-37127463 on Build GRCh38
rs763646939 Chr.22: 37127638-37127638 on Build GRCh38
rs228941 Chr.22: 37127681-37127681 on Build GRCh38
rs947854041 Chr.22: 37127644-37127644 on Build GRCh38
rs889569289 Chr.22: 37127401-37127401 on Build GRCh38
rs759000981 Chr.22: 37127568-37127568 on Build GRCh38
rs947905991 Chr.22: 37127421-37127421 on Build GRCh38
rs3218332 Chr.22: 37127391-37127391 on Build GRCh38
rs1052270910 Chr.22: 37127706-37127706 on Build GRCh38
rs749092330 Chr.22: 37127384-37127384 on Build GRCh38
rs3218329 Chr.22: 37127652-37127652 on Build GRCh38
rs986361388 Chr.22: 37127800-37127800 on Build GRCh38
rs374788096 Chr.22: 37127633-37127633 on Build GRCh38
rs185417988 Chr.22: 37127596-37127596 on Build GRCh38
rs1030488863 Chr.22: 37127794-37127794 on Build GRCh38
rs780198407 Chr.22: 37127813-37127813 on Build GRCh38
rs999335960 Chr.22: 37127461-37127461 on Build GRCh38
rs569839053 Chr.22: 37127551-37127551 on Build GRCh38
rs567514277 Chr.22: 37127629-37127629 on Build GRCh38
rs765532862 Chr.22: 37127752-37127752 on Build GRCh38
rs192627099 Chr.22: 37127584-37127584 on Build GRCh38
rs910774279 Chr.22: 37127802-37127802 on Build GRCh38
rs542924493 Chr.22: 37127682-37127682 on Build GRCh38
rs192146486 Chr.22: 37127821-37127821 on Build GRCh38
rs936616299 Chr.22: 37127381-37127381 on Build GRCh38
rs1050783948 Chr.22: 37127399-37127399 on Build GRCh38
rs979026667 Chr.22: 37127820-37127820 on Build GRCh38
rs34875471 Chr.22: 37127744-37127744 on Build GRCh38
rs536909831 Chr.22: 37127558-37127558 on Build GRCh38
rs551335257 Chr.22: 37127474-37127474 on Build GRCh38
rs545487206 Chr.22: 37127626-37127626 on Build GRCh38
rs755295087 Chr.22: 37127824-37127824 on Build GRCh38
rs1006580532 Chr.22: 37127428-37127428 on Build GRCh38
rs189536875 Chr.22: 37127724-37127724 on Build GRCh38
rs532844221 Chr.22: 37127756-37127756 on Build GRCh38
rs1023632265 Chr.22: 37127541-37127541 on Build GRCh38
rs935087388 Chr.22: 37127831-37127831 on Build GRCh38
rs960615980 Chr.22: 37127467-37127467 on Build GRCh38

View Full Product Details