Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37137621-37138160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RB
Entrez Gene ID
3560
Gene Name
interleukin 2 receptor subunit beta
Gene Aliases
CD122, IL15RB, P70-75
UniGene
Hs.474787
Gene Chromosome Location
Chr.22: 37125838-37149922 on Build GRCh38
Transcripts
NM_000878.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4103913 c.447C>T p.N149N Chr.22: 37137677-37137677 on Build GRCh38
135919 c.455G>A p.W152* Chr.22: 37137669-37137669 on Build GRCh38
240225 c.392G>A p.R131H Chr.22: 37137732-37137732 on Build GRCh38
228754 c.470C>T p.A157V Chr.22: 37137654-37137654 on Build GRCh38
4493199 c.410C>T p.S137F Chr.22: 37137714-37137714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781374953 Chr.22: 37137733-37137733 on Build GRCh38
rs117168830 Chr.22: 37138136-37138136 on Build GRCh38
rs1023804475 Chr.22: 37137979-37137979 on Build GRCh38
rs2284033 Chr.22: 37137994-37137994 on Build GRCh38
rs896624558 Chr.22: 37137951-37137951 on Build GRCh38
rs228957 Chr.22: 37137746-37137746 on Build GRCh38
rs3218354 Chr.22: 37137826-37137826 on Build GRCh38
rs746806656 Chr.22: 37137730-37137730 on Build GRCh38
rs764464996 Chr.22: 37137785-37137785 on Build GRCh38
rs79572807 Chr.22: 37138094-37138094 on Build GRCh38
rs867355828 Chr.22: 37137713-37137713 on Build GRCh38
rs889792718 Chr.22: 37137841-37137841 on Build GRCh38
rs367622513 Chr.22: 37137749-37137749 on Build GRCh38
rs1046664679 Chr.22: 37137816-37137816 on Build GRCh38
rs1043867397 Chr.22: 37137878-37137878 on Build GRCh38
rs763434888 Chr.22: 37137710-37137710 on Build GRCh38
rs768430184 Chr.22: 37137750-37137750 on Build GRCh38
rs1031605297 Chr.22: 37137938-37137938 on Build GRCh38
rs564308508 Chr.22: 37138090-37138090 on Build GRCh38
rs1053636405 Chr.22: 37137795-37137795 on Build GRCh38
rs774603661 Chr.22: 37137787-37137787 on Build GRCh38
rs1016661045 Chr.22: 37138008-37138008 on Build GRCh38
rs916792395 Chr.22: 37137762-37137762 on Build GRCh38
rs941234124 Chr.22: 37137791-37137791 on Build GRCh38
rs918401552 Chr.22: 37137803-37137803 on Build GRCh38
rs776134569 Chr.22: 37137781-37137781 on Build GRCh38
rs1031469496 Chr.22: 37138060-37138060 on Build GRCh38
rs541569355 Chr.22: 37138056-37138056 on Build GRCh38
rs778736826 Chr.22: 37138070-37138070 on Build GRCh38
rs751839490 Chr.22: 37137827-37137827 on Build GRCh38
rs771510617 Chr.22: 37137651-37137651 on Build GRCh38
rs528614736 Chr.22: 37138088-37138088 on Build GRCh38
rs571524244 Chr.22: 37138091-37138091 on Build GRCh38
rs777770000 Chr.22: 37137964-37137964 on Build GRCh38
rs770273658 Chr.22: 37137775-37137775 on Build GRCh38
rs530825663 Chr.22: 37137893-37137893 on Build GRCh38
rs200697945 Chr.22: 37137743-37137743 on Build GRCh38
rs942673877 Chr.22: 37137864-37137864 on Build GRCh38
rs757260348 Chr.22: 37138058-37138058 on Build GRCh38
rs765775857 Chr.22: 37137678-37137678 on Build GRCh38
rs371794346 Chr.22: 37137653-37137653 on Build GRCh38
rs982980989 Chr.22: 37138134-37138134 on Build GRCh38
rs760228358 Chr.22: 37137756-37137756 on Build GRCh38
rs149724439 Chr.22: 37137783-37137783 on Build GRCh38
rs752982148 Chr.22: 37137697-37137697 on Build GRCh38
rs372164885 Chr.22: 37137752-37137752 on Build GRCh38
rs979733184 Chr.22: 37138000-37138000 on Build GRCh38
rs755194037 Chr.22: 37137839-37137839 on Build GRCh38
rs930185783 Chr.22: 37137857-37137857 on Build GRCh38
rs961230216 Chr.22: 37137738-37137738 on Build GRCh38
rs149258462 Chr.22: 37137665-37137665 on Build GRCh38
rs148746744 Chr.22: 37138081-37138081 on Build GRCh38
rs972958214 Chr.22: 37137800-37137800 on Build GRCh38
rs886205367 Chr.22: 37137899-37137899 on Build GRCh38
rs903925618 Chr.22: 37137880-37137880 on Build GRCh38
rs228958 Chr.22: 37137755-37137755 on Build GRCh38
rs972479428 Chr.22: 37138061-37138061 on Build GRCh38
rs117687194 Chr.22: 37138102-37138102 on Build GRCh38
rs764222346 Chr.22: 37137718-37137718 on Build GRCh38
rs551388617 Chr.22: 37138127-37138127 on Build GRCh38
rs935428147 Chr.22: 37138106-37138106 on Build GRCh38
rs1030275515 Chr.22: 37138032-37138032 on Build GRCh38
rs962792692 Chr.22: 37138032-37138032 on Build GRCh38
rs1006269184 Chr.22: 37137939-37137939 on Build GRCh38
rs201441730 Chr.22: 37137723-37137723 on Build GRCh38

View Full Product Details