Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112464377-112464891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTLA
Entrez Gene ID
151888
Gene Name
B and T lymphocyte associated
Gene Aliases
BTLA1, CD272
UniGene
Hs.445162
Gene Chromosome Location
Chr.3: 112458790-112499756 on Build GRCh38
Transcripts
NM_001085357.1 NM_181780.3 XM_011512447.2 XM_017005748.1

SNP Detail

SNP ID Chromosome Location
rs769335874 Chr.3: 112464747-112464747 on Build GRCh38
rs745477310 Chr.3: 112464542-112464542 on Build GRCh38
rs868368190 Chr.3: 112464717-112464717 on Build GRCh38
rs995132138 Chr.3: 112464861-112464861 on Build GRCh38
rs1006229270 Chr.3: 112464825-112464826 on Build GRCh38
rs367691267 Chr.3: 112464798-112464798 on Build GRCh38
rs544467464 Chr.3: 112464829-112464829 on Build GRCh38
rs766533087 Chr.3: 112464623-112464623 on Build GRCh38
rs944883827 Chr.3: 112464825-112464825 on Build GRCh38
rs988046114 Chr.3: 112464666-112464666 on Build GRCh38
rs141341566 Chr.3: 112464646-112464646 on Build GRCh38
rs1043338181 Chr.3: 112464572-112464572 on Build GRCh38
rs1050503877 Chr.3: 112464800-112464800 on Build GRCh38
rs913795517 Chr.3: 112464700-112464700 on Build GRCh38
rs528401354 Chr.3: 112464830-112464830 on Build GRCh38
rs397744204 Chr.3: 112464829-112464830 on Build GRCh38
rs776885125 Chr.3: 112464660-112464660 on Build GRCh38
rs71631391 Chr.3: 112464793-112464794 on Build GRCh38
rs1001915996 Chr.3: 112464556-112464556 on Build GRCh38
rs764110433 Chr.3: 112464734-112464734 on Build GRCh38
rs969099549 Chr.3: 112464521-112464521 on Build GRCh38
rs144039869 Chr.3: 112464543-112464543 on Build GRCh38
rs777748219 Chr.3: 112464455-112464455 on Build GRCh38
rs146465696 Chr.3: 112464611-112464611 on Build GRCh38
rs572294197 Chr.3: 112464446-112464446 on Build GRCh38
rs984073124 Chr.3: 112464422-112464422 on Build GRCh38
rs1036563905 Chr.3: 112464828-112464828 on Build GRCh38
rs61381878 Chr.3: 112464799-112464800 on Build GRCh38
rs992672774 Chr.3: 112464794-112464794 on Build GRCh38
rs774440200 Chr.3: 112464800-112464801 on Build GRCh38
rs932093940 Chr.3: 112464411-112464411 on Build GRCh38
rs546113337 Chr.3: 112464742-112464742 on Build GRCh38
rs56752404 Chr.3: 112464826-112464829 on Build GRCh38
rs762753442 Chr.3: 112464727-112464727 on Build GRCh38
rs746520958 Chr.3: 112464456-112464456 on Build GRCh38
rs761025046 Chr.3: 112464567-112464567 on Build GRCh38
rs912042654 Chr.3: 112464816-112464816 on Build GRCh38
rs570533860 Chr.3: 112464741-112464743 on Build GRCh38
rs1050575700 Chr.3: 112464415-112464415 on Build GRCh38
rs776435626 Chr.3: 112464826-112464828 on Build GRCh38
rs181984803 Chr.3: 112464462-112464462 on Build GRCh38
rs1017593156 Chr.3: 112464827-112464828 on Build GRCh38
rs994900967 Chr.3: 112464503-112464503 on Build GRCh38
rs979168304 Chr.3: 112464765-112464765 on Build GRCh38
rs933763099 Chr.3: 112464519-112464519 on Build GRCh38
rs574805614 Chr.3: 112464464-112464464 on Build GRCh38
rs913594504 Chr.3: 112464544-112464544 on Build GRCh38
rs574017917 Chr.3: 112464766-112464766 on Build GRCh38
rs772382532 Chr.3: 112464489-112464489 on Build GRCh38
rs767437251 Chr.3: 112464826-112464827 on Build GRCh38
rs1057066556 Chr.3: 112464709-112464709 on Build GRCh38
rs202063221 Chr.3: 112464827-112464830 on Build GRCh38
rs139355057 Chr.3: 112464798-112464799 on Build GRCh38
rs189468208 Chr.3: 112464451-112464451 on Build GRCh38
rs56224196 Chr.3: 112464513-112464513 on Build GRCh38

View Full Product Details