Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129038610-129039101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
UniGene
Hs.134807
Gene Chromosome Location
Chr.3: 129001604-129040742 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513161.2 XM_011513164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1038031 c.287C>T p.S96L Chr.3: 129038838-129038838 on Build GRCh38
3059916 c.1615A>G p.K539E Chr.3: 129038855-129038855 on Build GRCh38
5418365 c.340C>T p.P114S Chr.3: 129038891-129038891 on Build GRCh38
1149603 c.1624C>T p.L542L Chr.3: 129038864-129038864 on Build GRCh38
4669957 c.1659C>T p.H553H Chr.3: 129038899-129038899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920550381 Chr.3: 129039015-129039015 on Build GRCh38
rs545756939 Chr.3: 129038852-129038852 on Build GRCh38
rs760550317 Chr.3: 129038915-129038915 on Build GRCh38
rs377257335 Chr.3: 129038885-129038885 on Build GRCh38
rs368823118 Chr.3: 129038907-129038907 on Build GRCh38
rs551232704 Chr.3: 129038919-129038919 on Build GRCh38
rs574287825 Chr.3: 129038748-129038748 on Build GRCh38
rs960641307 Chr.3: 129038989-129038989 on Build GRCh38
rs541649794 Chr.3: 129038750-129038750 on Build GRCh38
rs551709763 Chr.3: 129038937-129038937 on Build GRCh38
rs1022636964 Chr.3: 129038933-129038933 on Build GRCh38
rs968833231 Chr.3: 129038940-129038940 on Build GRCh38
rs1050490351 Chr.3: 129038752-129038752 on Build GRCh38
rs1035331267 Chr.3: 129038637-129038637 on Build GRCh38
rs752091923 Chr.3: 129038879-129038879 on Build GRCh38
rs552888025 Chr.3: 129038732-129038732 on Build GRCh38
rs1031434779 Chr.3: 129038949-129038949 on Build GRCh38
rs557885282 Chr.3: 129038707-129038707 on Build GRCh38
rs746502865 Chr.3: 129038751-129038751 on Build GRCh38
rs942137624 Chr.3: 129038793-129038793 on Build GRCh38
rs374722185 Chr.3: 129038878-129038878 on Build GRCh38
rs768081619 Chr.3: 129038944-129038944 on Build GRCh38
rs2306529 Chr.3: 129038717-129038717 on Build GRCh38
rs770055006 Chr.3: 129038834-129038834 on Build GRCh38
rs889194523 Chr.3: 129038772-129038772 on Build GRCh38
rs575259155 Chr.3: 129038839-129038839 on Build GRCh38
rs954022785 Chr.3: 129039026-129039026 on Build GRCh38
rs768416477 Chr.3: 129038845-129038845 on Build GRCh38
rs910556300 Chr.3: 129039065-129039065 on Build GRCh38
rs754531443 Chr.3: 129038872-129038872 on Build GRCh38
rs923852834 Chr.3: 129038943-129038943 on Build GRCh38
rs746292150 Chr.3: 129038889-129038889 on Build GRCh38
rs372442195 Chr.3: 129038910-129038910 on Build GRCh38
rs36093369 Chr.3: 129038632-129038633 on Build GRCh38
rs750776457 Chr.3: 129038671-129038671 on Build GRCh38
rs867391984 Chr.3: 129038979-129038979 on Build GRCh38
rs778350924 Chr.3: 129038783-129038783 on Build GRCh38
rs749276603 Chr.3: 129038906-129038906 on Build GRCh38
rs781518640 Chr.3: 129038886-129038886 on Build GRCh38
rs533976895 Chr.3: 129039033-129039033 on Build GRCh38
rs367935536 Chr.3: 129038782-129038782 on Build GRCh38
rs757641354 Chr.3: 129038786-129038786 on Build GRCh38
rs781353174 Chr.3: 129038796-129038796 on Build GRCh38
rs955859352 Chr.3: 129038959-129038959 on Build GRCh38
rs370605612 Chr.3: 129038802-129038802 on Build GRCh38
rs1047792217 Chr.3: 129039079-129039079 on Build GRCh38
rs575797318 Chr.3: 129038833-129038833 on Build GRCh38
rs760390838 Chr.3: 129038857-129038857 on Build GRCh38
rs750722827 Chr.3: 129038946-129038946 on Build GRCh38
rs563929853 Chr.3: 129038855-129038855 on Build GRCh38
rs748061270 Chr.3: 129038965-129038965 on Build GRCh38
rs755916552 Chr.3: 129038924-129038925 on Build GRCh38
rs774366140 Chr.3: 129038847-129038847 on Build GRCh38
rs529491405 Chr.3: 129038902-129038902 on Build GRCh38
rs539551115 Chr.3: 129038934-129038934 on Build GRCh38
rs752617267 Chr.3: 129038806-129038806 on Build GRCh38
rs991200227 Chr.3: 129038654-129038654 on Build GRCh38
rs1044496361 Chr.3: 129039013-129039013 on Build GRCh38
rs1023119354 Chr.3: 129038670-129038670 on Build GRCh38
rs566740929 Chr.3: 129038947-129038947 on Build GRCh38
rs747319412 Chr.3: 129038784-129038784 on Build GRCh38
rs976648971 Chr.3: 129038725-129038725 on Build GRCh38
rs138572817 Chr.3: 129038899-129038899 on Build GRCh38
rs773135739 Chr.3: 129038914-129038914 on Build GRCh38
rs991938134 Chr.3: 129038991-129038991 on Build GRCh38
rs959988937 Chr.3: 129038639-129038639 on Build GRCh38
rs772194455 Chr.3: 129038912-129038912 on Build GRCh38
rs916438173 Chr.3: 129038996-129038996 on Build GRCh38
rs267599600 Chr.3: 129038838-129038838 on Build GRCh38
rs964526379 Chr.3: 129038650-129038650 on Build GRCh38
rs746062614 Chr.3: 129038823-129038823 on Build GRCh38
rs946600868 Chr.3: 129038690-129038690 on Build GRCh38
rs528755361 Chr.3: 129038861-129038861 on Build GRCh38
rs758041125 Chr.3: 129038710-129038710 on Build GRCh38
rs544762122 Chr.3: 129038932-129038932 on Build GRCh38
rs758929534 Chr.3: 129038928-129038928 on Build GRCh38
rs756520622 Chr.3: 129038948-129038948 on Build GRCh38
rs754483940 Chr.3: 129038871-129038871 on Build GRCh38
rs758627432 Chr.3: 129039005-129039005 on Build GRCh38
rs774610017 Chr.3: 129038909-129038909 on Build GRCh38
rs922507593 Chr.3: 129038988-129038988 on Build GRCh38
rs761656680 Chr.3: 129038850-129038850 on Build GRCh38
rs534611070 Chr.3: 129038731-129038731 on Build GRCh38

View Full Product Details