Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160226681-160227190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf80
Entrez Gene ID
401097
Gene Name
chromosome 3 open reading frame 80
Gene Aliases
-
UniGene
Hs.729755
Gene Chromosome Location
Chr.3: 160225636-160228213 on Build GRCh38
Transcripts
NM_001168214.1

SNP Detail

SNP ID Chromosome Location
rs144190121 Chr.3: 160226731-160226731 on Build GRCh38
rs867434639 Chr.3: 160226778-160226778 on Build GRCh38
rs139400428 Chr.3: 160227105-160227105 on Build GRCh38
rs540866066 Chr.3: 160226925-160226925 on Build GRCh38
rs1044219279 Chr.3: 160226806-160226806 on Build GRCh38
rs1023982036 Chr.3: 160226782-160226782 on Build GRCh38
rs934011613 Chr.3: 160226979-160226979 on Build GRCh38
rs923949263 Chr.3: 160226712-160226712 on Build GRCh38
rs989686331 Chr.3: 160226893-160226893 on Build GRCh38
rs1007034604 Chr.3: 160226970-160226970 on Build GRCh38
rs752098599 Chr.3: 160227119-160227119 on Build GRCh38
rs933934344 Chr.3: 160226708-160226708 on Build GRCh38
rs896809496 Chr.3: 160226878-160226878 on Build GRCh38
rs866861201 Chr.3: 160227128-160227128 on Build GRCh38
rs1031706452 Chr.3: 160227018-160227018 on Build GRCh38
rs928498562 Chr.3: 160226824-160226824 on Build GRCh38
rs146766340 Chr.3: 160227064-160227064 on Build GRCh38
rs978551258 Chr.3: 160226968-160226968 on Build GRCh38
rs1022431313 Chr.3: 160226894-160226894 on Build GRCh38
rs138522487 Chr.3: 160227153-160227154 on Build GRCh38
rs547396093 Chr.3: 160226860-160226860 on Build GRCh38
rs942410889 Chr.3: 160227124-160227124 on Build GRCh38
rs922643939 Chr.3: 160226978-160226978 on Build GRCh38
rs1048548232 Chr.3: 160227110-160227110 on Build GRCh38
rs959603229 Chr.3: 160226880-160226880 on Build GRCh38
rs906833805 Chr.3: 160226833-160226833 on Build GRCh38
rs564832819 Chr.3: 160226843-160226843 on Build GRCh38
rs551047971 Chr.3: 160227077-160227077 on Build GRCh38
rs989904002 Chr.3: 160227023-160227023 on Build GRCh38
rs528757511 Chr.3: 160226877-160226877 on Build GRCh38
rs985415995 Chr.3: 160227022-160227022 on Build GRCh38
rs763344923 Chr.3: 160226734-160226734 on Build GRCh38
rs916652933 Chr.3: 160226745-160226745 on Build GRCh38
rs918503204 Chr.3: 160227100-160227100 on Build GRCh38
rs562132495 Chr.3: 160226948-160226948 on Build GRCh38
rs764193987 Chr.3: 160226946-160226949 on Build GRCh38
rs972641387 Chr.3: 160227091-160227091 on Build GRCh38
rs535152574 Chr.3: 160227018-160227018 on Build GRCh38
rs929755302 Chr.3: 160227075-160227075 on Build GRCh38
rs911173579 Chr.3: 160227051-160227051 on Build GRCh38
rs761975467 Chr.3: 160227055-160227055 on Build GRCh38
rs896944222 Chr.3: 160226767-160226767 on Build GRCh38
rs551784002 Chr.3: 160227158-160227158 on Build GRCh38
rs569294957 Chr.3: 160227086-160227086 on Build GRCh38
rs1049606155 Chr.3: 160226720-160226720 on Build GRCh38
rs1038171536 Chr.3: 160227062-160227062 on Build GRCh38
rs889683972 Chr.3: 160226728-160226728 on Build GRCh38
rs901335167 Chr.3: 160227140-160227140 on Build GRCh38

View Full Product Details