Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194688776-194689287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM43A
Entrez Gene ID
131583
Gene Name
family with sequence similarity 43 member A
Gene Aliases
-
UniGene
Hs.435080
Gene Chromosome Location
Chr.3: 194685893-194689037 on Build GRCh38
Transcripts
NM_153690.4

SNP Detail

SNP ID Chromosome Location
rs909624412 Chr.3: 194689080-194689080 on Build GRCh38
rs966850517 Chr.3: 194689028-194689028 on Build GRCh38
rs934910163 Chr.3: 194689046-194689046 on Build GRCh38
rs755679247 Chr.3: 194688888-194688888 on Build GRCh38
rs986229999 Chr.3: 194688909-194688909 on Build GRCh38
rs538901701 Chr.3: 194688864-194688864 on Build GRCh38
rs1036655778 Chr.3: 194689107-194689107 on Build GRCh38
rs975377917 Chr.3: 194689060-194689060 on Build GRCh38
rs1022767596 Chr.3: 194689012-194689012 on Build GRCh38
rs968734527 Chr.3: 194688947-194688947 on Build GRCh38
rs897057618 Chr.3: 194688825-194688825 on Build GRCh38
rs367752446 Chr.3: 194689236-194689238 on Build GRCh38
rs888239593 Chr.3: 194689103-194689103 on Build GRCh38
rs917419913 Chr.3: 194689061-194689061 on Build GRCh38
rs1000240679 Chr.3: 194689241-194689241 on Build GRCh38
rs1033708185 Chr.3: 194688988-194688988 on Build GRCh38
rs1044148973 Chr.3: 194689063-194689063 on Build GRCh38
rs746095224 Chr.3: 194688901-194688904 on Build GRCh38
rs76584419 Chr.3: 194689231-194689231 on Build GRCh38
rs768285655 Chr.3: 194689100-194689100 on Build GRCh38
rs538067671 Chr.3: 194688886-194688886 on Build GRCh38
rs998522424 Chr.3: 194689163-194689163 on Build GRCh38
rs948866797 Chr.3: 194689062-194689062 on Build GRCh38
rs1050998101 Chr.3: 194689167-194689167 on Build GRCh38
rs115571843 Chr.3: 194689214-194689214 on Build GRCh38
rs569229979 Chr.3: 194688978-194688978 on Build GRCh38
rs139694031 Chr.3: 194689102-194689102 on Build GRCh38
rs1006713750 Chr.3: 194689203-194689203 on Build GRCh38
rs765042646 Chr.3: 194688860-194688860 on Build GRCh38
rs143080437 Chr.3: 194688819-194688819 on Build GRCh38
rs779514823 Chr.3: 194688944-194688944 on Build GRCh38
rs924927266 Chr.3: 194689016-194689016 on Build GRCh38
rs749001689 Chr.3: 194689068-194689068 on Build GRCh38
rs1011282160 Chr.3: 194689222-194689222 on Build GRCh38
rs1022292022 Chr.3: 194689223-194689223 on Build GRCh38
rs555039668 Chr.3: 194689047-194689047 on Build GRCh38
rs1015434160 Chr.3: 194688907-194688907 on Build GRCh38
rs896781536 Chr.3: 194689162-194689162 on Build GRCh38
rs989592044 Chr.3: 194689000-194689000 on Build GRCh38
rs536754927 Chr.3: 194688999-194688999 on Build GRCh38
rs1004011761 Chr.3: 194688898-194688898 on Build GRCh38
rs550005414 Chr.3: 194688996-194688996 on Build GRCh38
rs756424779 Chr.3: 194688878-194688878 on Build GRCh38
rs998070058 Chr.3: 194688880-194688880 on Build GRCh38
rs978750159 Chr.3: 194688960-194688960 on Build GRCh38
rs1033983636 Chr.3: 194689211-194689211 on Build GRCh38
rs953873153 Chr.3: 194688900-194688900 on Build GRCh38
rs1003478320 Chr.3: 194689208-194689208 on Build GRCh38
rs1003450257 Chr.3: 194688952-194688952 on Build GRCh38
rs889639277 Chr.3: 194689183-194689183 on Build GRCh38
rs932275895 Chr.3: 194689077-194689077 on Build GRCh38
rs150683111 Chr.3: 194688942-194688942 on Build GRCh38
rs180745316 Chr.3: 194689059-194689059 on Build GRCh38
rs559019426 Chr.3: 194689166-194689166 on Build GRCh38

View Full Product Details