Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8440673-8441168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX3
Entrez Gene ID
8310
Gene Name
acyl-CoA oxidase 3, pristanoyl
Gene Aliases
-
UniGene
Hs.479122
Gene Chromosome Location
Chr.4: 8366263-8440725 on Build GRCh38
Transcripts
NM_001101667.1 NM_003501.2 NM_152544.2 XM_005247950.3 XM_011513406.2 XM_011513407.2 XM_011513408.1 XM_011513409.2 XM_011513410.2 XM_017007811.1

Gene Symbol
TRMT44
Entrez Gene ID
152992
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases
C4orf23, METTL19, TRM44
UniGene
Hs.566191
Gene Chromosome Location
Chr.4: 8440774-8512531 on Build GRCh38
Transcripts
NM_001101667.1 NM_003501.2 NM_152544.2 XM_005247950.3 XM_011513406.2 XM_011513407.2 XM_011513408.1 XM_011513409.2 XM_011513410.2 XM_017007811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836294 c.52C>T p.Q18* Chr.4: 8440874-8440874 on Build GRCh38
7284553 c.56G>A p.G19D Chr.4: 8440878-8440878 on Build GRCh38
5697955 c.120C>A p.G40G Chr.4: 8440942-8440942 on Build GRCh38
5658469 c.335C>T p.A112V Chr.4: 8441157-8441157 on Build GRCh38
6435716 c.148G>A p.A50T Chr.4: 8440970-8440970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894171272 Chr.4: 8440913-8440913 on Build GRCh38
rs748917670 Chr.4: 8441039-8441039 on Build GRCh38
rs929858516 Chr.4: 8440859-8440859 on Build GRCh38
rs758774873 Chr.4: 8440938-8440938 on Build GRCh38
rs907119402 Chr.4: 8440718-8440718 on Build GRCh38
rs568852952 Chr.4: 8440753-8440753 on Build GRCh38
rs974469579 Chr.4: 8441002-8441002 on Build GRCh38
rs1027805819 Chr.4: 8440878-8440878 on Build GRCh38
rs774200820 Chr.4: 8440770-8440770 on Build GRCh38
rs547899288 Chr.4: 8440947-8440947 on Build GRCh38
rs137937521 Chr.4: 8440936-8440936 on Build GRCh38
rs939038418 Chr.4: 8440846-8440846 on Build GRCh38
rs184964039 Chr.4: 8441061-8441061 on Build GRCh38
rs768450088 Chr.4: 8441048-8441048 on Build GRCh38
rs538138449 Chr.4: 8441146-8441146 on Build GRCh38
rs1053431548 Chr.4: 8440700-8440700 on Build GRCh38
rs564114043 Chr.4: 8441055-8441055 on Build GRCh38
rs766147645 Chr.4: 8440842-8440842 on Build GRCh38
rs1018518544 Chr.4: 8440948-8440948 on Build GRCh38
rs761804350 Chr.4: 8440882-8440882 on Build GRCh38
rs1033997156 Chr.4: 8440750-8440750 on Build GRCh38
rs977038786 Chr.4: 8440794-8440794 on Build GRCh38
rs533050698 Chr.4: 8440966-8440966 on Build GRCh38
rs928501910 Chr.4: 8441068-8441068 on Build GRCh38
rs1002709191 Chr.4: 8440727-8440727 on Build GRCh38
rs1042434906 Chr.4: 8440714-8440714 on Build GRCh38
rs1011657696 Chr.4: 8440925-8440925 on Build GRCh38
rs1033945166 Chr.4: 8440740-8440740 on Build GRCh38
rs565278788 Chr.4: 8440811-8440811 on Build GRCh38
rs978657744 Chr.4: 8440804-8440804 on Build GRCh38
rs778924433 Chr.4: 8440928-8440928 on Build GRCh38
rs983577868 Chr.4: 8440773-8440773 on Build GRCh38
rs550198706 Chr.4: 8440701-8440701 on Build GRCh38
rs959493705 Chr.4: 8440741-8440741 on Build GRCh38
rs560407049 Chr.4: 8440963-8440963 on Build GRCh38
rs866002811 Chr.4: 8441109-8441109 on Build GRCh38
rs1055625929 Chr.4: 8440898-8440898 on Build GRCh38
rs760517125 Chr.4: 8440812-8440812 on Build GRCh38
rs768774841 Chr.4: 8440699-8440699 on Build GRCh38
rs915822512 Chr.4: 8441136-8441136 on Build GRCh38
rs968285397 Chr.4: 8440796-8440796 on Build GRCh38
rs543494278 Chr.4: 8440807-8440807 on Build GRCh38
rs1047011725 Chr.4: 8440705-8440705 on Build GRCh38
rs762808462 Chr.4: 8441083-8441083 on Build GRCh38
rs751244950 Chr.4: 8441143-8441143 on Build GRCh38
rs754040852 Chr.4: 8440863-8440863 on Build GRCh38
rs942663981 Chr.4: 8440833-8440833 on Build GRCh38
rs778957821 Chr.4: 8440853-8440853 on Build GRCh38
rs1047344300 Chr.4: 8440870-8440870 on Build GRCh38
rs1011312168 Chr.4: 8440760-8440760 on Build GRCh38
rs771835830 Chr.4: 8441072-8441072 on Build GRCh38
rs995000711 Chr.4: 8440754-8440754 on Build GRCh38
rs995116528 Chr.4: 8440875-8440875 on Build GRCh38
rs966082695 Chr.4: 8440780-8440780 on Build GRCh38
rs773190135 Chr.4: 8441080-8441080 on Build GRCh38
rs955495737 Chr.4: 8440939-8440939 on Build GRCh38
rs895392489 Chr.4: 8440873-8440873 on Build GRCh38
rs771312952 Chr.4: 8441016-8441016 on Build GRCh38
rs911238388 Chr.4: 8440832-8440832 on Build GRCh38
rs757845577 Chr.4: 8440804-8440805 on Build GRCh38
rs964078370 Chr.4: 8440984-8440984 on Build GRCh38
rs778350913 Chr.4: 8440956-8440956 on Build GRCh38
rs1009354716 Chr.4: 8440710-8440710 on Build GRCh38
rs893556577 Chr.4: 8440708-8440708 on Build GRCh38
rs1021775650 Chr.4: 8440788-8440788 on Build GRCh38
rs760581652 Chr.4: 8441091-8441091 on Build GRCh38
rs540073637 Chr.4: 8441095-8441095 on Build GRCh38
rs894117569 Chr.4: 8440752-8440752 on Build GRCh38
rs939131288 Chr.4: 8441099-8441099 on Build GRCh38
rs796210014 Chr.4: 8440773-8440773 on Build GRCh38
rs776660551 Chr.4: 8440769-8440769 on Build GRCh38
rs929778714 Chr.4: 8440698-8440698 on Build GRCh38
rs1008512865 Chr.4: 8440940-8440940 on Build GRCh38
rs974207302 Chr.4: 8440836-8440836 on Build GRCh38
rs974714975 Chr.4: 8440974-8440974 on Build GRCh38
rs987026826 Chr.4: 8440826-8440826 on Build GRCh38
rs927738808 Chr.4: 8441093-8441093 on Build GRCh38
rs919901636 Chr.4: 8441012-8441012 on Build GRCh38
rs920000715 Chr.4: 8440851-8440851 on Build GRCh38
rs759257986 Chr.4: 8441116-8441116 on Build GRCh38
rs758445075 Chr.4: 8440929-8440929 on Build GRCh38
rs576395482 Chr.4: 8440775-8440775 on Build GRCh38
rs752952184 Chr.4: 8440915-8440915 on Build GRCh38
rs759705526 Chr.4: 8441127-8441127 on Build GRCh38
rs781458066 Chr.4: 8441023-8441023 on Build GRCh38
rs989382425 Chr.4: 8441059-8441059 on Build GRCh38
rs561376380 Chr.4: 8440720-8440720 on Build GRCh38
rs755237936 Chr.4: 8440871-8440871 on Build GRCh38
rs527445129 Chr.4: 8441001-8441001 on Build GRCh38
rs750302647 Chr.4: 8441017-8441017 on Build GRCh38
rs902940185 Chr.4: 8440934-8440934 on Build GRCh38
rs938615961 Chr.4: 8440891-8440891 on Build GRCh38
rs766169556 Chr.4: 8441101-8441101 on Build GRCh38

View Full Product Details