Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8449804-8450318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT44
Entrez Gene ID
152992
Gene Name
tRNA methyltransferase 44 homolog (S. cerevisiae)
Gene Aliases
C4orf23, METTL19, TRM44
UniGene
Hs.566191
Gene Chromosome Location
Chr.4: 8440774-8512531 on Build GRCh38
Transcripts
NM_152544.2 XM_005247950.3 XM_011513406.2 XM_011513407.2 XM_011513408.1 XM_011513409.2 XM_011513410.2 XM_017007811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5673215 c.168C>T p.I56I Chr.4: 8449825-8449825 on Build GRCh38
5673214 c.891C>T p.I297I Chr.4: 8449825-8449825 on Build GRCh38
5636932 c.165C>A p.L55L Chr.4: 8449822-8449822 on Build GRCh38
6361708 c.941A>T p.K314M Chr.4: 8449875-8449875 on Build GRCh38
5636931 c.888C>A p.L296L Chr.4: 8449822-8449822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370167389 Chr.4: 8450009-8450009 on Build GRCh38
rs749712570 Chr.4: 8449930-8449934 on Build GRCh38
rs978012053 Chr.4: 8449945-8449945 on Build GRCh38
rs1045481785 Chr.4: 8450102-8450102 on Build GRCh38
rs780728114 Chr.4: 8449872-8449872 on Build GRCh38
rs200112885 Chr.4: 8449949-8449949 on Build GRCh38
rs878957980 Chr.4: 8449956-8449957 on Build GRCh38
rs73211378 Chr.4: 8450229-8450229 on Build GRCh38
rs1002106340 Chr.4: 8449852-8449852 on Build GRCh38
rs879082918 Chr.4: 8449953-8449953 on Build GRCh38
rs575266276 Chr.4: 8450168-8450168 on Build GRCh38
rs957691869 Chr.4: 8449885-8449885 on Build GRCh38
rs923413041 Chr.4: 8450031-8450031 on Build GRCh38
rs538900924 Chr.4: 8449995-8449995 on Build GRCh38
rs551535320 Chr.4: 8450001-8450001 on Build GRCh38
rs377180019 Chr.4: 8449934-8449934 on Build GRCh38
rs113205933 Chr.4: 8450082-8450082 on Build GRCh38
rs1046686564 Chr.4: 8450121-8450121 on Build GRCh38
rs898727955 Chr.4: 8450142-8450142 on Build GRCh38
rs919352416 Chr.4: 8450109-8450109 on Build GRCh38
rs1055041386 Chr.4: 8450175-8450175 on Build GRCh38
rs201131284 Chr.4: 8449954-8449954 on Build GRCh38
rs759657898 Chr.4: 8450100-8450100 on Build GRCh38
rs1020595534 Chr.4: 8450190-8450190 on Build GRCh38
rs1021133035 Chr.4: 8449903-8449903 on Build GRCh38
rs369885914 Chr.4: 8449944-8449944 on Build GRCh38
rs921835052 Chr.4: 8449918-8449918 on Build GRCh38
rs975447677 Chr.4: 8449897-8449897 on Build GRCh38
rs866443648 Chr.4: 8449965-8449965 on Build GRCh38
rs912516060 Chr.4: 8449862-8449872 on Build GRCh38
rs925294698 Chr.4: 8450285-8450285 on Build GRCh38
rs745915221 Chr.4: 8449950-8449960 on Build GRCh38
rs73211375 Chr.4: 8449832-8449832 on Build GRCh38
rs554579483 Chr.4: 8450123-8450123 on Build GRCh38
rs540496636 Chr.4: 8450255-8450255 on Build GRCh38
rs954987584 Chr.4: 8450042-8450042 on Build GRCh38
rs893695665 Chr.4: 8450176-8450176 on Build GRCh38
rs62287383 Chr.4: 8450073-8450073 on Build GRCh38
rs151141713 Chr.4: 8450183-8450183 on Build GRCh38
rs373326908 Chr.4: 8449944-8449944 on Build GRCh38
rs975395439 Chr.4: 8450278-8450278 on Build GRCh38
rs778179208 Chr.4: 8449929-8449930 on Build GRCh38
rs536370576 Chr.4: 8450114-8450114 on Build GRCh38
rs377530926 Chr.4: 8449985-8449985 on Build GRCh38
rs56676930 Chr.4: 8449953-8449954 on Build GRCh38
rs746127004 Chr.4: 8449951-8449952 on Build GRCh38
rs189786402 Chr.4: 8450021-8450021 on Build GRCh38
rs749051136 Chr.4: 8450226-8450226 on Build GRCh38
rs773439992 Chr.4: 8450244-8450244 on Build GRCh38
rs1033366137 Chr.4: 8449876-8449876 on Build GRCh38
rs867598157 Chr.4: 8449960-8449960 on Build GRCh38
rs373530850 Chr.4: 8449949-8449949 on Build GRCh38
rs182350895 Chr.4: 8450107-8450107 on Build GRCh38
rs986912479 Chr.4: 8449858-8449858 on Build GRCh38
rs754608030 Chr.4: 8449928-8449929 on Build GRCh38
rs202074531 Chr.4: 8449959-8449959 on Build GRCh38
rs370642267 Chr.4: 8449939-8449939 on Build GRCh38
rs561941451 Chr.4: 8450289-8450289 on Build GRCh38
rs906459524 Chr.4: 8449840-8449840 on Build GRCh38
rs534552153 Chr.4: 8450039-8450039 on Build GRCh38
rs572879785 Chr.4: 8450232-8450232 on Build GRCh38
rs540935559 Chr.4: 8449847-8449847 on Build GRCh38
rs775466390 Chr.4: 8449934-8449934 on Build GRCh38
rs386671369 Chr.4: 8449998-8450001 on Build GRCh38
rs1002163663 Chr.4: 8450124-8450124 on Build GRCh38
rs997932759 Chr.4: 8450218-8450218 on Build GRCh38
rs760906386 Chr.4: 8449937-8449939 on Build GRCh38
rs185822187 Chr.4: 8449892-8449892 on Build GRCh38
rs761311536 Chr.4: 8449949-8449950 on Build GRCh38
rs35973516 Chr.4: 8449961-8449962 on Build GRCh38
rs935929903 Chr.4: 8450026-8450026 on Build GRCh38
rs910806871 Chr.4: 8450234-8450234 on Build GRCh38
rs550606251 Chr.4: 8449998-8449998 on Build GRCh38
rs772128524 Chr.4: 8449934-8449939 on Build GRCh38
rs562531628 Chr.4: 8449999-8449999 on Build GRCh38
rs769750640 Chr.4: 8449926-8449926 on Build GRCh38
rs966876302 Chr.4: 8450216-8450216 on Build GRCh38
rs546754700 Chr.4: 8450059-8450059 on Build GRCh38
rs533474285 Chr.4: 8450000-8450000 on Build GRCh38
rs756750049 Chr.4: 8449839-8449839 on Build GRCh38

View Full Product Details