Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81251203-81251708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7201609 c.99C>A p.N33K Chr.5: 81251231-81251231 on Build GRCh38
5808581 c.105G>C p.Q35H Chr.5: 81251237-81251237 on Build GRCh38
6303266 c.113G>T p.C38F Chr.5: 81251245-81251245 on Build GRCh38
4904294 c.126G>A p.R42R Chr.5: 81251258-81251258 on Build GRCh38
4895083 c.91C>T p.L31L Chr.5: 81251223-81251223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749939043 Chr.5: 81251320-81251320 on Build GRCh38
rs373137473 Chr.5: 81251231-81251231 on Build GRCh38
rs1003526377 Chr.5: 81251614-81251614 on Build GRCh38
rs772325145 Chr.5: 81251256-81251256 on Build GRCh38
rs373158462 Chr.5: 81251325-81251325 on Build GRCh38
rs942412528 Chr.5: 81251532-81251532 on Build GRCh38
rs555034687 Chr.5: 81251674-81251674 on Build GRCh38
rs929793260 Chr.5: 81251574-81251574 on Build GRCh38
rs377452879 Chr.5: 81251321-81251321 on Build GRCh38
rs931004632 Chr.5: 81251266-81251266 on Build GRCh38
rs1031912287 Chr.5: 81251406-81251406 on Build GRCh38
rs34345589 Chr.5: 81251651-81251652 on Build GRCh38
rs747057295 Chr.5: 81251262-81251262 on Build GRCh38
rs535448521 Chr.5: 81251653-81251653 on Build GRCh38
rs549949512 Chr.5: 81251581-81251581 on Build GRCh38
rs570621839 Chr.5: 81251544-81251544 on Build GRCh38
rs749932126 Chr.5: 81251225-81251225 on Build GRCh38
rs767191068 Chr.5: 81251316-81251316 on Build GRCh38
rs141935817 Chr.5: 81251257-81251257 on Build GRCh38
rs901995019 Chr.5: 81251665-81251665 on Build GRCh38
rs978751041 Chr.5: 81251420-81251420 on Build GRCh38
rs774133863 Chr.5: 81251294-81251294 on Build GRCh38
rs749410402 Chr.5: 81251458-81251458 on Build GRCh38
rs968367153 Chr.5: 81251412-81251412 on Build GRCh38
rs970510837 Chr.5: 81251489-81251489 on Build GRCh38
rs769025358 Chr.5: 81251480-81251480 on Build GRCh38
rs569752200 Chr.5: 81251639-81251639 on Build GRCh38
rs990146989 Chr.5: 81251457-81251457 on Build GRCh38
rs1022966010 Chr.5: 81251485-81251485 on Build GRCh38
rs548278772 Chr.5: 81251433-81251433 on Build GRCh38
rs562033855 Chr.5: 81251513-81251513 on Build GRCh38
rs533292553 Chr.5: 81251456-81251456 on Build GRCh38
rs895270465 Chr.5: 81251617-81251617 on Build GRCh38
rs138839566 Chr.5: 81251222-81251222 on Build GRCh38
rs939900129 Chr.5: 81251516-81251516 on Build GRCh38
rs886446804 Chr.5: 81251309-81251309 on Build GRCh38
rs75970007 Chr.5: 81251267-81251267 on Build GRCh38
rs1019502728 Chr.5: 81251372-81251372 on Build GRCh38
rs755356788 Chr.5: 81251242-81251242 on Build GRCh38
rs762948369 Chr.5: 81251274-81251274 on Build GRCh38
rs999044596 Chr.5: 81251384-81251384 on Build GRCh38
rs981571239 Chr.5: 81251492-81251492 on Build GRCh38
rs943762038 Chr.5: 81251622-81251622 on Build GRCh38
rs776537512 Chr.5: 81251325-81251326 on Build GRCh38
rs531627306 Chr.5: 81251354-81251354 on Build GRCh38
rs199576376 Chr.5: 81251673-81251673 on Build GRCh38
rs990978695 Chr.5: 81251345-81251345 on Build GRCh38
rs55727032 Chr.5: 81251487-81251487 on Build GRCh38
rs368914826 Chr.5: 81251226-81251226 on Build GRCh38
rs928442232 Chr.5: 81251493-81251493 on Build GRCh38
rs780505629 Chr.5: 81251232-81251232 on Build GRCh38
rs539634331 Chr.5: 81251545-81251545 on Build GRCh38
rs910997740 Chr.5: 81251526-81251526 on Build GRCh38
rs202021346 Chr.5: 81251233-81251233 on Build GRCh38
rs907097325 Chr.5: 81251613-81251614 on Build GRCh38
rs1040822904 Chr.5: 81251652-81251652 on Build GRCh38
rs748332374 Chr.5: 81251250-81251250 on Build GRCh38
rs779296426 Chr.5: 81251249-81251249 on Build GRCh38
rs902027598 Chr.5: 81251379-81251379 on Build GRCh38
rs972638919 Chr.5: 81251520-81251520 on Build GRCh38
rs768344797 Chr.5: 81251285-81251285 on Build GRCh38
rs924180072 Chr.5: 81251422-81251422 on Build GRCh38
rs889473102 Chr.5: 81251582-81251582 on Build GRCh38
rs769904311 Chr.5: 81251265-81251265 on Build GRCh38
rs545932325 Chr.5: 81251302-81251302 on Build GRCh38
rs562637382 Chr.5: 81251324-81251324 on Build GRCh38
rs755420070 Chr.5: 81251326-81251326 on Build GRCh38
rs1034972240 Chr.5: 81251614-81251617 on Build GRCh38
rs554828214 Chr.5: 81251542-81251542 on Build GRCh38

View Full Product Details