Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132694000-132694513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3A
Entrez Gene ID
11127
Gene Name
kinesin family member 3A
Gene Aliases
FLA10, KLP-20
UniGene
Hs.43670
Gene Chromosome Location
Chr.5: 132688808-132737633 on Build GRCh38
Transcripts
NM_001300791.1 NM_001300792.1 NM_007054.6 XM_006714526.3 XM_017008995.1 XM_017008996.1

SNP Detail

SNP ID Chromosome Location
rs761518148 Chr.5: 132694444-132694444 on Build GRCh38
rs1002062070 Chr.5: 132694085-132694085 on Build GRCh38
rs112058399 Chr.5: 132694116-132694116 on Build GRCh38
rs891355975 Chr.5: 132694092-132694092 on Build GRCh38
rs1002516987 Chr.5: 132694238-132694238 on Build GRCh38
rs905946924 Chr.5: 132694077-132694077 on Build GRCh38
rs569666136 Chr.5: 132694212-132694212 on Build GRCh38
rs949283727 Chr.5: 132694490-132694490 on Build GRCh38
rs549830857 Chr.5: 132694130-132694130 on Build GRCh38
rs893267221 Chr.5: 132694283-132694283 on Build GRCh38
rs966281110 Chr.5: 132694325-132694325 on Build GRCh38
rs921638059 Chr.5: 132694163-132694163 on Build GRCh38
rs145403816 Chr.5: 132694112-132694112 on Build GRCh38
rs940537275 Chr.5: 132694176-132694176 on Build GRCh38
rs768197971 Chr.5: 132694330-132694330 on Build GRCh38
rs1040358113 Chr.5: 132694411-132694411 on Build GRCh38
rs1036241783 Chr.5: 132694182-132694182 on Build GRCh38
rs942990189 Chr.5: 132694398-132694398 on Build GRCh38
rs1001611350 Chr.5: 132694278-132694278 on Build GRCh38
rs979496213 Chr.5: 132694331-132694331 on Build GRCh38
rs777445965 Chr.5: 132694196-132694196 on Build GRCh38
rs535584587 Chr.5: 132694230-132694230 on Build GRCh38
rs975740102 Chr.5: 132694161-132694161 on Build GRCh38
rs923991083 Chr.5: 132694350-132694350 on Build GRCh38
rs150074408 Chr.5: 132694054-132694054 on Build GRCh38
rs1020553488 Chr.5: 132694142-132694142 on Build GRCh38
rs560797037 Chr.5: 132694437-132694437 on Build GRCh38
rs148412631 Chr.5: 132694354-132694354 on Build GRCh38
rs940500369 Chr.5: 132694377-132694377 on Build GRCh38
rs909017617 Chr.5: 132694174-132694174 on Build GRCh38
rs905894494 Chr.5: 132694267-132694267 on Build GRCh38
rs1024308556 Chr.5: 132694066-132694066 on Build GRCh38
rs564422478 Chr.5: 132694091-132694091 on Build GRCh38
rs776103248 Chr.5: 132694343-132694343 on Build GRCh38
rs748318132 Chr.5: 132694088-132694088 on Build GRCh38
rs898698286 Chr.5: 132694412-132694412 on Build GRCh38
rs1020501070 Chr.5: 132694324-132694324 on Build GRCh38
rs1033531919 Chr.5: 132694280-132694280 on Build GRCh38
rs1010830112 Chr.5: 132694286-132694286 on Build GRCh38
rs776519953 Chr.5: 132694363-132694363 on Build GRCh38
rs565591584 Chr.5: 132694386-132694386 on Build GRCh38
rs931732639 Chr.5: 132694169-132694169 on Build GRCh38
rs984976387 Chr.5: 132694170-132694170 on Build GRCh38
rs1018747189 Chr.5: 132694177-132694177 on Build GRCh38
rs908958302 Chr.5: 132694376-132694376 on Build GRCh38
rs1060980 Chr.5: 132694100-132694100 on Build GRCh38
rs761049387 Chr.5: 132694311-132694311 on Build GRCh38
rs965669558 Chr.5: 132694144-132694144 on Build GRCh38
rs953176577 Chr.5: 132694359-132694359 on Build GRCh38
rs896414284 Chr.5: 132694201-132694201 on Build GRCh38
rs989056062 Chr.5: 132694140-132694140 on Build GRCh38
rs931429535 Chr.5: 132694449-132694449 on Build GRCh38
rs190048812 Chr.5: 132694026-132694026 on Build GRCh38
rs1035619600 Chr.5: 132694262-132694262 on Build GRCh38

View Full Product Details