Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160251927-160252463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJL
Entrez Gene ID
79616
Gene Name
cyclin J like
Gene Aliases
-
UniGene
Hs.14070
Gene Chromosome Location
Chr.5: 160251652-160339592 on Build GRCh38
Transcripts
NM_001308173.1 NM_024565.6 NR_131769.1 XM_006714917.3 XM_011534646.2 XM_011534647.2 XM_017009846.1 XM_017009847.1 XM_017009848.1 XM_017009849.1

SNP Detail

SNP ID Chromosome Location
rs1022842354 Chr.5: 160252284-160252284 on Build GRCh38
rs527564718 Chr.5: 160252351-160252351 on Build GRCh38
rs955709016 Chr.5: 160252337-160252337 on Build GRCh38
rs924305516 Chr.5: 160252122-160252122 on Build GRCh38
rs921579865 Chr.5: 160252307-160252307 on Build GRCh38
rs1045986087 Chr.5: 160252441-160252441 on Build GRCh38
rs1054171077 Chr.5: 160252154-160252154 on Build GRCh38
rs565622307 Chr.5: 160251989-160251989 on Build GRCh38
rs1036988694 Chr.5: 160252217-160252217 on Build GRCh38
rs967546582 Chr.5: 160252218-160252218 on Build GRCh38
rs987040857 Chr.5: 160251956-160251956 on Build GRCh38
rs575744375 Chr.5: 160252000-160252000 on Build GRCh38
rs576950479 Chr.5: 160251971-160251971 on Build GRCh38
rs907617865 Chr.5: 160252361-160252361 on Build GRCh38
rs1019829556 Chr.5: 160252257-160252257 on Build GRCh38
rs1027493988 Chr.5: 160252019-160252019 on Build GRCh38
rs532409552 Chr.5: 160252135-160252135 on Build GRCh38
rs191365321 Chr.5: 160252305-160252305 on Build GRCh38
rs924524609 Chr.5: 160252322-160252322 on Build GRCh38
rs116679396 Chr.5: 160252338-160252338 on Build GRCh38
rs921632055 Chr.5: 160251991-160251991 on Build GRCh38
rs975702496 Chr.5: 160252290-160252290 on Build GRCh38
rs1012732780 Chr.5: 160252281-160252281 on Build GRCh38
rs893159612 Chr.5: 160252167-160252167 on Build GRCh38
rs1015572062 Chr.5: 160252003-160252003 on Build GRCh38
rs934328534 Chr.5: 160252150-160252150 on Build GRCh38
rs967976836 Chr.5: 160251954-160251954 on Build GRCh38
rs964280687 Chr.5: 160252275-160252275 on Build GRCh38
rs981099077 Chr.5: 160252111-160252111 on Build GRCh38
rs985591141 Chr.5: 160252422-160252422 on Build GRCh38
rs914323691 Chr.5: 160252169-160252169 on Build GRCh38
rs944220425 Chr.5: 160252442-160252442 on Build GRCh38
rs560056450 Chr.5: 160252207-160252207 on Build GRCh38
rs907134031 Chr.5: 160252055-160252055 on Build GRCh38
rs991751766 Chr.5: 160252040-160252040 on Build GRCh38
rs912764285 Chr.5: 160252442-160252446 on Build GRCh38
rs546717287 Chr.5: 160252063-160252063 on Build GRCh38
rs552006835 Chr.5: 160252181-160252181 on Build GRCh38
rs771372261 Chr.5: 160252200-160252203 on Build GRCh38
rs17057564 Chr.5: 160252192-160252192 on Build GRCh38
rs1001459335 Chr.5: 160252081-160252081 on Build GRCh38
rs561396009 Chr.5: 160252015-160252015 on Build GRCh38
rs3749798 Chr.5: 160251988-160251988 on Build GRCh38
rs150568515 Chr.5: 160252052-160252052 on Build GRCh38
rs940510307 Chr.5: 160252419-160252419 on Build GRCh38
rs188618874 Chr.5: 160251990-160251990 on Build GRCh38
rs538011262 Chr.5: 160252324-160252324 on Build GRCh38
rs940533966 Chr.5: 160252026-160252026 on Build GRCh38
rs372227502 Chr.5: 160252375-160252375 on Build GRCh38
rs1045629303 Chr.5: 160252054-160252054 on Build GRCh38
rs551978293 Chr.5: 160252352-160252352 on Build GRCh38
rs1027049270 Chr.5: 160252224-160252224 on Build GRCh38
rs1008473068 Chr.5: 160252208-160252208 on Build GRCh38
rs995917827 Chr.5: 160252223-160252223 on Build GRCh38
rs534324366 Chr.5: 160252248-160252248 on Build GRCh38
rs372563844 Chr.5: 160252006-160252006 on Build GRCh38
rs139894583 Chr.5: 160252104-160252104 on Build GRCh38
rs1008813674 Chr.5: 160252246-160252246 on Build GRCh38

View Full Product Details