Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160253172-160253667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJL
Entrez Gene ID
79616
Gene Name
cyclin J like
Gene Aliases
-
UniGene
Hs.14070
Gene Chromosome Location
Chr.5: 160251652-160339592 on Build GRCh38
Transcripts
NM_001308173.1 NM_024565.6 NR_131769.1 XM_006714917.3 XM_011534646.2 XM_011534647.2 XM_017009846.1 XM_017009847.1 XM_017009848.1 XM_017009849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4964326 c.1162T>C p.L388L Chr.5: 160253524-160253524 on Build GRCh38
6623051 c.1247C>T p.T416I Chr.5: 160253439-160253439 on Build GRCh38
1435677 c.1236C>T p.C412C Chr.5: 160253450-160253450 on Build GRCh38
5068606 c.1287_1288insC p.T430fs*5 Chr.5: 160253398-160253399 on Build GRCh38
136509 c.1186C>T p.P396S Chr.5: 160253500-160253500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755201303 Chr.5: 160253455-160253455 on Build GRCh38
rs765269177 Chr.5: 160253345-160253347 on Build GRCh38
rs762420687 Chr.5: 160253408-160253408 on Build GRCh38
rs764300195 Chr.5: 160253600-160253600 on Build GRCh38
rs191171075 Chr.5: 160253247-160253247 on Build GRCh38
rs756009636 Chr.5: 160253373-160253373 on Build GRCh38
rs779624773 Chr.5: 160253627-160253627 on Build GRCh38
rs753664479 Chr.5: 160253510-160253510 on Build GRCh38
rs527440214 Chr.5: 160253475-160253475 on Build GRCh38
rs370421409 Chr.5: 160253593-160253593 on Build GRCh38
rs199879462 Chr.5: 160253509-160253509 on Build GRCh38
rs778551885 Chr.5: 160253561-160253561 on Build GRCh38
rs765219491 Chr.5: 160253578-160253578 on Build GRCh38
rs200717032 Chr.5: 160253544-160253544 on Build GRCh38
rs749845640 Chr.5: 160253491-160253491 on Build GRCh38
rs774961375 Chr.5: 160253572-160253572 on Build GRCh38
rs944012205 Chr.5: 160253271-160253271 on Build GRCh38
rs767353761 Chr.5: 160253422-160253422 on Build GRCh38
rs375350128 Chr.5: 160253329-160253329 on Build GRCh38
rs183560299 Chr.5: 160253411-160253411 on Build GRCh38
rs767452083 Chr.5: 160253360-160253360 on Build GRCh38
rs375710423 Chr.5: 160253394-160253394 on Build GRCh38
rs762090969 Chr.5: 160253298-160253298 on Build GRCh38
rs779046128 Chr.5: 160253382-160253382 on Build GRCh38
rs576946259 Chr.5: 160253326-160253326 on Build GRCh38
rs373648473 Chr.5: 160253534-160253534 on Build GRCh38
rs763986715 Chr.5: 160253430-160253430 on Build GRCh38
rs1031976654 Chr.5: 160253450-160253450 on Build GRCh38
rs921260841 Chr.5: 160253456-160253456 on Build GRCh38
rs759656265 Chr.5: 160253585-160253585 on Build GRCh38
rs1003780227 Chr.5: 160253377-160253377 on Build GRCh38
rs758569380 Chr.5: 160253473-160253474 on Build GRCh38
rs754310300 Chr.5: 160253603-160253603 on Build GRCh38
rs760394370 Chr.5: 160253573-160253573 on Build GRCh38
rs377553131 Chr.5: 160253396-160253396 on Build GRCh38
rs779603501 Chr.5: 160253613-160253613 on Build GRCh38
rs377363287 Chr.5: 160253592-160253592 on Build GRCh38
rs752501513 Chr.5: 160253423-160253423 on Build GRCh38
rs750482616 Chr.5: 160253434-160253434 on Build GRCh38
rs377179668 Chr.5: 160253636-160253636 on Build GRCh38
rs368325119 Chr.5: 160253341-160253341 on Build GRCh38
rs371574226 Chr.5: 160253507-160253507 on Build GRCh38
rs770276155 Chr.5: 160253405-160253405 on Build GRCh38
rs751379130 Chr.5: 160253347-160253347 on Build GRCh38
rs751707240 Chr.5: 160253519-160253519 on Build GRCh38
rs372596521 Chr.5: 160253343-160253343 on Build GRCh38
rs1006113971 Chr.5: 160253447-160253447 on Build GRCh38
rs769691898 Chr.5: 160253639-160253639 on Build GRCh38
rs900805034 Chr.5: 160253414-160253414 on Build GRCh38
rs540158833 Chr.5: 160253229-160253229 on Build GRCh38
rs374113107 Chr.5: 160253579-160253579 on Build GRCh38
rs746572826 Chr.5: 160253632-160253632 on Build GRCh38
rs755182751 Chr.5: 160253521-160253521 on Build GRCh38
rs775331235 Chr.5: 160253506-160253506 on Build GRCh38
rs541758503 Chr.5: 160253409-160253409 on Build GRCh38
rs375241736 Chr.5: 160253609-160253609 on Build GRCh38
rs747988573 Chr.5: 160253638-160253638 on Build GRCh38
rs984575156 Chr.5: 160253484-160253484 on Build GRCh38
rs373229199 Chr.5: 160253359-160253359 on Build GRCh38
rs745991868 Chr.5: 160253385-160253385 on Build GRCh38
rs984318616 Chr.5: 160253288-160253288 on Build GRCh38
rs942537903 Chr.5: 160253556-160253556 on Build GRCh38
rs778185097 Chr.5: 160253481-160253481 on Build GRCh38
rs759256343 Chr.5: 160253504-160253504 on Build GRCh38
rs771632303 Chr.5: 160253555-160253555 on Build GRCh38
rs752778414 Chr.5: 160253448-160253449 on Build GRCh38
rs141268151 Chr.5: 160253543-160253543 on Build GRCh38
rs965274125 Chr.5: 160253454-160253454 on Build GRCh38
rs187903121 Chr.5: 160253574-160253574 on Build GRCh38
rs374780099 Chr.5: 160253446-160253446 on Build GRCh38
rs1035375381 Chr.5: 160253194-160253194 on Build GRCh38
rs747381182 Chr.5: 160253568-160253578 on Build GRCh38
rs976761167 Chr.5: 160253293-160253293 on Build GRCh38
rs761737257 Chr.5: 160253577-160253577 on Build GRCh38
rs199835106 Chr.5: 160253503-160253503 on Build GRCh38
rs553816630 Chr.5: 160253261-160253261 on Build GRCh38
rs770516651 Chr.5: 160253492-160253492 on Build GRCh38
rs781619971 Chr.5: 160253459-160253459 on Build GRCh38
rs958436120 Chr.5: 160253202-160253202 on Build GRCh38
rs201164652 Chr.5: 160253395-160253395 on Build GRCh38
rs991224036 Chr.5: 160253204-160253204 on Build GRCh38
rs373108109 Chr.5: 160253564-160253564 on Build GRCh38
rs533035690 Chr.5: 160253614-160253614 on Build GRCh38
rs1048547183 Chr.5: 160253315-160253315 on Build GRCh38
rs371210198 Chr.5: 160253442-160253442 on Build GRCh38
rs756062568 Chr.5: 160253429-160253429 on Build GRCh38
rs765432135 Chr.5: 160253310-160253310 on Build GRCh38
rs940157997 Chr.5: 160253518-160253518 on Build GRCh38
rs771724000 Chr.5: 160253568-160253568 on Build GRCh38
rs759284864 Chr.5: 160253418-160253418 on Build GRCh38
rs753076404 Chr.5: 160253536-160253536 on Build GRCh38
rs757793736 Chr.5: 160253624-160253624 on Build GRCh38
rs780291160 Chr.5: 160253393-160253393 on Build GRCh38
rs748522355 Chr.5: 160253465-160253465 on Build GRCh38
rs772768116 Chr.5: 160253407-160253407 on Build GRCh38
rs756373783 Chr.5: 160253597-160253597 on Build GRCh38
rs777995694 Chr.5: 160253479-160253480 on Build GRCh38
rs901110449 Chr.5: 160253565-160253565 on Build GRCh38
rs921293370 Chr.5: 160253300-160253300 on Build GRCh38
rs73308655 Chr.5: 160253221-160253221 on Build GRCh38
rs574688763 Chr.5: 160253226-160253226 on Build GRCh38
rs982326910 Chr.5: 160253486-160253486 on Build GRCh38
rs369592831 Chr.5: 160253512-160253512 on Build GRCh38
rs758595466 Chr.5: 160253388-160253388 on Build GRCh38
rs780126271 Chr.5: 160253445-160253445 on Build GRCh38
rs939603948 Chr.5: 160253559-160253559 on Build GRCh38
rs954058104 Chr.5: 160253467-160253467 on Build GRCh38
rs557707300 Chr.5: 160253237-160253237 on Build GRCh38
rs772605627 Chr.5: 160253339-160253339 on Build GRCh38
rs993841807 Chr.5: 160253262-160253262 on Build GRCh38
rs756394538 Chr.5: 160253537-160253537 on Build GRCh38
rs773096728 Chr.5: 160253645-160253645 on Build GRCh38

View Full Product Details