Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26234504-26234999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H1D
Entrez Gene ID
3007
Gene Name
histone cluster 1, H1d
Gene Aliases
H1.3, H1D, H1F3, H1s-2
UniGene
Hs.136857
Gene Chromosome Location
Chr.6: 26234212-26234988 on Build GRCh38
Transcripts
NM_005320.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5404962 c.361G>A p.A121T Chr.6: 26234573-26234573 on Build GRCh38
6241278 c.137C>A p.T46N Chr.6: 26234797-26234797 on Build GRCh38
5404964 c.50A>T p.K17I Chr.6: 26234884-26234884 on Build GRCh38
6407142 c.202G>T p.A68S Chr.6: 26234732-26234732 on Build GRCh38
6680195 c.54A>G p.T18T Chr.6: 26234880-26234880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765867901 Chr.6: 26234688-26234688 on Build GRCh38
rs779392662 Chr.6: 26234964-26234964 on Build GRCh38
rs768617477 Chr.6: 26234674-26234674 on Build GRCh38
rs771890248 Chr.6: 26234908-26234908 on Build GRCh38
rs376493714 Chr.6: 26234565-26234565 on Build GRCh38
rs142772662 Chr.6: 26234897-26234897 on Build GRCh38
rs369074504 Chr.6: 26234816-26234816 on Build GRCh38
rs765086445 Chr.6: 26234898-26234898 on Build GRCh38
rs781696897 Chr.6: 26234878-26234878 on Build GRCh38
rs770792589 Chr.6: 26234747-26234747 on Build GRCh38
rs750569345 Chr.6: 26234958-26234958 on Build GRCh38
rs757799635 Chr.6: 26234770-26234770 on Build GRCh38
rs139832189 Chr.6: 26234594-26234594 on Build GRCh38
rs776308039 Chr.6: 26234619-26234619 on Build GRCh38
rs778834682 Chr.6: 26234704-26234704 on Build GRCh38
rs749549303 Chr.6: 26234623-26234623 on Build GRCh38
rs774642794 Chr.6: 26234564-26234568 on Build GRCh38
rs75255708 Chr.6: 26234701-26234701 on Build GRCh38
rs201679688 Chr.6: 26234827-26234827 on Build GRCh38
rs772955921 Chr.6: 26234549-26234549 on Build GRCh38
rs759908465 Chr.6: 26234749-26234749 on Build GRCh38
rs540981031 Chr.6: 26234541-26234541 on Build GRCh38
rs745686818 Chr.6: 26234804-26234804 on Build GRCh38
rs2050950 Chr.6: 26234655-26234655 on Build GRCh38
rs201889816 Chr.6: 26234862-26234862 on Build GRCh38
rs201632466 Chr.6: 26234875-26234875 on Build GRCh38
rs754760490 Chr.6: 26234866-26234866 on Build GRCh38
rs767014847 Chr.6: 26234961-26234961 on Build GRCh38
rs143234965 Chr.6: 26234596-26234596 on Build GRCh38
rs770606111 Chr.6: 26234591-26234591 on Build GRCh38
rs986624611 Chr.6: 26234622-26234622 on Build GRCh38
rs374396004 Chr.6: 26234656-26234656 on Build GRCh38
rs780673857 Chr.6: 26234936-26234936 on Build GRCh38
rs138719852 Chr.6: 26234880-26234880 on Build GRCh38
rs749992324 Chr.6: 26234839-26234839 on Build GRCh38
rs370735023 Chr.6: 26234592-26234592 on Build GRCh38
rs767659677 Chr.6: 26234654-26234654 on Build GRCh38
rs754813347 Chr.6: 26234535-26234535 on Build GRCh38
rs775417205 Chr.6: 26234860-26234868 on Build GRCh38
rs747596738 Chr.6: 26234736-26234736 on Build GRCh38
rs200751379 Chr.6: 26234681-26234681 on Build GRCh38
rs750696083 Chr.6: 26234629-26234633 on Build GRCh38
rs768427982 Chr.6: 26234952-26234952 on Build GRCh38
rs549014505 Chr.6: 26234607-26234607 on Build GRCh38
rs778271229 Chr.6: 26234691-26234691 on Build GRCh38
rs147157401 Chr.6: 26234597-26234597 on Build GRCh38
rs200643557 Chr.6: 26234613-26234613 on Build GRCh38
rs770409593 Chr.6: 26234912-26234912 on Build GRCh38
rs2050949 Chr.6: 26234711-26234711 on Build GRCh38
rs1013776709 Chr.6: 26234546-26234546 on Build GRCh38
rs919969528 Chr.6: 26234657-26234657 on Build GRCh38
rs757285227 Chr.6: 26234925-26234925 on Build GRCh38
rs200289213 Chr.6: 26234558-26234558 on Build GRCh38
rs772093987 Chr.6: 26234738-26234738 on Build GRCh38
rs760795695 Chr.6: 26234959-26234959 on Build GRCh38
rs761990052 Chr.6: 26234595-26234596 on Build GRCh38
rs774668728 Chr.6: 26234758-26234758 on Build GRCh38
rs957420095 Chr.6: 26234948-26234948 on Build GRCh38
rs1038204406 Chr.6: 26234562-26234562 on Build GRCh38
rs375359922 Chr.6: 26234797-26234797 on Build GRCh38
rs145483692 Chr.6: 26234651-26234651 on Build GRCh38
rs890781296 Chr.6: 26234821-26234821 on Build GRCh38
rs753614104 Chr.6: 26234865-26234865 on Build GRCh38
rs772606876 Chr.6: 26234815-26234815 on Build GRCh38
rs752285481 Chr.6: 26234769-26234769 on Build GRCh38
rs755565497 Chr.6: 26234699-26234701 on Build GRCh38
rs776581922 Chr.6: 26234896-26234896 on Build GRCh38
rs747123489 Chr.6: 26234540-26234540 on Build GRCh38
rs372659859 Chr.6: 26234823-26234823 on Build GRCh38
rs766806485 Chr.6: 26234698-26234710 on Build GRCh38
rs760280711 Chr.6: 26234551-26234551 on Build GRCh38
rs377088507 Chr.6: 26234785-26234785 on Build GRCh38
rs150400552 Chr.6: 26234748-26234748 on Build GRCh38
rs769294524 Chr.6: 26234625-26234625 on Build GRCh38
rs551606847 Chr.6: 26234817-26234817 on Build GRCh38
rs754129847 Chr.6: 26234698-26234699 on Build GRCh38
rs751429232 Chr.6: 26234921-26234921 on Build GRCh38
rs749753570 Chr.6: 26234608-26234608 on Build GRCh38
rs759703508 Chr.6: 26234714-26234714 on Build GRCh38
rs749696539 Chr.6: 26234574-26234574 on Build GRCh38
rs150746370 Chr.6: 26234902-26234902 on Build GRCh38
rs774839714 Chr.6: 26234783-26234783 on Build GRCh38
rs988923936 Chr.6: 26234616-26234616 on Build GRCh38
rs754910001 Chr.6: 26234934-26234934 on Build GRCh38
rs763387017 Chr.6: 26234543-26234543 on Build GRCh38
rs774247984 Chr.6: 26234973-26234973 on Build GRCh38
rs201093098 Chr.6: 26234830-26234830 on Build GRCh38
rs147516990 Chr.6: 26234859-26234859 on Build GRCh38
rs772646340 Chr.6: 26234892-26234892 on Build GRCh38
rs769313433 Chr.6: 26234815-26234815 on Build GRCh38
rs755594152 Chr.6: 26234658-26234658 on Build GRCh38
rs371627397 Chr.6: 26234777-26234777 on Build GRCh38
rs202171620 Chr.6: 26234776-26234776 on Build GRCh38
rs746317850 Chr.6: 26234904-26234904 on Build GRCh38
rs770222362 Chr.6: 26234879-26234879 on Build GRCh38
rs767650060 Chr.6: 26234622-26234623 on Build GRCh38
rs373606239 Chr.6: 26234822-26234822 on Build GRCh38
rs751859791 Chr.6: 26234692-26234692 on Build GRCh38
rs866405895 Chr.6: 26234544-26234545 on Build GRCh38
rs372255160 Chr.6: 26234756-26234756 on Build GRCh38
rs769240912 Chr.6: 26234968-26234968 on Build GRCh38
rs779975489 Chr.6: 26234969-26234969 on Build GRCh38
rs779386969 Chr.6: 26234704-26234704 on Build GRCh38
rs199968673 Chr.6: 26234917-26234917 on Build GRCh38
rs201283893 Chr.6: 26234962-26234962 on Build GRCh38
rs530322275 Chr.6: 26234976-26234976 on Build GRCh38
rs769329952 Chr.6: 26234669-26234669 on Build GRCh38
rs749405977 Chr.6: 26234781-26234781 on Build GRCh38
rs774471687 Chr.6: 26234812-26234812 on Build GRCh38
rs374182906 Chr.6: 26234967-26234967 on Build GRCh38
rs201392898 Chr.6: 26234581-26234581 on Build GRCh38
rs951535539 Chr.6: 26234858-26234858 on Build GRCh38
rs572160234 Chr.6: 26234539-26234539 on Build GRCh38
rs1014468965 Chr.6: 26234578-26234578 on Build GRCh38
rs765137824 Chr.6: 26234916-26234916 on Build GRCh38
rs750506011 Chr.6: 26234604-26234604 on Build GRCh38
rs970772285 Chr.6: 26234930-26234930 on Build GRCh38
rs2050948 Chr.6: 26234724-26234724 on Build GRCh38
rs752526833 Chr.6: 26234538-26234538 on Build GRCh38
rs144291537 Chr.6: 26234700-26234701 on Build GRCh38
rs200830065 Chr.6: 26234725-26234725 on Build GRCh38
rs776944538 Chr.6: 26234712-26234712 on Build GRCh38
rs762573768 Chr.6: 26234819-26234819 on Build GRCh38
rs746877459 Chr.6: 26234772-26234772 on Build GRCh38
rs370746969 Chr.6: 26234687-26234687 on Build GRCh38
rs779699844 Chr.6: 26234728-26234728 on Build GRCh38
rs571636717 Chr.6: 26234664-26234664 on Build GRCh38
rs201686429 Chr.6: 26234689-26234689 on Build GRCh38
rs1000795773 Chr.6: 26234709-26234709 on Build GRCh38
rs866323036 Chr.6: 26234877-26234877 on Build GRCh38
rs144176369 Chr.6: 26234612-26234612 on Build GRCh38
rs949598040 Chr.6: 26234698-26234698 on Build GRCh38
rs746824443 Chr.6: 26234744-26234744 on Build GRCh38
rs766318051 Chr.6: 26234864-26234864 on Build GRCh38
rs765746746 Chr.6: 26234552-26234552 on Build GRCh38
rs756175093 Chr.6: 26234605-26234605 on Build GRCh38
rs543699809 Chr.6: 26234786-26234786 on Build GRCh38
rs754453249 Chr.6: 26234938-26234938 on Build GRCh38
rs751105243 Chr.6: 26234793-26234793 on Build GRCh38
rs539123260 Chr.6: 26234873-26234873 on Build GRCh38
rs371497835 Chr.6: 26234726-26234726 on Build GRCh38
rs756770691 Chr.6: 26234707-26234721 on Build GRCh38
rs148166404 Chr.6: 26234533-26234533 on Build GRCh38
rs142030602 Chr.6: 26234537-26234537 on Build GRCh38
rs111312434 Chr.6: 26234945-26234945 on Build GRCh38
rs771890315 Chr.6: 26234544-26234544 on Build GRCh38
rs758741358 Chr.6: 26234666-26234666 on Build GRCh38
rs761023941 Chr.6: 26234683-26234683 on Build GRCh38
rs367665999 Chr.6: 26234832-26234832 on Build GRCh38
rs766988164 Chr.6: 26234894-26234894 on Build GRCh38
rs536681516 Chr.6: 26234860-26234860 on Build GRCh38
rs146809745 Chr.6: 26234731-26234731 on Build GRCh38
rs768502952 Chr.6: 26234935-26234936 on Build GRCh38
rs774094633 Chr.6: 26234855-26234855 on Build GRCh38
rs1025083265 Chr.6: 26234922-26234922 on Build GRCh38
rs200463287 Chr.6: 26234799-26234799 on Build GRCh38
rs762754388 Chr.6: 26234950-26234950 on Build GRCh38
rs141394027 Chr.6: 26234891-26234891 on Build GRCh38
rs377468893 Chr.6: 26234945-26234945 on Build GRCh38
rs760334048 Chr.6: 26234584-26234584 on Build GRCh38
rs778098526 Chr.6: 26234834-26234834 on Build GRCh38
rs752852087 Chr.6: 26234871-26234871 on Build GRCh38
rs138815183 Chr.6: 26234813-26234813 on Build GRCh38
rs181724833 Chr.6: 26234954-26234954 on Build GRCh38
rs762699214 Chr.6: 26234920-26234920 on Build GRCh38
rs771897377 Chr.6: 26234705-26234705 on Build GRCh38
rs548822473 Chr.6: 26234805-26234805 on Build GRCh38
rs758111146 Chr.6: 26234850-26234850 on Build GRCh38
rs985392066 Chr.6: 26234650-26234651 on Build GRCh38
rs767608650 Chr.6: 26234955-26234955 on Build GRCh38
rs759916366 Chr.6: 26234854-26234854 on Build GRCh38
rs754488594 Chr.6: 26234730-26234730 on Build GRCh38
rs140433263 Chr.6: 26234599-26234599 on Build GRCh38
rs768557134 Chr.6: 26234782-26234782 on Build GRCh38
rs775775058 Chr.6: 26234542-26234542 on Build GRCh38
rs549847001 Chr.6: 26234572-26234572 on Build GRCh38
rs200195730 Chr.6: 26234835-26234835 on Build GRCh38
rs113622900 Chr.6: 26234977-26234977 on Build GRCh38
rs770130258 Chr.6: 26234750-26234750 on Build GRCh38
rs150822634 Chr.6: 26234906-26234906 on Build GRCh38
rs140636459 Chr.6: 26234742-26234742 on Build GRCh38
rs1047971723 Chr.6: 26234570-26234571 on Build GRCh38
rs750781968 Chr.6: 26234765-26234765 on Build GRCh38
rs372987662 Chr.6: 26234843-26234843 on Build GRCh38
rs151016458 Chr.6: 26234660-26234660 on Build GRCh38
rs768101896 Chr.6: 26234703-26234703 on Build GRCh38
rs758150131 Chr.6: 26234564-26234564 on Build GRCh38
rs745562029 Chr.6: 26234802-26234803 on Build GRCh38
rs377237102 Chr.6: 26234729-26234729 on Build GRCh38
rs764666462 Chr.6: 26234771-26234771 on Build GRCh38
rs749647274 Chr.6: 26234907-26234907 on Build GRCh38
rs752533232 Chr.6: 26234559-26234559 on Build GRCh38
rs747537217 Chr.6: 26234849-26234849 on Build GRCh38
rs780725404 Chr.6: 26234667-26234667 on Build GRCh38
rs753815669 Chr.6: 26234553-26234553 on Build GRCh38
rs759059672 Chr.6: 26234593-26234593 on Build GRCh38
rs752984127 Chr.6: 26234965-26234965 on Build GRCh38
rs764772054 Chr.6: 26234767-26234767 on Build GRCh38
rs76322151 Chr.6: 26234702-26234702 on Build GRCh38
rs150694983 Chr.6: 26234569-26234569 on Build GRCh38
rs1027218137 Chr.6: 26234856-26234856 on Build GRCh38
rs759527585 Chr.6: 26234915-26234915 on Build GRCh38
rs905354305 Chr.6: 26234706-26234706 on Build GRCh38
rs368646122 Chr.6: 26234751-26234751 on Build GRCh38
rs41266815 Chr.6: 26234923-26234923 on Build GRCh38
rs200372842 Chr.6: 26234975-26234975 on Build GRCh38
rs1045298001 Chr.6: 26234700-26234700 on Build GRCh38
rs544145459 Chr.6: 26234949-26234949 on Build GRCh38
rs201092413 Chr.6: 26234779-26234779 on Build GRCh38
rs377367782 Chr.6: 26234852-26234852 on Build GRCh38
rs376631713 Chr.6: 26234900-26234900 on Build GRCh38
rs200922448 Chr.6: 26234911-26234911 on Build GRCh38
rs200514070 Chr.6: 26234867-26234867 on Build GRCh38
rs763579474 Chr.6: 26234792-26234792 on Build GRCh38
rs780578769 Chr.6: 26234573-26234573 on Build GRCh38
rs775308102 Chr.6: 26234918-26234918 on Build GRCh38
rs200822000 Chr.6: 26234840-26234840 on Build GRCh38
rs772878475 Chr.6: 26234909-26234909 on Build GRCh38
rs756679839 Chr.6: 26234905-26234905 on Build GRCh38
rs537346008 Chr.6: 26234679-26234679 on Build GRCh38
rs756975638 Chr.6: 26234966-26234966 on Build GRCh38
rs761793622 Chr.6: 26234958-26234959 on Build GRCh38
rs867520326 Chr.6: 26234627-26234627 on Build GRCh38
rs762947217 Chr.6: 26234635-26234635 on Build GRCh38
rs758615680 Chr.6: 26234872-26234872 on Build GRCh38
rs760200626 Chr.6: 26234893-26234893 on Build GRCh38
rs760358653 Chr.6: 26234710-26234710 on Build GRCh38
rs369209295 Chr.6: 26234787-26234787 on Build GRCh38
rs553068566 Chr.6: 26234863-26234863 on Build GRCh38
rs771386189 Chr.6: 26234876-26234876 on Build GRCh38
rs368415674 Chr.6: 26234940-26234940 on Build GRCh38
rs775041809 Chr.6: 26234673-26234673 on Build GRCh38
rs757476262 Chr.6: 26234601-26234601 on Build GRCh38
rs780628089 Chr.6: 26234740-26234742 on Build GRCh38
rs143191814 Chr.6: 26234846-26234846 on Build GRCh38
rs936871497 Chr.6: 26234798-26234798 on Build GRCh38
rs749035508 Chr.6: 26234882-26234882 on Build GRCh38
rs143379113 Chr.6: 26234807-26234807 on Build GRCh38
rs200619490 Chr.6: 26234791-26234791 on Build GRCh38
rs775414427 Chr.6: 26234632-26234632 on Build GRCh38
rs755396668 Chr.6: 26234610-26234610 on Build GRCh38
rs200015980 Chr.6: 26234903-26234903 on Build GRCh38
rs767802986 Chr.6: 26234764-26234764 on Build GRCh38
rs768461049 Chr.6: 26234972-26234972 on Build GRCh38
rs374651708 Chr.6: 26234899-26234899 on Build GRCh38
rs1054213579 Chr.6: 26234719-26234719 on Build GRCh38
rs777303503 Chr.6: 26234944-26234944 on Build GRCh38
rs373384671 Chr.6: 26234563-26234563 on Build GRCh38
rs773009233 Chr.6: 26234583-26234583 on Build GRCh38
rs745461263 Chr.6: 26234668-26234668 on Build GRCh38
rs955234705 Chr.6: 26234643-26234643 on Build GRCh38
rs112515836 Chr.6: 26234545-26234546 on Build GRCh38
rs148478928 Chr.6: 26234652-26234652 on Build GRCh38
rs565142442 Chr.6: 26234942-26234942 on Build GRCh38
rs772368103 Chr.6: 26234974-26234974 on Build GRCh38
rs762174884 Chr.6: 26234763-26234763 on Build GRCh38
rs754681281 Chr.6: 26234766-26234766 on Build GRCh38
rs745352725 Chr.6: 26234946-26234946 on Build GRCh38
rs755810082 Chr.6: 26234693-26234693 on Build GRCh38
rs748585271 Chr.6: 26234704-26234705 on Build GRCh38
rs767698067 Chr.6: 26234929-26234929 on Build GRCh38
rs756730837 Chr.6: 26234935-26234935 on Build GRCh38
rs367925017 Chr.6: 26234682-26234682 on Build GRCh38
rs779646561 Chr.6: 26234695-26234695 on Build GRCh38
rs896719006 Chr.6: 26234545-26234545 on Build GRCh38
rs576081113 Chr.6: 26234924-26234924 on Build GRCh38
rs745396335 Chr.6: 26234620-26234620 on Build GRCh38
rs750325803 Chr.6: 26234828-26234828 on Build GRCh38
rs200716311 Chr.6: 26234721-26234721 on Build GRCh38
rs756251364 Chr.6: 26234635-26234655 on Build GRCh38
rs757336092 Chr.6: 26234570-26234570 on Build GRCh38
rs896107272 Chr.6: 26234571-26234571 on Build GRCh38
rs759453204 Chr.6: 26234556-26234556 on Build GRCh38
rs764003776 Chr.6: 26234638-26234638 on Build GRCh38
rs781195252 Chr.6: 26234775-26234775 on Build GRCh38
rs140975803 Chr.6: 26234718-26234718 on Build GRCh38
rs757259808 Chr.6: 26234774-26234774 on Build GRCh38
rs138119279 Chr.6: 26234790-26234790 on Build GRCh38
rs749400507 Chr.6: 26234806-26234806 on Build GRCh38
rs774228817 Chr.6: 26234676-26234676 on Build GRCh38

View Full Product Details