Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26376265-26376758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001197246.2 NM_001197247.2 NM_001197248.2 NM_001197249.2 NM_007047.4 XM_005248827.3 XM_005248831.3 XM_005248832.3 XM_006714979.3 XM_006714980.3 XM_006714981.3 XM_006714982.3 XM_011514267.2 XM_011514268.2 XM_011514269.2 XM_011514270.2 XM_011514271.2 XM_017010211.1 XM_017010212.1 XM_017010213.1 XM_017010214.1 XM_017010215.1 XM_017010216.1 XM_017010217.1

SNP Detail

SNP ID Chromosome Location
rs1007709744 Chr.6: 26376553-26376553 on Build GRCh38
rs538852052 Chr.6: 26376695-26376695 on Build GRCh38
rs965538370 Chr.6: 26376678-26376678 on Build GRCh38
rs936566491 Chr.6: 26376484-26376484 on Build GRCh38
rs530923674 Chr.6: 26376396-26376396 on Build GRCh38
rs9393716 Chr.6: 26376640-26376640 on Build GRCh38
rs368400868 Chr.6: 26376587-26376587 on Build GRCh38
rs1009688565 Chr.6: 26376501-26376501 on Build GRCh38
rs947576985 Chr.6: 26376340-26376340 on Build GRCh38
rs1017858319 Chr.6: 26376342-26376342 on Build GRCh38
rs754160156 Chr.6: 26376720-26376720 on Build GRCh38
rs551007404 Chr.6: 26376644-26376644 on Build GRCh38
rs567744466 Chr.6: 26376415-26376415 on Build GRCh38
rs914788922 Chr.6: 26376313-26376314 on Build GRCh38
rs201645186 Chr.6: 26376467-26376467 on Build GRCh38
rs551291128 Chr.6: 26376727-26376727 on Build GRCh38
rs530113292 Chr.6: 26376465-26376465 on Build GRCh38
rs953802096 Chr.6: 26376696-26376696 on Build GRCh38
rs536719897 Chr.6: 26376439-26376439 on Build GRCh38
rs984252834 Chr.6: 26376700-26376700 on Build GRCh38
rs565823023 Chr.6: 26376450-26376450 on Build GRCh38
rs1053641998 Chr.6: 26376495-26376495 on Build GRCh38
rs1017763302 Chr.6: 26376568-26376568 on Build GRCh38
rs552010050 Chr.6: 26376352-26376352 on Build GRCh38
rs1015361110 Chr.6: 26376433-26376433 on Build GRCh38
rs968947943 Chr.6: 26376505-26376505 on Build GRCh38
rs906351229 Chr.6: 26376449-26376449 on Build GRCh38
rs924387378 Chr.6: 26376360-26376360 on Build GRCh38
rs559705655 Chr.6: 26376593-26376593 on Build GRCh38
rs12214031 Chr.6: 26376400-26376400 on Build GRCh38
rs917359024 Chr.6: 26376463-26376463 on Build GRCh38
rs958957828 Chr.6: 26376469-26376469 on Build GRCh38
rs926521433 Chr.6: 26376473-26376473 on Build GRCh38
rs577266243 Chr.6: 26376498-26376498 on Build GRCh38
rs41266833 Chr.6: 26376645-26376645 on Build GRCh38
rs1045328964 Chr.6: 26376466-26376466 on Build GRCh38
rs973669914 Chr.6: 26376372-26376372 on Build GRCh38
rs936512876 Chr.6: 26376660-26376660 on Build GRCh38
rs530141928 Chr.6: 26376652-26376652 on Build GRCh38
rs189964435 Chr.6: 26376574-26376574 on Build GRCh38
rs888361878 Chr.6: 26376405-26376405 on Build GRCh38
rs780869237 Chr.6: 26376395-26376395 on Build GRCh38
rs13218591 Chr.6: 26376604-26376604 on Build GRCh38
rs536630019 Chr.6: 26376499-26376499 on Build GRCh38
rs950743592 Chr.6: 26376597-26376597 on Build GRCh38
rs144182156 Chr.6: 26376733-26376733 on Build GRCh38
rs769615055 Chr.6: 26376378-26376378 on Build GRCh38
rs976916480 Chr.6: 26376534-26376534 on Build GRCh38
rs534814181 Chr.6: 26376726-26376726 on Build GRCh38
rs375635004 Chr.6: 26376580-26376580 on Build GRCh38
rs115643731 Chr.6: 26376626-26376626 on Build GRCh38
rs924792557 Chr.6: 26376519-26376519 on Build GRCh38
rs571707261 Chr.6: 26376362-26376362 on Build GRCh38
rs1004030510 Chr.6: 26376407-26376407 on Build GRCh38
rs954899827 Chr.6: 26376329-26376329 on Build GRCh38
rs1030499995 Chr.6: 26376302-26376302 on Build GRCh38
rs1036878231 Chr.6: 26376598-26376598 on Build GRCh38
rs943753064 Chr.6: 26376370-26376370 on Build GRCh38
rs548930843 Chr.6: 26376443-26376443 on Build GRCh38
rs569939238 Chr.6: 26376520-26376520 on Build GRCh38
rs565554111 Chr.6: 26376629-26376629 on Build GRCh38
rs894554515 Chr.6: 26376639-26376639 on Build GRCh38
rs775902280 Chr.6: 26376677-26376677 on Build GRCh38
rs111353898 Chr.6: 26376706-26376706 on Build GRCh38
rs528782425 Chr.6: 26376341-26376341 on Build GRCh38
rs547033648 Chr.6: 26376691-26376691 on Build GRCh38
rs548594777 Chr.6: 26376513-26376513 on Build GRCh38
rs1051891246 Chr.6: 26376715-26376715 on Build GRCh38
rs1003336609 Chr.6: 26376605-26376605 on Build GRCh38
rs976945455 Chr.6: 26376686-26376686 on Build GRCh38

View Full Product Details