Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151838559-151839058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD4D
Entrez Gene ID
100191040
Gene Name
C2 calcium dependent domain containing 4D
Gene Aliases
FAM148D
UniGene
Hs.694717
Gene Chromosome Location
Chr.1: 151837818-151840557 on Build GRCh38
Transcripts
NM_001136003.1 XM_011509055.2 XM_016999989.1 NR_024237.1

Gene Symbol
LOC100132111
Entrez Gene ID
100132111
Gene Name
uncharacterized LOC100132111
Gene Aliases
-
UniGene
Hs.697059
Gene Chromosome Location
Chr.1: 151838469-151844165 on Build GRCh38
Transcripts
NM_001136003.1 XM_011509055.2 XM_016999989.1 NR_024237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275290 c.6G>A p.W2* Chr.1: 151838984-151838984 on Build GRCh38
5689007 c.394C>A p.L132M Chr.1: 151838596-151838596 on Build GRCh38
7266298 c.40G>A p.A14T Chr.1: 151838950-151838950 on Build GRCh38
3740654 c.118C>T p.P40S Chr.1: 151838872-151838872 on Build GRCh38
5817512 c.338C>G p.A113G Chr.1: 151838652-151838652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183029853 Chr.1: 151838958-151838958 on Build GRCh38
rs931416427 Chr.1: 151838761-151838761 on Build GRCh38
rs1047652770 Chr.1: 151838927-151838927 on Build GRCh38
rs753860529 Chr.1: 151838921-151838921 on Build GRCh38
rs950088856 Chr.1: 151838645-151838645 on Build GRCh38
rs994721869 Chr.1: 151838728-151838728 on Build GRCh38
rs918719629 Chr.1: 151838609-151838609 on Build GRCh38
rs761128301 Chr.1: 151838758-151838758 on Build GRCh38
rs982090073 Chr.1: 151838708-151838708 on Build GRCh38
rs781312077 Chr.1: 151838920-151838920 on Build GRCh38
rs752385267 Chr.1: 151838896-151838896 on Build GRCh38
rs1041305189 Chr.1: 151838828-151838828 on Build GRCh38
rs776992898 Chr.1: 151839008-151839008 on Build GRCh38
rs1037125976 Chr.1: 151838604-151838604 on Build GRCh38
rs950058783 Chr.1: 151838787-151838787 on Build GRCh38
rs900213424 Chr.1: 151839003-151839003 on Build GRCh38
rs1025675883 Chr.1: 151838999-151838999 on Build GRCh38
rs1025898040 Chr.1: 151838731-151838731 on Build GRCh38
rs777238973 Chr.1: 151838824-151838824 on Build GRCh38
rs539333912 Chr.1: 151838850-151838850 on Build GRCh38
rs1008588917 Chr.1: 151838935-151838935 on Build GRCh38
rs931488476 Chr.1: 151838635-151838635 on Build GRCh38
rs747308407 Chr.1: 151838970-151838970 on Build GRCh38
rs926119380 Chr.1: 151838751-151838751 on Build GRCh38
rs775646422 Chr.1: 151838757-151838757 on Build GRCh38
rs537061891 Chr.1: 151838644-151838644 on Build GRCh38
rs911435747 Chr.1: 151838799-151838799 on Build GRCh38
rs772556969 Chr.1: 151838753-151838753 on Build GRCh38
rs972717365 Chr.1: 151838965-151838965 on Build GRCh38
rs1055449744 Chr.1: 151838684-151838684 on Build GRCh38
rs752964206 Chr.1: 151838676-151838676 on Build GRCh38
rs1010634766 Chr.1: 151838901-151838901 on Build GRCh38
rs1018727037 Chr.1: 151838735-151838735 on Build GRCh38
rs6673556 Chr.1: 151838750-151838750 on Build GRCh38
rs755968172 Chr.1: 151838919-151838919 on Build GRCh38
rs771787977 Chr.1: 151838959-151838959 on Build GRCh38
rs753697778 Chr.1: 151838661-151838661 on Build GRCh38
rs950407117 Chr.1: 151838732-151838732 on Build GRCh38
rs1045827719 Chr.1: 151838649-151838649 on Build GRCh38
rs777798208 Chr.1: 151838672-151838672 on Build GRCh38
rs981913915 Chr.1: 151838793-151838793 on Build GRCh38
rs757353692 Chr.1: 151838751-151838752 on Build GRCh38
rs1023886637 Chr.1: 151838904-151838904 on Build GRCh38
rs769007185 Chr.1: 151838776-151838776 on Build GRCh38
rs750177782 Chr.1: 151838764-151838764 on Build GRCh38
rs774118895 Chr.1: 151838743-151838743 on Build GRCh38
rs746137221 Chr.1: 151838962-151838962 on Build GRCh38
rs758488906 Chr.1: 151838705-151838705 on Build GRCh38
rs1019109809 Chr.1: 151838963-151838963 on Build GRCh38
rs927439572 Chr.1: 151838823-151838823 on Build GRCh38
rs756083266 Chr.1: 151838977-151838977 on Build GRCh38
rs1052103219 Chr.1: 151838854-151838854 on Build GRCh38
rs564914374 Chr.1: 151838660-151838660 on Build GRCh38
rs960032222 Chr.1: 151838739-151838739 on Build GRCh38
rs930540012 Chr.1: 151838878-151838878 on Build GRCh38
rs1053073013 Chr.1: 151838591-151838591 on Build GRCh38
rs1010706765 Chr.1: 151838694-151838694 on Build GRCh38
rs755938162 Chr.1: 151838661-151838661 on Build GRCh38
rs779790861 Chr.1: 151838967-151838967 on Build GRCh38
rs1023495953 Chr.1: 151838696-151838696 on Build GRCh38
rs763094959 Chr.1: 151838867-151838867 on Build GRCh38
rs532247828 Chr.1: 151838945-151838945 on Build GRCh38
rs765360637 Chr.1: 151838851-151838851 on Build GRCh38
rs900793190 Chr.1: 151838844-151838844 on Build GRCh38
rs999194511 Chr.1: 151838666-151838666 on Build GRCh38
rs747988147 Chr.1: 151838610-151838610 on Build GRCh38
rs756812351 Chr.1: 151838933-151838933 on Build GRCh38
rs942941595 Chr.1: 151838827-151838827 on Build GRCh38
rs965226515 Chr.1: 151838748-151838748 on Build GRCh38
rs781095843 Chr.1: 151838734-151838734 on Build GRCh38
rs913206234 Chr.1: 151838785-151838785 on Build GRCh38
rs766601318 Chr.1: 151838868-151838868 on Build GRCh38
rs972790854 Chr.1: 151838747-151838747 on Build GRCh38

View Full Product Details