Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57181942-57182438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG2
Entrez Gene ID
9532
Gene Name
BCL2 associated athanogene 2
Gene Aliases
BAG-2, dJ417I1.2
UniGene
Hs.745046
Gene Chromosome Location
Chr.6: 57172306-57185215 on Build GRCh38
Transcripts
NM_004282.3 XM_005249490.3 XM_011514998.2 XM_011514999.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445508 c.160G>T p.E54* Chr.6: 57182078-57182078 on Build GRCh38
1445509 c.201G>A p.Q67Q Chr.6: 57182119-57182119 on Build GRCh38
3715559 c.222C>T p.D74D Chr.6: 57182140-57182140 on Build GRCh38
1672536 c.179T>C p.I60T Chr.6: 57182097-57182097 on Build GRCh38
7327230 c.150G>A p.E50E Chr.6: 57182068-57182068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35189709 Chr.6: 57182041-57182041 on Build GRCh38
rs778577656 Chr.6: 57182028-57182028 on Build GRCh38
rs987733126 Chr.6: 57182394-57182394 on Build GRCh38
rs980908031 Chr.6: 57182196-57182198 on Build GRCh38
rs574420183 Chr.6: 57182228-57182228 on Build GRCh38
rs746743643 Chr.6: 57182000-57182000 on Build GRCh38
rs958463272 Chr.6: 57182360-57182360 on Build GRCh38
rs186921168 Chr.6: 57182365-57182365 on Build GRCh38
rs932380077 Chr.6: 57182142-57182142 on Build GRCh38
rs77741301 Chr.6: 57182414-57182414 on Build GRCh38
rs753608649 Chr.6: 57182138-57182138 on Build GRCh38
rs576878604 Chr.6: 57182221-57182221 on Build GRCh38
rs954425714 Chr.6: 57181998-57181998 on Build GRCh38
rs766755089 Chr.6: 57182083-57182083 on Build GRCh38
rs12197258 Chr.6: 57182383-57182383 on Build GRCh38
rs776898508 Chr.6: 57182024-57182024 on Build GRCh38
rs181118137 Chr.6: 57182294-57182294 on Build GRCh38
rs1048286601 Chr.6: 57182180-57182180 on Build GRCh38
rs764847840 Chr.6: 57182141-57182141 on Build GRCh38
rs375376850 Chr.6: 57182154-57182154 on Build GRCh38
rs769328263 Chr.6: 57182073-57182076 on Build GRCh38
rs753883085 Chr.6: 57181967-57181967 on Build GRCh38
rs939844616 Chr.6: 57182191-57182191 on Build GRCh38
rs567906439 Chr.6: 57182398-57182398 on Build GRCh38
rs905317476 Chr.6: 57182265-57182265 on Build GRCh38
rs745854137 Chr.6: 57181984-57181984 on Build GRCh38
rs41267687 Chr.6: 57182008-57182008 on Build GRCh38
rs755065373 Chr.6: 57182371-57182371 on Build GRCh38
rs758241444 Chr.6: 57181992-57181992 on Build GRCh38
rs767979775 Chr.6: 57182134-57182134 on Build GRCh38
rs1032405634 Chr.6: 57182332-57182332 on Build GRCh38
rs540757936 Chr.6: 57182236-57182236 on Build GRCh38
rs553248975 Chr.6: 57182244-57182244 on Build GRCh38
rs145764571 Chr.6: 57182400-57182400 on Build GRCh38
rs377471840 Chr.6: 57182109-57182109 on Build GRCh38
rs900028853 Chr.6: 57182369-57182369 on Build GRCh38
rs558216538 Chr.6: 57182195-57182195 on Build GRCh38
rs1024923878 Chr.6: 57182264-57182264 on Build GRCh38
rs202083861 Chr.6: 57182149-57182149 on Build GRCh38
rs1052426127 Chr.6: 57182237-57182237 on Build GRCh38
rs976373918 Chr.6: 57182045-57182045 on Build GRCh38
rs574671212 Chr.6: 57182278-57182278 on Build GRCh38
rs370574275 Chr.6: 57182113-57182113 on Build GRCh38
rs188737182 Chr.6: 57182187-57182187 on Build GRCh38
rs749850943 Chr.6: 57182146-57182146 on Build GRCh38
rs1008568020 Chr.6: 57182415-57182415 on Build GRCh38
rs773144401 Chr.6: 57182064-57182064 on Build GRCh38
rs1013579520 Chr.6: 57182251-57182251 on Build GRCh38
rs779859251 Chr.6: 57181993-57181993 on Build GRCh38
rs758364398 Chr.6: 57182148-57182148 on Build GRCh38
rs1050226427 Chr.6: 57182405-57182405 on Build GRCh38
rs149704905 Chr.6: 57182082-57182082 on Build GRCh38
rs371537828 Chr.6: 57182169-57182169 on Build GRCh38
rs928017845 Chr.6: 57182201-57182201 on Build GRCh38
rs756825488 Chr.6: 57182140-57182140 on Build GRCh38
rs754836702 Chr.6: 57182182-57182182 on Build GRCh38

View Full Product Details