Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157091960-157092454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV3L
Entrez Gene ID
440695
Gene Name
ETS variant 3 like
Gene Aliases
-
UniGene
Hs.127138
Gene Chromosome Location
Chr.1: 157092043-157099808 on Build GRCh38
Transcripts
NM_001004341.2

SNP Detail

SNP ID Chromosome Location
rs988725972 Chr.1: 157092011-157092011 on Build GRCh38
rs1000461633 Chr.1: 157091983-157091983 on Build GRCh38
rs955862758 Chr.1: 157092352-157092352 on Build GRCh38
rs370946042 Chr.1: 157092187-157092187 on Build GRCh38
rs562957919 Chr.1: 157092024-157092024 on Build GRCh38
rs578209622 Chr.1: 157092018-157092018 on Build GRCh38
rs941099646 Chr.1: 157092095-157092095 on Build GRCh38
rs533358183 Chr.1: 157092045-157092045 on Build GRCh38
rs771591359 Chr.1: 157092022-157092022 on Build GRCh38
rs537336581 Chr.1: 157091993-157091993 on Build GRCh38
rs956186545 Chr.1: 157091990-157091990 on Build GRCh38
rs968335436 Chr.1: 157092051-157092051 on Build GRCh38
rs184632730 Chr.1: 157092027-157092027 on Build GRCh38
rs978945708 Chr.1: 157092070-157092070 on Build GRCh38
rs1055106 Chr.1: 157092181-157092181 on Build GRCh38
rs142442117 Chr.1: 157092245-157092245 on Build GRCh38
rs767630418 Chr.1: 157092297-157092297 on Build GRCh38
rs7534856 Chr.1: 157092113-157092113 on Build GRCh38
rs542620275 Chr.1: 157092026-157092026 on Build GRCh38
rs1038993461 Chr.1: 157092272-157092272 on Build GRCh38
rs1033299692 Chr.1: 157092319-157092319 on Build GRCh38
rs114790584 Chr.1: 157092294-157092294 on Build GRCh38
rs565164494 Chr.1: 157092282-157092282 on Build GRCh38
rs867534128 Chr.1: 157092379-157092379 on Build GRCh38
rs565128298 Chr.1: 157092225-157092225 on Build GRCh38
rs189344641 Chr.1: 157092392-157092392 on Build GRCh38
rs111355391 Chr.1: 157091984-157091984 on Build GRCh38
rs920764591 Chr.1: 157092149-157092149 on Build GRCh38
rs1024197814 Chr.1: 157092431-157092431 on Build GRCh38
rs538486131 Chr.1: 157092401-157092401 on Build GRCh38
rs192087262 Chr.1: 157092023-157092023 on Build GRCh38
rs531156333 Chr.1: 157092407-157092407 on Build GRCh38
rs1010167819 Chr.1: 157092378-157092378 on Build GRCh38
rs2766080 Chr.1: 157092180-157092180 on Build GRCh38
rs974634096 Chr.1: 157092279-157092279 on Build GRCh38
rs573986705 Chr.1: 157092025-157092025 on Build GRCh38
rs550290007 Chr.1: 157092287-157092287 on Build GRCh38
rs1055107 Chr.1: 157092169-157092169 on Build GRCh38
rs1039061783 Chr.1: 157092021-157092021 on Build GRCh38
rs890470321 Chr.1: 157092171-157092171 on Build GRCh38
rs75257253 Chr.1: 157092166-157092166 on Build GRCh38
rs141946749 Chr.1: 157092022-157092022 on Build GRCh38
rs542316015 Chr.1: 157092064-157092064 on Build GRCh38
rs939395994 Chr.1: 157092400-157092400 on Build GRCh38

View Full Product Details