Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157098611-157099104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV3L
Entrez Gene ID
440695
Gene Name
ETS variant 3 like
Gene Aliases
-
UniGene
Hs.127138
Gene Chromosome Location
Chr.1: 157092043-157099808 on Build GRCh38
Transcripts
NM_001004341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3802285 c.422A>C p.H141P Chr.1: 157098770-157098770 on Build GRCh38
6444060 c.397G>A p.E133K Chr.1: 157098795-157098795 on Build GRCh38
897500 c.375C>T p.I125I Chr.1: 157098817-157098817 on Build GRCh38
5477672 c.335G>A p.G112D Chr.1: 157098857-157098857 on Build GRCh38
897499 c.403C>T p.R135W Chr.1: 157098789-157098789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772728724 Chr.1: 157098732-157098733 on Build GRCh38
rs762590849 Chr.1: 157098759-157098759 on Build GRCh38
rs375485140 Chr.1: 157098926-157098926 on Build GRCh38
rs147712441 Chr.1: 157098740-157098740 on Build GRCh38
rs16838072 Chr.1: 157098880-157098880 on Build GRCh38
rs776003709 Chr.1: 157098668-157098668 on Build GRCh38
rs760681245 Chr.1: 157098881-157098881 on Build GRCh38
rs765556753 Chr.1: 157098785-157098785 on Build GRCh38
rs772728404 Chr.1: 157098667-157098667 on Build GRCh38
rs577101931 Chr.1: 157098788-157098788 on Build GRCh38
rs1009701753 Chr.1: 157098826-157098826 on Build GRCh38
rs371804655 Chr.1: 157098816-157098816 on Build GRCh38
rs972430686 Chr.1: 157099018-157099018 on Build GRCh38
rs943798563 Chr.1: 157098851-157098851 on Build GRCh38
rs909099537 Chr.1: 157098980-157098980 on Build GRCh38
rs952650102 Chr.1: 157099047-157099047 on Build GRCh38
rs550855165 Chr.1: 157098733-157098733 on Build GRCh38
rs778098669 Chr.1: 157098823-157098823 on Build GRCh38
rs116859604 Chr.1: 157098724-157098724 on Build GRCh38
rs953028422 Chr.1: 157098938-157098938 on Build GRCh38
rs572957328 Chr.1: 157098925-157098925 on Build GRCh38
rs753660679 Chr.1: 157098888-157098888 on Build GRCh38
rs12083811 Chr.1: 157098741-157098741 on Build GRCh38
rs957332282 Chr.1: 157098644-157098644 on Build GRCh38
rs764832204 Chr.1: 157098689-157098689 on Build GRCh38
rs367802331 Chr.1: 157098929-157098929 on Build GRCh38
rs371229211 Chr.1: 157098907-157098907 on Build GRCh38
rs747406102 Chr.1: 157098739-157098740 on Build GRCh38
rs758434202 Chr.1: 157098732-157098732 on Build GRCh38
rs769453836 Chr.1: 157098745-157098745 on Build GRCh38
rs752507586 Chr.1: 157098723-157098723 on Build GRCh38
rs745957498 Chr.1: 157098725-157098725 on Build GRCh38
rs749020214 Chr.1: 157098838-157098838 on Build GRCh38
rs896523703 Chr.1: 157098770-157098770 on Build GRCh38
rs552196259 Chr.1: 157098771-157098771 on Build GRCh38
rs762918639 Chr.1: 157098789-157098789 on Build GRCh38
rs544819258 Chr.1: 157098951-157098951 on Build GRCh38
rs372395455 Chr.1: 157098720-157098720 on Build GRCh38
rs754950481 Chr.1: 157098906-157098906 on Build GRCh38
rs982893160 Chr.1: 157099052-157099052 on Build GRCh38
rs759556960 Chr.1: 157098773-157098773 on Build GRCh38
rs548190247 Chr.1: 157098756-157098756 on Build GRCh38
rs183986526 Chr.1: 157098697-157098697 on Build GRCh38
rs535850379 Chr.1: 157098753-157098753 on Build GRCh38
rs940499961 Chr.1: 157098757-157098757 on Build GRCh38
rs149030440 Chr.1: 157098784-157098784 on Build GRCh38
rs374328445 Chr.1: 157098947-157098947 on Build GRCh38
rs761525642 Chr.1: 157099073-157099073 on Build GRCh38
rs61735500 Chr.1: 157098803-157098803 on Build GRCh38
rs1011683437 Chr.1: 157098673-157098673 on Build GRCh38
rs1020423069 Chr.1: 157098710-157098710 on Build GRCh38
rs766466276 Chr.1: 157098899-157098899 on Build GRCh38
rs991263515 Chr.1: 157099015-157099015 on Build GRCh38
rs566470571 Chr.1: 157099003-157099003 on Build GRCh38
rs753262856 Chr.1: 157098910-157098910 on Build GRCh38
rs370076655 Chr.1: 157098941-157098941 on Build GRCh38
rs967906791 Chr.1: 157098973-157098973 on Build GRCh38
rs747335015 Chr.1: 157098670-157098670 on Build GRCh38
rs776843674 Chr.1: 157098680-157098680 on Build GRCh38
rs547827441 Chr.1: 157099002-157099002 on Build GRCh38
rs769129043 Chr.1: 157098675-157098675 on Build GRCh38
rs748521800 Chr.1: 157098809-157098809 on Build GRCh38
rs796424681 Chr.1: 157098832-157098832 on Build GRCh38
rs767316136 Chr.1: 157098781-157098781 on Build GRCh38
rs144577755 Chr.1: 157099035-157099035 on Build GRCh38
rs769353508 Chr.1: 157098698-157098698 on Build GRCh38
rs761496027 Chr.1: 157098687-157098687 on Build GRCh38
rs751677051 Chr.1: 157098902-157098902 on Build GRCh38
rs201139806 Chr.1: 157098769-157098769 on Build GRCh38
rs530432055 Chr.1: 157098979-157098979 on Build GRCh38
rs909153669 Chr.1: 157098751-157098751 on Build GRCh38
rs758920690 Chr.1: 157098865-157098865 on Build GRCh38
rs769015280 Chr.1: 157098734-157098734 on Build GRCh38
rs763272800 Chr.1: 157098896-157098896 on Build GRCh38
rs770460497 Chr.1: 157098844-157098844 on Build GRCh38
rs563012315 Chr.1: 157098952-157098952 on Build GRCh38
rs201223286 Chr.1: 157098738-157098738 on Build GRCh38
rs569157105 Chr.1: 157098945-157098945 on Build GRCh38
rs773940808 Chr.1: 157098855-157098855 on Build GRCh38
rs754859635 Chr.1: 157098699-157098699 on Build GRCh38
rs767395485 Chr.1: 157098719-157098719 on Build GRCh38
rs770638515 Chr.1: 157098933-157098933 on Build GRCh38
rs551366020 Chr.1: 157098874-157098874 on Build GRCh38
rs574694747 Chr.1: 157098833-157098833 on Build GRCh38
rs532552968 Chr.1: 157098942-157098942 on Build GRCh38
rs558730616 Chr.1: 157098786-157098786 on Build GRCh38
rs1002921950 Chr.1: 157098981-157098981 on Build GRCh38
rs764035372 Chr.1: 157098886-157098886 on Build GRCh38
rs775805690 Chr.1: 157098737-157098737 on Build GRCh38

View Full Product Details