Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157099034-157099527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV3L
Entrez Gene ID
440695
Gene Name
ETS variant 3 like
Gene Aliases
-
UniGene
Hs.127138
Gene Chromosome Location
Chr.1: 157092043-157099808 on Build GRCh38
Transcripts
NM_001004341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750451 c.75T>C p.D25D Chr.1: 157099362-157099362 on Build GRCh38
346546 c.223G>A p.E75K Chr.1: 157099214-157099214 on Build GRCh38
6478212 c.279G>T p.K93N Chr.1: 157099158-157099158 on Build GRCh38
3802289 c.157G>A p.E53K Chr.1: 157099280-157099280 on Build GRCh38
5435616 c.164G>A p.R55H Chr.1: 157099273-157099273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764533881 Chr.1: 157099146-157099146 on Build GRCh38
rs762633924 Chr.1: 157099242-157099243 on Build GRCh38
rs914706303 Chr.1: 157099415-157099415 on Build GRCh38
rs867561121 Chr.1: 157099170-157099170 on Build GRCh38
rs568226360 Chr.1: 157099250-157099250 on Build GRCh38
rs775621115 Chr.1: 157099321-157099321 on Build GRCh38
rs761901267 Chr.1: 157099401-157099401 on Build GRCh38
rs759791604 Chr.1: 157099129-157099129 on Build GRCh38
rs776369446 Chr.1: 157099193-157099193 on Build GRCh38
rs908162279 Chr.1: 157099090-157099090 on Build GRCh38
rs761460239 Chr.1: 157099119-157099119 on Build GRCh38
rs901435637 Chr.1: 157099367-157099367 on Build GRCh38
rs752164549 Chr.1: 157099443-157099443 on Build GRCh38
rs758152885 Chr.1: 157099267-157099267 on Build GRCh38
rs369834258 Chr.1: 157099103-157099103 on Build GRCh38
rs372227275 Chr.1: 157099304-157099304 on Build GRCh38
rs538008233 Chr.1: 157099196-157099196 on Build GRCh38
rs921046376 Chr.1: 157099361-157099361 on Build GRCh38
rs763344237 Chr.1: 157099357-157099357 on Build GRCh38
rs148267509 Chr.1: 157099482-157099482 on Build GRCh38
rs866279665 Chr.1: 157099105-157099105 on Build GRCh38
rs777030540 Chr.1: 157099110-157099111 on Build GRCh38
rs80223807 Chr.1: 157099423-157099423 on Build GRCh38
rs774881884 Chr.1: 157099485-157099485 on Build GRCh38
rs1042114857 Chr.1: 157099239-157099239 on Build GRCh38
rs763541073 Chr.1: 157099429-157099429 on Build GRCh38
rs368401445 Chr.1: 157099328-157099328 on Build GRCh38
rs747280743 Chr.1: 157099391-157099391 on Build GRCh38
rs150322506 Chr.1: 157099273-157099273 on Build GRCh38
rs768680694 Chr.1: 157099110-157099110 on Build GRCh38
rs530277050 Chr.1: 157099398-157099398 on Build GRCh38
rs761525642 Chr.1: 157099073-157099073 on Build GRCh38
rs745621994 Chr.1: 157099456-157099456 on Build GRCh38
rs769536170 Chr.1: 157099197-157099197 on Build GRCh38
rs760882467 Chr.1: 157099327-157099327 on Build GRCh38
rs552221977 Chr.1: 157099470-157099470 on Build GRCh38
rs760688466 Chr.1: 157099241-157099241 on Build GRCh38
rs762710188 Chr.1: 157099111-157099111 on Build GRCh38
rs61730132 Chr.1: 157099342-157099342 on Build GRCh38
rs140222573 Chr.1: 157099185-157099185 on Build GRCh38
rs199499287 Chr.1: 157099366-157099366 on Build GRCh38
rs773869771 Chr.1: 157099406-157099406 on Build GRCh38
rs16838075 Chr.1: 157099362-157099362 on Build GRCh38
rs769976233 Chr.1: 157099479-157099479 on Build GRCh38
rs756646645 Chr.1: 157099144-157099144 on Build GRCh38
rs947661413 Chr.1: 157099230-157099230 on Build GRCh38
rs367691485 Chr.1: 157099296-157099296 on Build GRCh38
rs74938633 Chr.1: 157099450-157099450 on Build GRCh38
rs759146244 Chr.1: 157099440-157099440 on Build GRCh38
rs766899991 Chr.1: 157099430-157099430 on Build GRCh38
rs375452315 Chr.1: 157099442-157099442 on Build GRCh38
rs759817053 Chr.1: 157099451-157099451 on Build GRCh38
rs374533520 Chr.1: 157099290-157099290 on Build GRCh38
rs775567685 Chr.1: 157099217-157099217 on Build GRCh38
rs368745197 Chr.1: 157099086-157099086 on Build GRCh38
rs199807521 Chr.1: 157099128-157099128 on Build GRCh38
rs777706159 Chr.1: 157099386-157099386 on Build GRCh38
rs750838692 Chr.1: 157099347-157099347 on Build GRCh38
rs755177088 Chr.1: 157099461-157099461 on Build GRCh38
rs779331270 Chr.1: 157099157-157099157 on Build GRCh38
rs149508240 Chr.1: 157099481-157099481 on Build GRCh38
rs765974958 Chr.1: 157099393-157099394 on Build GRCh38
rs953139938 Chr.1: 157099211-157099211 on Build GRCh38
rs752744015 Chr.1: 157099142-157099142 on Build GRCh38
rs767688626 Chr.1: 157099208-157099208 on Build GRCh38
rs368973328 Chr.1: 157099495-157099495 on Build GRCh38
rs776746126 Chr.1: 157099341-157099341 on Build GRCh38
rs770776463 Chr.1: 157099390-157099390 on Build GRCh38
rs750766156 Chr.1: 157099263-157099263 on Build GRCh38
rs768731373 Chr.1: 157099189-157099189 on Build GRCh38
rs752214159 Chr.1: 157099375-157099375 on Build GRCh38
rs755581979 Chr.1: 157099447-157099447 on Build GRCh38
rs745561256 Chr.1: 157099175-157099175 on Build GRCh38
rs111662736 Chr.1: 157099346-157099346 on Build GRCh38
rs757160790 Chr.1: 157099455-157099455 on Build GRCh38
rs759795311 Chr.1: 157099205-157099205 on Build GRCh38
rs892681618 Chr.1: 157099155-157099155 on Build GRCh38
rs767633624 Chr.1: 157099138-157099138 on Build GRCh38
rs370221827 Chr.1: 157099274-157099274 on Build GRCh38
rs777462387 Chr.1: 157099312-157099312 on Build GRCh38
rs139716725 Chr.1: 157099202-157099202 on Build GRCh38
rs746989914 Chr.1: 157099108-157099108 on Build GRCh38
rs1005803442 Chr.1: 157099238-157099238 on Build GRCh38
rs756030311 Chr.1: 157099383-157099383 on Build GRCh38
rs768958093 Chr.1: 157099105-157099105 on Build GRCh38
rs751945362 Chr.1: 157099437-157099437 on Build GRCh38
rs766632785 Chr.1: 157099127-157099127 on Build GRCh38
rs765566393 Chr.1: 157099253-157099253 on Build GRCh38
rs374905841 Chr.1: 157099319-157099319 on Build GRCh38
rs894601470 Chr.1: 157099471-157099471 on Build GRCh38
rs751315681 Chr.1: 157099364-157099364 on Build GRCh38
rs771528327 Chr.1: 157099484-157099484 on Build GRCh38
rs780610906 Chr.1: 157099368-157099368 on Build GRCh38
rs764665765 Chr.1: 157099242-157099242 on Build GRCh38
rs535746301 Chr.1: 157099251-157099251 on Build GRCh38
rs1003386769 Chr.1: 157099262-157099262 on Build GRCh38
rs770208936 Chr.1: 157099413-157099413 on Build GRCh38
rs893426995 Chr.1: 157099204-157099204 on Build GRCh38
rs367920616 Chr.1: 157099150-157099150 on Build GRCh38
rs757496608 Chr.1: 157099151-157099151 on Build GRCh38
rs749083227 Chr.1: 157099389-157099389 on Build GRCh38
rs780744286 Chr.1: 157099295-157099295 on Build GRCh38
rs61730131 Chr.1: 157099176-157099176 on Build GRCh38
rs746475053 Chr.1: 157099186-157099186 on Build GRCh38
rs752789582 Chr.1: 157099496-157099496 on Build GRCh38
rs16838078 Chr.1: 157099469-157099469 on Build GRCh38
rs145414360 Chr.1: 157099270-157099270 on Build GRCh38

View Full Product Details