Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157099337-157099838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV3L
Entrez Gene ID
440695
Gene Name
ETS variant 3 like
Gene Aliases
-
UniGene
Hs.127138
Gene Chromosome Location
Chr.1: 157092043-157099808 on Build GRCh38
Transcripts
NM_001004341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750451 c.75T>C p.D25D Chr.1: 157099362-157099362 on Build GRCh38
5905395 c.98C>T p.S33F Chr.1: 157099339-157099339 on Build GRCh38
897502 c.53T>C p.I18T Chr.1: 157099471-157099471 on Build GRCh38
6501760 c.84C>G p.Y28* Chr.1: 157099353-157099353 on Build GRCh38
4558783 c.77G>A p.W26* Chr.1: 157099360-157099360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765974958 Chr.1: 157099393-157099394 on Build GRCh38
rs568246199 Chr.1: 157099672-157099672 on Build GRCh38
rs914706303 Chr.1: 157099415-157099415 on Build GRCh38
rs554019154 Chr.1: 157099727-157099727 on Build GRCh38
rs915876558 Chr.1: 157099781-157099781 on Build GRCh38
rs375595524 Chr.1: 157099534-157099534 on Build GRCh38
rs965664043 Chr.1: 157099576-157099576 on Build GRCh38
rs976449105 Chr.1: 157099581-157099581 on Build GRCh38
rs888512400 Chr.1: 157099802-157099802 on Build GRCh38
rs535334128 Chr.1: 157099688-157099688 on Build GRCh38
rs370224118 Chr.1: 157099533-157099533 on Build GRCh38
rs764527023 Chr.1: 157099774-157099774 on Build GRCh38
rs977979251 Chr.1: 157099762-157099762 on Build GRCh38
rs989550473 Chr.1: 157099723-157099723 on Build GRCh38
rs368973328 Chr.1: 157099495-157099495 on Build GRCh38
rs367562492 Chr.1: 157099570-157099570 on Build GRCh38
rs761901267 Chr.1: 157099401-157099401 on Build GRCh38
rs770776463 Chr.1: 157099390-157099390 on Build GRCh38
rs79218294 Chr.1: 157099796-157099796 on Build GRCh38
rs750196282 Chr.1: 157099554-157099554 on Build GRCh38
rs752214159 Chr.1: 157099375-157099375 on Build GRCh38
rs758985619 Chr.1: 157099546-157099547 on Build GRCh38
rs755581979 Chr.1: 157099447-157099447 on Build GRCh38
rs901435637 Chr.1: 157099367-157099367 on Build GRCh38
rs1037267311 Chr.1: 157099808-157099808 on Build GRCh38
rs752164549 Chr.1: 157099443-157099443 on Build GRCh38
rs1057060954 Chr.1: 157099776-157099776 on Build GRCh38
rs757160790 Chr.1: 157099455-157099455 on Build GRCh38
rs921046376 Chr.1: 157099361-157099361 on Build GRCh38
rs112315179 Chr.1: 157099805-157099805 on Build GRCh38
rs577652050 Chr.1: 157099678-157099678 on Build GRCh38
rs763344237 Chr.1: 157099357-157099357 on Build GRCh38
rs148267509 Chr.1: 157099482-157099482 on Build GRCh38
rs779701367 Chr.1: 157099566-157099566 on Build GRCh38
rs80223807 Chr.1: 157099423-157099423 on Build GRCh38
rs774881884 Chr.1: 157099485-157099485 on Build GRCh38
rs763541073 Chr.1: 157099429-157099429 on Build GRCh38
rs747280743 Chr.1: 157099391-157099391 on Build GRCh38
rs750389631 Chr.1: 157099713-157099713 on Build GRCh38
rs530277050 Chr.1: 157099398-157099398 on Build GRCh38
rs756030311 Chr.1: 157099383-157099383 on Build GRCh38
rs745621994 Chr.1: 157099456-157099456 on Build GRCh38
rs1015536556 Chr.1: 157099572-157099572 on Build GRCh38
rs552221977 Chr.1: 157099470-157099470 on Build GRCh38
rs751945362 Chr.1: 157099437-157099437 on Build GRCh38
rs894601470 Chr.1: 157099471-157099471 on Build GRCh38
rs751315681 Chr.1: 157099364-157099364 on Build GRCh38
rs16838075 Chr.1: 157099362-157099362 on Build GRCh38
rs199499287 Chr.1: 157099366-157099366 on Build GRCh38
rs773869771 Chr.1: 157099406-157099406 on Build GRCh38
rs112858973 Chr.1: 157099786-157099786 on Build GRCh38
rs780610906 Chr.1: 157099368-157099368 on Build GRCh38
rs769976233 Chr.1: 157099479-157099479 on Build GRCh38
rs771528327 Chr.1: 157099484-157099484 on Build GRCh38
rs901129324 Chr.1: 157099809-157099809 on Build GRCh38
rs770208936 Chr.1: 157099413-157099413 on Build GRCh38
rs977166422 Chr.1: 157099778-157099778 on Build GRCh38
rs74938633 Chr.1: 157099450-157099450 on Build GRCh38
rs757216253 Chr.1: 157099550-157099550 on Build GRCh38
rs1033509795 Chr.1: 157099629-157099629 on Build GRCh38
rs565665759 Chr.1: 157099745-157099745 on Build GRCh38
rs759146244 Chr.1: 157099440-157099440 on Build GRCh38
rs766899991 Chr.1: 157099430-157099430 on Build GRCh38
rs756373212 Chr.1: 157099573-157099573 on Build GRCh38
rs554389622 Chr.1: 157099634-157099634 on Build GRCh38
rs968936364 Chr.1: 157099752-157099752 on Build GRCh38
rs759889745 Chr.1: 157099508-157099508 on Build GRCh38
rs375452315 Chr.1: 157099442-157099442 on Build GRCh38
rs758282544 Chr.1: 157099562-157099562 on Build GRCh38
rs759817053 Chr.1: 157099451-157099451 on Build GRCh38
rs953779193 Chr.1: 157099663-157099663 on Build GRCh38
rs775276186 Chr.1: 157099522-157099522 on Build GRCh38
rs749083227 Chr.1: 157099389-157099389 on Build GRCh38
rs371844064 Chr.1: 157099800-157099800 on Build GRCh38
rs1027990712 Chr.1: 157099605-157099605 on Build GRCh38
rs373161167 Chr.1: 157099536-157099536 on Build GRCh38
rs571468238 Chr.1: 157099551-157099551 on Build GRCh38
rs777706159 Chr.1: 157099386-157099386 on Build GRCh38
rs752789582 Chr.1: 157099496-157099496 on Build GRCh38
rs16838078 Chr.1: 157099469-157099469 on Build GRCh38
rs576878216 Chr.1: 157099787-157099787 on Build GRCh38
rs531442967 Chr.1: 157099509-157099509 on Build GRCh38
rs75500098 Chr.1: 157099769-157099769 on Build GRCh38
rs755177088 Chr.1: 157099461-157099461 on Build GRCh38
rs149508240 Chr.1: 157099481-157099481 on Build GRCh38

View Full Product Details