Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161781827-161782332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF6
Entrez Gene ID
22926
Gene Name
activating transcription factor 6
Gene Aliases
ACHM7, ATF6A
UniGene
Hs.492740
Gene Chromosome Location
Chr.1: 161766244-161964070 on Build GRCh38
Transcripts
NM_007348.3 XM_006711224.1 XM_011509308.1 XM_011509309.1 XM_011509310.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272765 c.201G>T p.M67I Chr.1: 161781953-161781953 on Build GRCh38
3750535 c.199A>G p.M67V Chr.1: 161781951-161781951 on Build GRCh38
898605 c.247G>T p.V83F Chr.1: 161781999-161781999 on Build GRCh38
3418245 c.160G>A p.E54K Chr.1: 161781912-161781912 on Build GRCh38
1756838 c.160-2A>T p.? Chr.1: 161781910-161781910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968136920 Chr.1: 161782122-161782122 on Build GRCh38
rs72706043 Chr.1: 161782075-161782075 on Build GRCh38
rs1033701447 Chr.1: 161782142-161782142 on Build GRCh38
rs759324557 Chr.1: 161782015-161782015 on Build GRCh38
rs748580731 Chr.1: 161781945-161781945 on Build GRCh38
rs775097907 Chr.1: 161782019-161782019 on Build GRCh38
rs774498650 Chr.1: 161782005-161782005 on Build GRCh38
rs1135982 Chr.1: 161781953-161781953 on Build GRCh38
rs763859732 Chr.1: 161781863-161781863 on Build GRCh38
rs749575306 Chr.1: 161781989-161781989 on Build GRCh38
rs758122208 Chr.1: 161782037-161782037 on Build GRCh38
rs182140282 Chr.1: 161782294-161782294 on Build GRCh38
rs201520644 Chr.1: 161782017-161782017 on Build GRCh38
rs146083374 Chr.1: 161782278-161782278 on Build GRCh38
rs992139941 Chr.1: 161782154-161782154 on Build GRCh38
rs887975226 Chr.1: 161782244-161782244 on Build GRCh38
rs1020774524 Chr.1: 161782104-161782104 on Build GRCh38
rs1014991903 Chr.1: 161781986-161781986 on Build GRCh38
rs1040652145 Chr.1: 161781870-161781870 on Build GRCh38
rs373888481 Chr.1: 161781908-161781908 on Build GRCh38
rs765000484 Chr.1: 161782030-161782031 on Build GRCh38
rs144808590 Chr.1: 161782083-161782083 on Build GRCh38
rs551851042 Chr.1: 161781894-161781894 on Build GRCh38
rs751509864 Chr.1: 161782297-161782297 on Build GRCh38
rs756390478 Chr.1: 161781870-161781874 on Build GRCh38
rs886878754 Chr.1: 161781891-161781891 on Build GRCh38
rs932278208 Chr.1: 161782214-161782214 on Build GRCh38
rs764783403 Chr.1: 161782038-161782038 on Build GRCh38
rs562413208 Chr.1: 161782097-161782097 on Build GRCh38
rs112736858 Chr.1: 161782176-161782176 on Build GRCh38
rs761406989 Chr.1: 161782091-161782091 on Build GRCh38
rs757029182 Chr.1: 161782035-161782035 on Build GRCh38
rs748820567 Chr.1: 161782209-161782209 on Build GRCh38
rs778633759 Chr.1: 161781934-161781934 on Build GRCh38
rs959631349 Chr.1: 161782149-161782149 on Build GRCh38
rs753596270 Chr.1: 161781904-161781904 on Build GRCh38
rs1005962230 Chr.1: 161781974-161781974 on Build GRCh38
rs747457868 Chr.1: 161781939-161781939 on Build GRCh38
rs983598152 Chr.1: 161782231-161782231 on Build GRCh38
rs1050548015 Chr.1: 161782238-161782238 on Build GRCh38
rs990277255 Chr.1: 161782143-161782143 on Build GRCh38
rs750146012 Chr.1: 161782033-161782034 on Build GRCh38
rs144743551 Chr.1: 161781997-161781997 on Build GRCh38
rs746565808 Chr.1: 161781857-161781857 on Build GRCh38
rs913204831 Chr.1: 161782144-161782144 on Build GRCh38
rs370780754 Chr.1: 161781861-161781861 on Build GRCh38
rs923529041 Chr.1: 161782206-161782206 on Build GRCh38
rs532192842 Chr.1: 161781999-161781999 on Build GRCh38
rs760494664 Chr.1: 161782022-161782022 on Build GRCh38
rs773389442 Chr.1: 161781978-161781978 on Build GRCh38
rs138607905 Chr.1: 161781995-161781995 on Build GRCh38
rs899491046 Chr.1: 161781992-161781992 on Build GRCh38
rs189503932 Chr.1: 161782265-161782265 on Build GRCh38
rs547742427 Chr.1: 161782197-161782197 on Build GRCh38
rs377686597 Chr.1: 161781944-161781944 on Build GRCh38
rs755526870 Chr.1: 161781940-161781940 on Build GRCh38
rs1058405 Chr.1: 161781951-161781951 on Build GRCh38
rs764023093 Chr.1: 161782024-161782024 on Build GRCh38
rs779771526 Chr.1: 161782040-161782041 on Build GRCh38
rs138658313 Chr.1: 161782130-161782130 on Build GRCh38

View Full Product Details