Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 87236818-87237363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNBD1
Entrez Gene ID
168975
Gene Name
cyclic nucleotide binding domain containing 1
Gene Aliases
-
UniGene
Hs.246284
Gene Chromosome Location
Chr.8: 86866442-87428652 on Build GRCh38
Transcripts
NM_173538.2 XM_017013149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5358582 c.578-10delT p.? Chr.8: 87236909-87236909 on Build GRCh38
2789903 c.754C>A p.P252T Chr.8: 87237095-87237095 on Build GRCh38
2789901 c.739G>A p.A247T Chr.8: 87237080-87237080 on Build GRCh38
2789893 c.728A>G p.N243S Chr.8: 87237069-87237069 on Build GRCh38
3925681 c.758C>T p.S253L Chr.8: 87237099-87237099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919471294 Chr.8: 87237337-87237337 on Build GRCh38
rs1022385061 Chr.8: 87237294-87237294 on Build GRCh38
rs901781887 Chr.8: 87237234-87237234 on Build GRCh38
rs779617810 Chr.8: 87236954-87236954 on Build GRCh38
rs367726460 Chr.8: 87237023-87237023 on Build GRCh38
rs375254057 Chr.8: 87236992-87236992 on Build GRCh38
rs373719179 Chr.8: 87237244-87237244 on Build GRCh38
rs781442977 Chr.8: 87237063-87237063 on Build GRCh38
rs191023955 Chr.8: 87237170-87237170 on Build GRCh38
rs1031541960 Chr.8: 87237254-87237254 on Build GRCh38
rs751259462 Chr.8: 87236928-87236928 on Build GRCh38
rs115723202 Chr.8: 87236874-87236874 on Build GRCh38
rs569905871 Chr.8: 87237205-87237205 on Build GRCh38
rs376856009 Chr.8: 87236877-87236877 on Build GRCh38
rs561026350 Chr.8: 87236932-87236932 on Build GRCh38
rs1036518714 Chr.8: 87237124-87237124 on Build GRCh38
rs759171306 Chr.8: 87237099-87237099 on Build GRCh38
rs760362513 Chr.8: 87237113-87237113 on Build GRCh38
rs780194243 Chr.8: 87237049-87237049 on Build GRCh38
rs1011372877 Chr.8: 87237292-87237292 on Build GRCh38
rs767108876 Chr.8: 87237100-87237100 on Build GRCh38
rs200993835 Chr.8: 87236957-87236957 on Build GRCh38
rs571644256 Chr.8: 87236969-87236969 on Build GRCh38
rs770703761 Chr.8: 87237096-87237096 on Build GRCh38
rs34754092 Chr.8: 87237136-87237136 on Build GRCh38
rs767341880 Chr.8: 87237016-87237016 on Build GRCh38
rs182921012 Chr.8: 87237304-87237304 on Build GRCh38
rs182811386 Chr.8: 87236964-87236964 on Build GRCh38
rs765779447 Chr.8: 87236907-87236907 on Build GRCh38
rs961023448 Chr.8: 87237311-87237311 on Build GRCh38
rs943847962 Chr.8: 87237109-87237109 on Build GRCh38
rs768801750 Chr.8: 87236966-87236966 on Build GRCh38
rs772836598 Chr.8: 87237089-87237089 on Build GRCh38
rs778976241 Chr.8: 87236969-87236969 on Build GRCh38
rs1016238346 Chr.8: 87237287-87237297 on Build GRCh38
rs911980315 Chr.8: 87237326-87237327 on Build GRCh38
rs1003663889 Chr.8: 87237285-87237285 on Build GRCh38
rs899536039 Chr.8: 87236861-87236861 on Build GRCh38
rs538269843 Chr.8: 87237298-87237298 on Build GRCh38
rs752380725 Chr.8: 87237017-87237017 on Build GRCh38
rs60556175 Chr.8: 87237110-87237110 on Build GRCh38
rs761745228 Chr.8: 87236891-87236891 on Build GRCh38
rs776360066 Chr.8: 87236890-87236890 on Build GRCh38
rs780485864 Chr.8: 87237149-87237149 on Build GRCh38
rs930435241 Chr.8: 87237160-87237160 on Build GRCh38
rs375597027 Chr.8: 87237064-87237064 on Build GRCh38
rs370736889 Chr.8: 87236978-87236978 on Build GRCh38
rs758780978 Chr.8: 87237036-87237036 on Build GRCh38
rs772318363 Chr.8: 87237009-87237011 on Build GRCh38
rs1048507237 Chr.8: 87237147-87237147 on Build GRCh38
rs981143781 Chr.8: 87237069-87237069 on Build GRCh38
rs755086876 Chr.8: 87236908-87236909 on Build GRCh38
rs765939131 Chr.8: 87236985-87236985 on Build GRCh38
rs866621083 Chr.8: 87237224-87237224 on Build GRCh38
rs757688776 Chr.8: 87236946-87236946 on Build GRCh38
rs996915941 Chr.8: 87236864-87236864 on Build GRCh38
rs558961509 Chr.8: 87237214-87237214 on Build GRCh38
rs757608759 Chr.8: 87237051-87237051 on Build GRCh38
rs114244853 Chr.8: 87237123-87237123 on Build GRCh38
rs151276481 Chr.8: 87237156-87237156 on Build GRCh38
rs371272177 Chr.8: 87236887-87236887 on Build GRCh38
rs750314719 Chr.8: 87236904-87236904 on Build GRCh38
rs753378577 Chr.8: 87236908-87236908 on Build GRCh38
rs765227412 Chr.8: 87236893-87236893 on Build GRCh38
rs143210786 Chr.8: 87236896-87236896 on Build GRCh38
rs142384213 Chr.8: 87236871-87236871 on Build GRCh38
rs531680350 Chr.8: 87236963-87236963 on Build GRCh38
rs1041556655 Chr.8: 87237033-87237033 on Build GRCh38
rs957187299 Chr.8: 87237272-87237272 on Build GRCh38
rs1994834 Chr.8: 87237295-87237295 on Build GRCh38
rs764804468 Chr.8: 87236848-87236848 on Build GRCh38
rs373039361 Chr.8: 87236934-87236934 on Build GRCh38
rs982294821 Chr.8: 87236955-87236955 on Build GRCh38
rs779115343 Chr.8: 87237019-87237019 on Build GRCh38
rs577154569 Chr.8: 87236867-87236867 on Build GRCh38
rs780057407 Chr.8: 87236871-87236871 on Build GRCh38
rs370066698 Chr.8: 87237052-87237052 on Build GRCh38
rs754680070 Chr.8: 87237000-87237000 on Build GRCh38
rs1015721047 Chr.8: 87237240-87237240 on Build GRCh38
rs760776752 Chr.8: 87236886-87236886 on Build GRCh38
rs1040419725 Chr.8: 87237225-87237225 on Build GRCh38
rs769611950 Chr.8: 87237087-87237087 on Build GRCh38
rs765654545 Chr.8: 87237121-87237121 on Build GRCh38
rs748460241 Chr.8: 87236976-87236977 on Build GRCh38
rs970903913 Chr.8: 87237284-87237284 on Build GRCh38
rs754631029 Chr.8: 87236931-87236931 on Build GRCh38
rs368161700 Chr.8: 87237269-87237269 on Build GRCh38
rs566063414 Chr.8: 87237155-87237155 on Build GRCh38
rs537395017 Chr.8: 87237211-87237211 on Build GRCh38
rs972286742 Chr.8: 87237313-87237313 on Build GRCh38
rs998759171 Chr.8: 87237250-87237250 on Build GRCh38
rs757980108 Chr.8: 87236905-87236905 on Build GRCh38
rs888830891 Chr.8: 87237219-87237219 on Build GRCh38
rs761655174 Chr.8: 87236977-87236977 on Build GRCh38
rs767932359 Chr.8: 87237070-87237070 on Build GRCh38
rs758788306 Chr.8: 87237144-87237144 on Build GRCh38
rs1040743634 Chr.8: 87236850-87236850 on Build GRCh38
rs762963909 Chr.8: 87237092-87237092 on Build GRCh38
rs751897679 Chr.8: 87236982-87236982 on Build GRCh38
rs978616930 Chr.8: 87237317-87237317 on Build GRCh38

View Full Product Details