Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136489993-136490495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRS3
Entrez Gene ID
680
Gene Name
bombesin receptor subtype 3
Gene Aliases
BB3, BB3R
UniGene
Hs.121484
Gene Chromosome Location
Chr.X: 136487966-136492439 on Build GRCh38
Transcripts
NM_001727.1

SNP Detail

SNP ID Chromosome Location
rs201594031 Chr.X: 136490345-136490345 on Build GRCh38
rs5234 Chr.X: 136490183-136490183 on Build GRCh38
rs989597245 Chr.X: 136490301-136490301 on Build GRCh38
rs754915114 Chr.X: 136490056-136490056 on Build GRCh38
rs745804869 Chr.X: 136490220-136490220 on Build GRCh38
rs769565089 Chr.X: 136490101-136490101 on Build GRCh38
rs765593380 Chr.X: 136490386-136490386 on Build GRCh38
rs376273315 Chr.X: 136490442-136490442 on Build GRCh38
rs188653015 Chr.X: 136490033-136490033 on Build GRCh38
rs397705465 Chr.X: 136490066-136490067 on Build GRCh38
rs771020086 Chr.X: 136490271-136490271 on Build GRCh38
rs773884108 Chr.X: 136490094-136490094 on Build GRCh38
rs773971890 Chr.X: 136490273-136490273 on Build GRCh38
rs761016383 Chr.X: 136490107-136490107 on Build GRCh38
rs878863239 Chr.X: 136490230-136490230 on Build GRCh38
rs373773614 Chr.X: 136490093-136490093 on Build GRCh38
rs767845392 Chr.X: 136490449-136490449 on Build GRCh38
rs773654082 Chr.X: 136490322-136490322 on Build GRCh38
rs779667181 Chr.X: 136490122-136490123 on Build GRCh38
rs5235 Chr.X: 136490247-136490247 on Build GRCh38
rs752869358 Chr.X: 136490209-136490209 on Build GRCh38
rs371708251 Chr.X: 136490338-136490338 on Build GRCh38
rs964595064 Chr.X: 136490427-136490427 on Build GRCh38
rs906446771 Chr.X: 136490174-136490174 on Build GRCh38
rs994906106 Chr.X: 136490448-136490448 on Build GRCh38
rs367611736 Chr.X: 136490095-136490095 on Build GRCh38
rs761261309 Chr.X: 136490334-136490334 on Build GRCh38
rs748994645 Chr.X: 136490262-136490262 on Build GRCh38
rs140873258 Chr.X: 136490034-136490034 on Build GRCh38
rs768062989 Chr.X: 136490096-136490096 on Build GRCh38
rs781340230 Chr.X: 136490218-136490218 on Build GRCh38
rs957791519 Chr.X: 136490294-136490294 on Build GRCh38
rs200528448 Chr.X: 136490248-136490248 on Build GRCh38
rs761354565 Chr.X: 136490280-136490280 on Build GRCh38
rs766755736 Chr.X: 136490111-136490111 on Build GRCh38
rs150938152 Chr.X: 136490026-136490026 on Build GRCh38
rs192324772 Chr.X: 136490457-136490457 on Build GRCh38
rs774537704 Chr.X: 136490282-136490282 on Build GRCh38
rs924574930 Chr.X: 136490025-136490025 on Build GRCh38
rs779226345 Chr.X: 136490203-136490203 on Build GRCh38
rs373752983 Chr.X: 136490065-136490066 on Build GRCh38
rs202114962 Chr.X: 136490217-136490217 on Build GRCh38
rs759794275 Chr.X: 136490368-136490368 on Build GRCh38
rs769559411 Chr.X: 136490226-136490226 on Build GRCh38
rs773147695 Chr.X: 136490119-136490119 on Build GRCh38
rs3030366 Chr.X: 136490064-136490065 on Build GRCh38

View Full Product Details