Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 1177989-1178488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB2
Entrez Gene ID
105
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Gene Aliases
ADAR3, RED2
UniGene
Hs.586663
Gene Chromosome Location
Chr.10: 1177313-1737476 on Build GRCh38
Transcripts
NM_018702.3

SNP Detail

SNP ID Chromosome Location
rs950570126 Chr.10: 1178260-1178260 on Build GRCh38
rs965866234 Chr.10: 1178034-1178034 on Build GRCh38
rs567771408 Chr.10: 1178340-1178340 on Build GRCh38
rs60078522 Chr.10: 1178277-1178277 on Build GRCh38
rs757080078 Chr.10: 1178456-1178456 on Build GRCh38
rs900977943 Chr.10: 1178176-1178176 on Build GRCh38
rs61374726 Chr.10: 1178278-1178278 on Build GRCh38
rs908875855 Chr.10: 1178065-1178065 on Build GRCh38
rs913392408 Chr.10: 1178394-1178394 on Build GRCh38
rs751008865 Chr.10: 1178104-1178104 on Build GRCh38
rs994291648 Chr.10: 1178192-1178192 on Build GRCh38
rs984730495 Chr.10: 1178056-1178056 on Build GRCh38
rs573422651 Chr.10: 1178419-1178419 on Build GRCh38
rs199836862 Chr.10: 1178159-1178159 on Build GRCh38
rs369792357 Chr.10: 1178457-1178457 on Build GRCh38
rs888226932 Chr.10: 1178101-1178101 on Build GRCh38
rs754474216 Chr.10: 1178113-1178113 on Build GRCh38
rs916176928 Chr.10: 1178338-1178338 on Build GRCh38
rs34033894 Chr.10: 1178078-1178079 on Build GRCh38
rs560693711 Chr.10: 1178026-1178026 on Build GRCh38
rs1043250123 Chr.10: 1178397-1178397 on Build GRCh38
rs979845694 Chr.10: 1178103-1178103 on Build GRCh38
rs911491801 Chr.10: 1178398-1178398 on Build GRCh38
rs386739807 Chr.10: 1178310-1178311 on Build GRCh38
rs889748100 Chr.10: 1178172-1178172 on Build GRCh38
rs977470009 Chr.10: 1178291-1178291 on Build GRCh38
rs571598223 Chr.10: 1178194-1178194 on Build GRCh38
rs777505724 Chr.10: 1178380-1178380 on Build GRCh38
rs767065019 Chr.10: 1178314-1178314 on Build GRCh38
rs948990477 Chr.10: 1178371-1178371 on Build GRCh38
rs1041111295 Chr.10: 1178190-1178190 on Build GRCh38
rs200190573 Chr.10: 1178259-1178261 on Build GRCh38
rs748536127 Chr.10: 1178149-1178150 on Build GRCh38
rs938841688 Chr.10: 1178111-1178111 on Build GRCh38
rs992992135 Chr.10: 1178336-1178336 on Build GRCh38
rs546396561 Chr.10: 1178048-1178048 on Build GRCh38
rs947634797 Chr.10: 1178148-1178148 on Build GRCh38
rs201324201 Chr.10: 1178151-1178151 on Build GRCh38
rs980196623 Chr.10: 1178307-1178307 on Build GRCh38
rs188944007 Chr.10: 1178182-1178182 on Build GRCh38
rs762604787 Chr.10: 1178141-1178141 on Build GRCh38
rs537493160 Chr.10: 1178289-1178289 on Build GRCh38
rs1006965124 Chr.10: 1178020-1178020 on Build GRCh38
rs34381966 Chr.10: 1178156-1178157 on Build GRCh38
rs529717155 Chr.10: 1178191-1178191 on Build GRCh38
rs950601316 Chr.10: 1178049-1178049 on Build GRCh38
rs762255962 Chr.10: 1178018-1178018 on Build GRCh38
rs1048045512 Chr.10: 1178079-1178079 on Build GRCh38
rs938923798 Chr.10: 1178022-1178022 on Build GRCh38
rs752277438 Chr.10: 1178137-1178137 on Build GRCh38
rs939779641 Chr.10: 1178387-1178387 on Build GRCh38
rs115469149 Chr.10: 1178290-1178290 on Build GRCh38
rs868688497 Chr.10: 1178332-1178332 on Build GRCh38
rs752316968 Chr.10: 1178331-1178331 on Build GRCh38
rs969745994 Chr.10: 1178074-1178074 on Build GRCh38
rs1007305617 Chr.10: 1178185-1178185 on Build GRCh38
rs756534680 Chr.10: 1178304-1178304 on Build GRCh38
rs1026134914 Chr.10: 1178203-1178203 on Build GRCh38
rs1041429370 Chr.10: 1178413-1178413 on Build GRCh38
rs1006135787 Chr.10: 1178280-1178280 on Build GRCh38
rs879327000 Chr.10: 1178066-1178066 on Build GRCh38
rs942687656 Chr.10: 1178411-1178411 on Build GRCh38
rs532454262 Chr.10: 1178149-1178149 on Build GRCh38
rs913425414 Chr.10: 1178126-1178126 on Build GRCh38
rs183891805 Chr.10: 1178046-1178046 on Build GRCh38
rs6560716 Chr.10: 1178396-1178396 on Build GRCh38
rs77767853 Chr.10: 1178156-1178156 on Build GRCh38
rs192936934 Chr.10: 1178384-1178384 on Build GRCh38
rs566600558 Chr.10: 1178136-1178136 on Build GRCh38
rs1036710257 Chr.10: 1178393-1178393 on Build GRCh38
rs36093407 Chr.10: 1178150-1178151 on Build GRCh38

View Full Product Details