Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84201718-84202220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR1
Entrez Gene ID
92211
Gene Name
cadherin related family member 1
Gene Aliases
CORD15, PCDH21, PRCAD, RP65
UniGene
Hs.137556
Gene Chromosome Location
Chr.10: 84194635-84219621 on Build GRCh38
Transcripts
NM_001171971.2 NM_033100.3 XM_011540337.1 XM_011540338.1 XM_011540339.1 XM_011540340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807834 c.526-1G>A p.? Chr.10: 84201806-84201806 on Build GRCh38
4016305 c.639G>T p.K213N Chr.10: 84201920-84201920 on Build GRCh38
4554144 c.612G>A p.R204R Chr.10: 84201893-84201893 on Build GRCh38
539922 c.554G>A p.R185H Chr.10: 84201835-84201835 on Build GRCh38
539920 c.572G>C p.R191P Chr.10: 84201853-84201853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780642266 Chr.10: 84201799-84201799 on Build GRCh38
rs763409556 Chr.10: 84201904-84201904 on Build GRCh38
rs779510340 Chr.10: 84201746-84201746 on Build GRCh38
rs139313264 Chr.10: 84201881-84201881 on Build GRCh38
rs759140940 Chr.10: 84201851-84201851 on Build GRCh38
rs1042988195 Chr.10: 84202125-84202125 on Build GRCh38
rs756128159 Chr.10: 84201803-84201803 on Build GRCh38
rs535375339 Chr.10: 84201962-84201962 on Build GRCh38
rs758112482 Chr.10: 84201952-84201952 on Build GRCh38
rs899614534 Chr.10: 84201794-84201794 on Build GRCh38
rs561927881 Chr.10: 84202149-84202149 on Build GRCh38
rs890758133 Chr.10: 84201985-84201985 on Build GRCh38
rs777542147 Chr.10: 84201804-84201804 on Build GRCh38
rs939262359 Chr.10: 84201742-84201742 on Build GRCh38
rs768976866 Chr.10: 84201834-84201834 on Build GRCh38
rs773578231 Chr.10: 84201838-84201838 on Build GRCh38
rs758756753 Chr.10: 84201778-84201778 on Build GRCh38
rs147420731 Chr.10: 84201837-84201837 on Build GRCh38
rs747165743 Chr.10: 84201827-84201827 on Build GRCh38
rs759864852 Chr.10: 84201913-84201913 on Build GRCh38
rs190906755 Chr.10: 84201800-84201800 on Build GRCh38
rs939364566 Chr.10: 84201854-84201854 on Build GRCh38
rs781263335 Chr.10: 84201891-84201891 on Build GRCh38
rs765638275 Chr.10: 84201768-84201768 on Build GRCh38
rs879751360 Chr.10: 84201745-84201745 on Build GRCh38
rs769938919 Chr.10: 84201936-84201936 on Build GRCh38
rs974344308 Chr.10: 84201997-84201997 on Build GRCh38
rs142631189 Chr.10: 84201853-84201853 on Build GRCh38
rs962625789 Chr.10: 84202089-84202089 on Build GRCh38
rs751154635 Chr.10: 84201790-84201790 on Build GRCh38
rs140328843 Chr.10: 84202183-84202183 on Build GRCh38
rs745955360 Chr.10: 84201793-84201793 on Build GRCh38
rs748412274 Chr.10: 84201843-84201843 on Build GRCh38
rs756248913 Chr.10: 84201969-84201969 on Build GRCh38
rs745932099 Chr.10: 84201920-84201920 on Build GRCh38
rs760249323 Chr.10: 84201923-84201923 on Build GRCh38
rs188744482 Chr.10: 84201993-84201993 on Build GRCh38
rs908681006 Chr.10: 84201991-84201991 on Build GRCh38
rs373616602 Chr.10: 84201925-84201925 on Build GRCh38
rs986973475 Chr.10: 84201978-84201978 on Build GRCh38
rs908453789 Chr.10: 84202083-84202083 on Build GRCh38
rs932479139 Chr.10: 84202158-84202159 on Build GRCh38
rs755804592 Chr.10: 84202072-84202072 on Build GRCh38
rs35205289 Chr.10: 84201936-84201937 on Build GRCh38
rs974409155 Chr.10: 84202141-84202141 on Build GRCh38
rs767268640 Chr.10: 84201914-84201914 on Build GRCh38
rs955920045 Chr.10: 84201789-84201789 on Build GRCh38
rs764094310 Chr.10: 84202118-84202118 on Build GRCh38
rs200646223 Chr.10: 84201882-84201882 on Build GRCh38
rs762564825 Chr.10: 84202142-84202142 on Build GRCh38
rs199671784 Chr.10: 84201876-84201876 on Build GRCh38
rs145290418 Chr.10: 84201937-84201938 on Build GRCh38
rs747290719 Chr.10: 84201829-84201829 on Build GRCh38
rs773773216 Chr.10: 84201900-84201900 on Build GRCh38
rs201731584 Chr.10: 84201835-84201835 on Build GRCh38
rs1028446487 Chr.10: 84201938-84201938 on Build GRCh38
rs927854245 Chr.10: 84201740-84201740 on Build GRCh38
rs938407156 Chr.10: 84202047-84202047 on Build GRCh38
rs981284030 Chr.10: 84201849-84201849 on Build GRCh38
rs150979616 Chr.10: 84201860-84201860 on Build GRCh38
rs376739586 Chr.10: 84201887-84201887 on Build GRCh38
rs769702557 Chr.10: 84201892-84201892 on Build GRCh38
rs750307646 Chr.10: 84201948-84201948 on Build GRCh38
rs12415295 Chr.10: 84202044-84202044 on Build GRCh38
rs1022440260 Chr.10: 84202032-84202032 on Build GRCh38
rs199825352 Chr.10: 84201842-84201842 on Build GRCh38
rs200914741 Chr.10: 84201908-84201908 on Build GRCh38
rs1003113889 Chr.10: 84201758-84201758 on Build GRCh38
rs377408181 Chr.10: 84201769-84201769 on Build GRCh38
rs377285624 Chr.10: 84201871-84201871 on Build GRCh38
rs74145721 Chr.10: 84202058-84202058 on Build GRCh38
rs770839275 Chr.10: 84201820-84201820 on Build GRCh38
rs184241207 Chr.10: 84201823-84201823 on Build GRCh38
rs774747506 Chr.10: 84201909-84201909 on Build GRCh38
rs755578389 Chr.10: 84201856-84201856 on Build GRCh38
rs200880106 Chr.10: 84201915-84201915 on Build GRCh38
rs142109115 Chr.10: 84201828-84201828 on Build GRCh38
rs375359797 Chr.10: 84201942-84201942 on Build GRCh38
rs900414035 Chr.10: 84202132-84202132 on Build GRCh38
rs770348342 Chr.10: 84201870-84201870 on Build GRCh38
rs528514743 Chr.10: 84201812-84201812 on Build GRCh38
rs192787878 Chr.10: 84201999-84201999 on Build GRCh38
rs781489568 Chr.10: 84201958-84201958 on Build GRCh38
rs956854277 Chr.10: 84201831-84201831 on Build GRCh38
rs765231249 Chr.10: 84201943-84201943 on Build GRCh38
rs547134587 Chr.10: 84202033-84202033 on Build GRCh38
rs151204356 Chr.10: 84201852-84201852 on Build GRCh38
rs74145722 Chr.10: 84202190-84202190 on Build GRCh38
rs934208979 Chr.10: 84201839-84201839 on Build GRCh38
rs921071844 Chr.10: 84202155-84202155 on Build GRCh38
rs11200922 Chr.10: 84202002-84202002 on Build GRCh38
rs766076823 Chr.10: 84201779-84201779 on Build GRCh38
rs1052851841 Chr.10: 84201905-84201905 on Build GRCh38

View Full Product Details