Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122855556-122856065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM24B
Entrez Gene ID
196792
Gene Name
family with sequence similarity 24 member B
Gene Aliases
-
UniGene
Hs.114648
Gene Chromosome Location
Chr.10: 122849094-122879641 on Build GRCh38
Transcripts
NM_001204364.1 NM_152644.2 NR_037911.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_001204364.1 NM_152644.2 NR_037911.1 NR_037915.1

SNP Detail

SNP ID Chromosome Location
rs1046757008 Chr.10: 122855641-122855641 on Build GRCh38
rs935885915 Chr.10: 122855734-122855734 on Build GRCh38
rs144888191 Chr.10: 122855911-122855911 on Build GRCh38
rs578156183 Chr.10: 122855924-122855924 on Build GRCh38
rs1028483430 Chr.10: 122855909-122855909 on Build GRCh38
rs1004251422 Chr.10: 122855837-122855837 on Build GRCh38
rs1038648646 Chr.10: 122855842-122855842 on Build GRCh38
rs371416421 Chr.10: 122856011-122856011 on Build GRCh38
rs966745140 Chr.10: 122855966-122855966 on Build GRCh38
rs919497875 Chr.10: 122855598-122855598 on Build GRCh38
rs147512042 Chr.10: 122855937-122855937 on Build GRCh38
rs979835681 Chr.10: 122855998-122855998 on Build GRCh38
rs912320019 Chr.10: 122855645-122855645 on Build GRCh38
rs1023036443 Chr.10: 122855895-122855895 on Build GRCh38
rs915688612 Chr.10: 122855797-122855797 on Build GRCh38
rs921022556 Chr.10: 122856010-122856010 on Build GRCh38
rs1028452154 Chr.10: 122855585-122855585 on Build GRCh38
rs188041530 Chr.10: 122855633-122855633 on Build GRCh38
rs955425276 Chr.10: 122855610-122855610 on Build GRCh38
rs575303558 Chr.10: 122856041-122856041 on Build GRCh38
rs949813505 Chr.10: 122855799-122855799 on Build GRCh38
rs899633304 Chr.10: 122855744-122855744 on Build GRCh38
rs1021013458 Chr.10: 122855951-122855951 on Build GRCh38
rs886779526 Chr.10: 122855807-122855807 on Build GRCh38
rs191358220 Chr.10: 122855724-122855724 on Build GRCh38
rs998822308 Chr.10: 122855788-122855788 on Build GRCh38
rs549541877 Chr.10: 122855967-122855967 on Build GRCh38
rs750306225 Chr.10: 122855921-122855921 on Build GRCh38
rs1011392764 Chr.10: 122855754-122855754 on Build GRCh38
rs1045431092 Chr.10: 122855806-122855806 on Build GRCh38
rs1042864575 Chr.10: 122856030-122856030 on Build GRCh38
rs563352227 Chr.10: 122855752-122855752 on Build GRCh38
rs913917638 Chr.10: 122855646-122855646 on Build GRCh38
rs898740147 Chr.10: 122855845-122855845 on Build GRCh38
rs977371274 Chr.10: 122855681-122855681 on Build GRCh38
rs997049177 Chr.10: 122855876-122855876 on Build GRCh38
rs1043407005 Chr.10: 122855768-122855768 on Build GRCh38
rs74163513 Chr.10: 122855775-122855775 on Build GRCh38
rs967092298 Chr.10: 122855664-122855664 on Build GRCh38
rs943781103 Chr.10: 122855971-122855972 on Build GRCh38
rs1039574607 Chr.10: 122855732-122855732 on Build GRCh38
rs183885729 Chr.10: 122855745-122855745 on Build GRCh38
rs779816451 Chr.10: 122855961-122855961 on Build GRCh38
rs963903339 Chr.10: 122855907-122855907 on Build GRCh38
rs975187325 Chr.10: 122855982-122855982 on Build GRCh38

View Full Product Details