Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32588671-32589165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926368 c.345G>T p.K115N Chr.11: 32589042-32589042 on Build GRCh38
244230 c.360A>C p.R120S Chr.11: 32589057-32589057 on Build GRCh38
5626038 c.381T>G p.L127L Chr.11: 32589078-32589078 on Build GRCh38
5694987 c.334G>A p.G112R Chr.11: 32589031-32589031 on Build GRCh38
4393002 c.309G>A p.L103L Chr.11: 32588727-32588727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049742952 Chr.11: 32588860-32588860 on Build GRCh38
rs187873377 Chr.11: 32588906-32588906 on Build GRCh38
rs544993751 Chr.11: 32588788-32588788 on Build GRCh38
rs538625850 Chr.11: 32588870-32588870 on Build GRCh38
rs768121933 Chr.11: 32588738-32588739 on Build GRCh38
rs995662041 Chr.11: 32588879-32588879 on Build GRCh38
rs983768593 Chr.11: 32588923-32588923 on Build GRCh38
rs959797630 Chr.11: 32588926-32588926 on Build GRCh38
rs548130009 Chr.11: 32588847-32588847 on Build GRCh38
rs569935921 Chr.11: 32588857-32588857 on Build GRCh38
rs759393884 Chr.11: 32589021-32589021 on Build GRCh38
rs936701719 Chr.11: 32588888-32588888 on Build GRCh38
rs11031885 Chr.11: 32588921-32588921 on Build GRCh38
rs975455352 Chr.11: 32588865-32588865 on Build GRCh38
rs778496148 Chr.11: 32588772-32588772 on Build GRCh38
rs779629980 Chr.11: 32588774-32588774 on Build GRCh38
rs371010742 Chr.11: 32588700-32588700 on Build GRCh38
rs764132923 Chr.11: 32588996-32588996 on Build GRCh38
rs200317760 Chr.11: 32589061-32589061 on Build GRCh38
rs765290768 Chr.11: 32589012-32589012 on Build GRCh38
rs760813548 Chr.11: 32588711-32588711 on Build GRCh38
rs777430050 Chr.11: 32588762-32588762 on Build GRCh38
rs754828246 Chr.11: 32588745-32588745 on Build GRCh38
rs140562244 Chr.11: 32589071-32589071 on Build GRCh38
rs750405547 Chr.11: 32589004-32589004 on Build GRCh38
rs906688570 Chr.11: 32588957-32588957 on Build GRCh38
rs777148441 Chr.11: 32588694-32588694 on Build GRCh38
rs898887092 Chr.11: 32588995-32588995 on Build GRCh38
rs373235607 Chr.11: 32588788-32588791 on Build GRCh38
rs749149691 Chr.11: 32588790-32588791 on Build GRCh38
rs772318051 Chr.11: 32588705-32588705 on Build GRCh38
rs1027674370 Chr.11: 32588915-32588915 on Build GRCh38
rs761951404 Chr.11: 32589005-32589005 on Build GRCh38
rs776807796 Chr.11: 32588986-32588986 on Build GRCh38
rs763878201 Chr.11: 32589036-32589036 on Build GRCh38
rs374422722 Chr.11: 32588715-32588715 on Build GRCh38
rs979776609 Chr.11: 32588730-32588730 on Build GRCh38
rs780588966 Chr.11: 32589121-32589121 on Build GRCh38
rs747377742 Chr.11: 32589138-32589138 on Build GRCh38
rs1026243693 Chr.11: 32588801-32588801 on Build GRCh38
rs547173819 Chr.11: 32588835-32588835 on Build GRCh38
rs755749703 Chr.11: 32588758-32588758 on Build GRCh38
rs755915170 Chr.11: 32589031-32589031 on Build GRCh38
rs796395743 Chr.11: 32588908-32588908 on Build GRCh38
rs868284578 Chr.11: 32589066-32589066 on Build GRCh38
rs201877 Chr.11: 32588842-32588842 on Build GRCh38
rs748856256 Chr.11: 32588765-32588765 on Build GRCh38
rs748762443 Chr.11: 32588699-32588699 on Build GRCh38
rs780860172 Chr.11: 32588753-32588753 on Build GRCh38
rs187882113 Chr.11: 32588834-32588834 on Build GRCh38
rs192168173 Chr.11: 32588977-32588977 on Build GRCh38
rs531170953 Chr.11: 32588727-32588727 on Build GRCh38
rs772565763 Chr.11: 32588781-32588781 on Build GRCh38
rs988604545 Chr.11: 32588830-32588830 on Build GRCh38
rs764338329 Chr.11: 32588714-32588714 on Build GRCh38
rs118109062 Chr.11: 32589041-32589041 on Build GRCh38
rs201519949 Chr.11: 32588693-32588693 on Build GRCh38
rs752585370 Chr.11: 32588767-32588767 on Build GRCh38
rs184536848 Chr.11: 32588885-32588885 on Build GRCh38
rs755782729 Chr.11: 32588733-32588734 on Build GRCh38
rs752467349 Chr.11: 32588754-32588754 on Build GRCh38
rs753534895 Chr.11: 32589038-32589038 on Build GRCh38
rs771497291 Chr.11: 32589140-32589141 on Build GRCh38
rs761863631 Chr.11: 32588724-32588724 on Build GRCh38
rs748780627 Chr.11: 32588737-32588738 on Build GRCh38
rs751496316 Chr.11: 32588741-32588741 on Build GRCh38
rs564803222 Chr.11: 32588797-32588797 on Build GRCh38
rs965467339 Chr.11: 32588855-32588855 on Build GRCh38
rs140524517 Chr.11: 32588698-32588698 on Build GRCh38
rs760017065 Chr.11: 32588701-32588701 on Build GRCh38
rs867782695 Chr.11: 32588712-32588712 on Build GRCh38
rs779904593 Chr.11: 32588736-32588737 on Build GRCh38
rs192453112 Chr.11: 32588841-32588841 on Build GRCh38
rs891212644 Chr.11: 32588861-32588861 on Build GRCh38
rs897300187 Chr.11: 32588878-32588878 on Build GRCh38
rs746362517 Chr.11: 32588780-32588780 on Build GRCh38
rs544647585 Chr.11: 32589048-32589048 on Build GRCh38
rs117913882 Chr.11: 32588905-32588905 on Build GRCh38
rs769065555 Chr.11: 32589143-32589143 on Build GRCh38
rs775750589 Chr.11: 32588964-32588964 on Build GRCh38
rs775868574 Chr.11: 32588710-32588710 on Build GRCh38
rs115236034 Chr.11: 32588979-32588979 on Build GRCh38
rs752452780 Chr.11: 32589030-32589030 on Build GRCh38
rs751697287 Chr.11: 32589020-32589020 on Build GRCh38
rs770533017 Chr.11: 32588772-32588772 on Build GRCh38
rs374272570 Chr.11: 32588919-32588919 on Build GRCh38
rs747404370 Chr.11: 32588985-32588988 on Build GRCh38
rs749045406 Chr.11: 32588901-32588901 on Build GRCh38

View Full Product Details