Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32600440-32600938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7232629 c.906G>C p.Q302H Chr.11: 32600795-32600795 on Build GRCh38
4435334 c.831G>A p.M277I Chr.11: 32600720-32600720 on Build GRCh38
5734467 c.873A>T p.E291D Chr.11: 32600762-32600762 on Build GRCh38
5017045 c.882T>A p.F294L Chr.11: 32600771-32600771 on Build GRCh38
926373 c.800-1G>T p.? Chr.11: 32600688-32600688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374728625 Chr.11: 32600678-32600678 on Build GRCh38
rs11031888 Chr.11: 32600667-32600667 on Build GRCh38
rs773821094 Chr.11: 32600831-32600831 on Build GRCh38
rs1024095342 Chr.11: 32600669-32600669 on Build GRCh38
rs746226718 Chr.11: 32600800-32600801 on Build GRCh38
rs890507007 Chr.11: 32600604-32600604 on Build GRCh38
rs780809253 Chr.11: 32600675-32600675 on Build GRCh38
rs909835116 Chr.11: 32600478-32600478 on Build GRCh38
rs767736836 Chr.11: 32600875-32600875 on Build GRCh38
rs1005019738 Chr.11: 32600632-32600632 on Build GRCh38
rs758773505 Chr.11: 32600868-32600869 on Build GRCh38
rs866389469 Chr.11: 32600720-32600720 on Build GRCh38
rs927718352 Chr.11: 32600864-32600864 on Build GRCh38
rs867976365 Chr.11: 32600833-32600833 on Build GRCh38
rs142909545 Chr.11: 32600809-32600809 on Build GRCh38
rs367544364 Chr.11: 32600679-32600679 on Build GRCh38
rs1032311248 Chr.11: 32600624-32600624 on Build GRCh38
rs915108894 Chr.11: 32600907-32600907 on Build GRCh38
rs979764462 Chr.11: 32600766-32600766 on Build GRCh38
rs759680408 Chr.11: 32600868-32600868 on Build GRCh38
rs757172794 Chr.11: 32600707-32600707 on Build GRCh38
rs981767641 Chr.11: 32600855-32600855 on Build GRCh38
rs7121241 Chr.11: 32600519-32600519 on Build GRCh38
rs574103017 Chr.11: 32600531-32600531 on Build GRCh38
rs770200418 Chr.11: 32600831-32600832 on Build GRCh38
rs746656425 Chr.11: 32600784-32600784 on Build GRCh38
rs868817359 Chr.11: 32600832-32600832 on Build GRCh38
rs1007605568 Chr.11: 32600607-32600607 on Build GRCh38
rs773739161 Chr.11: 32600665-32600665 on Build GRCh38
rs543227525 Chr.11: 32600577-32600577 on Build GRCh38
rs779742418 Chr.11: 32600780-32600780 on Build GRCh38
rs548110979 Chr.11: 32600592-32600592 on Build GRCh38
rs779032979 Chr.11: 32600536-32600536 on Build GRCh38
rs868256741 Chr.11: 32600687-32600687 on Build GRCh38
rs192472717 Chr.11: 32600499-32600499 on Build GRCh38
rs1039046754 Chr.11: 32600608-32600608 on Build GRCh38
rs532065868 Chr.11: 32600619-32600619 on Build GRCh38
rs540367937 Chr.11: 32600514-32600514 on Build GRCh38
rs544206381 Chr.11: 32600622-32600622 on Build GRCh38
rs563006123 Chr.11: 32600610-32600610 on Build GRCh38
rs374872378 Chr.11: 32600805-32600805 on Build GRCh38
rs371343128 Chr.11: 32600709-32600709 on Build GRCh38
rs933791406 Chr.11: 32600590-32600590 on Build GRCh38
rs767506307 Chr.11: 32600680-32600680 on Build GRCh38
rs766647020 Chr.11: 32600480-32600480 on Build GRCh38
rs113796540 Chr.11: 32600813-32600813 on Build GRCh38
rs750259913 Chr.11: 32600764-32600764 on Build GRCh38
rs768291113 Chr.11: 32600799-32600799 on Build GRCh38
rs763473605 Chr.11: 32600841-32600841 on Build GRCh38
rs771405773 Chr.11: 32600848-32600848 on Build GRCh38
rs78916077 Chr.11: 32600866-32600866 on Build GRCh38
rs904806129 Chr.11: 32600611-32600611 on Build GRCh38
rs752753754 Chr.11: 32600685-32600685 on Build GRCh38
rs758185130 Chr.11: 32600770-32600770 on Build GRCh38
rs770475847 Chr.11: 32600829-32600829 on Build GRCh38
rs745694522 Chr.11: 32600708-32600708 on Build GRCh38
rs918467090 Chr.11: 32600837-32600837 on Build GRCh38

View Full Product Details