Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114582205-114582710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE2
Entrez Gene ID
120406
Gene Name
neurexophilin and PC-esterase domain family member 2
Gene Aliases
FAM55B
UniGene
Hs.446760
Gene Chromosome Location
Chr.11: 114464276-114813702 on Build GRCh38
Transcripts
XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 NM_001077639.1 NM_017678.2 XM_006718865.3 XM_011542881.2 XM_011542882.2 XM_011542883.2

Gene Symbol
NXPE4
Entrez Gene ID
54827
Gene Name
neurexophilin and PC-esterase domain family member 4
Gene Aliases
C11orf33, FAM55D
UniGene
Hs.179100
Gene Chromosome Location
Chr.11: 114570591-114596215 on Build GRCh38
Transcripts
XM_017017207.1 XM_017017208.1 XM_017017209.1 XM_017017210.1 NM_001077639.1 NM_017678.2 XM_006718865.3 XM_011542881.2 XM_011542882.2 XM_011542883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251919 c.449G>A p.G150D Chr.11: 114582669-114582669 on Build GRCh38
5453080 c.419C>T p.A140V Chr.11: 114582699-114582699 on Build GRCh38
6131335 c.606G>A p.V202V Chr.11: 114582512-114582512 on Build GRCh38
2103922 c.538C>T p.L180L Chr.11: 114582580-114582580 on Build GRCh38
70720 c.823T>C p.F275L Chr.11: 114582295-114582295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573837301 Chr.11: 114582281-114582281 on Build GRCh38
rs754295764 Chr.11: 114582685-114582685 on Build GRCh38
rs769071982 Chr.11: 114582390-114582390 on Build GRCh38
rs771763637 Chr.11: 114582298-114582298 on Build GRCh38
rs565606316 Chr.11: 114582341-114582341 on Build GRCh38
rs1004302011 Chr.11: 114582660-114582660 on Build GRCh38
rs982557099 Chr.11: 114582294-114582294 on Build GRCh38
rs570103651 Chr.11: 114582595-114582595 on Build GRCh38
rs757155010 Chr.11: 114582474-114582474 on Build GRCh38
rs763688583 Chr.11: 114582331-114582331 on Build GRCh38
rs114870261 Chr.11: 114582372-114582372 on Build GRCh38
rs772466532 Chr.11: 114582321-114582322 on Build GRCh38
rs756270770 Chr.11: 114582370-114582370 on Build GRCh38
rs765390878 Chr.11: 114582311-114582311 on Build GRCh38
rs199976393 Chr.11: 114582620-114582620 on Build GRCh38
rs370800079 Chr.11: 114582615-114582615 on Build GRCh38
rs201191135 Chr.11: 114582263-114582263 on Build GRCh38
rs369782046 Chr.11: 114582599-114582599 on Build GRCh38
rs761735843 Chr.11: 114582310-114582310 on Build GRCh38
rs746673669 Chr.11: 114582489-114582489 on Build GRCh38
rs749870159 Chr.11: 114582654-114582654 on Build GRCh38
rs892503497 Chr.11: 114582438-114582438 on Build GRCh38
rs753626303 Chr.11: 114582471-114582471 on Build GRCh38
rs745965247 Chr.11: 114582639-114582639 on Build GRCh38
rs1020325592 Chr.11: 114582355-114582355 on Build GRCh38
rs764525259 Chr.11: 114582587-114582587 on Build GRCh38
rs192956484 Chr.11: 114582667-114582667 on Build GRCh38
rs549986778 Chr.11: 114582563-114582563 on Build GRCh38
rs200052294 Chr.11: 114582307-114582307 on Build GRCh38
rs113447804 Chr.11: 114582329-114582329 on Build GRCh38
rs770373330 Chr.11: 114582516-114582516 on Build GRCh38
rs766253958 Chr.11: 114582666-114582666 on Build GRCh38
rs762949140 Chr.11: 114582593-114582593 on Build GRCh38
rs200548586 Chr.11: 114582681-114582681 on Build GRCh38
rs756550760 Chr.11: 114582622-114582622 on Build GRCh38
rs748551809 Chr.11: 114582510-114582510 on Build GRCh38
rs184063316 Chr.11: 114582367-114582367 on Build GRCh38
rs758007169 Chr.11: 114582374-114582374 on Build GRCh38
rs753809110 Chr.11: 114582352-114582352 on Build GRCh38
rs144600200 Chr.11: 114582235-114582235 on Build GRCh38
rs868796970 Chr.11: 114582533-114582533 on Build GRCh38
rs761620452 Chr.11: 114582575-114582575 on Build GRCh38
rs372688755 Chr.11: 114582399-114582399 on Build GRCh38
rs775972370 Chr.11: 114582414-114582414 on Build GRCh38
rs572641123 Chr.11: 114582549-114582549 on Build GRCh38
rs780727481 Chr.11: 114582617-114582617 on Build GRCh38
rs761532668 Chr.11: 114582655-114582655 on Build GRCh38
rs754632542 Chr.11: 114582498-114582498 on Build GRCh38
rs766649665 Chr.11: 114582422-114582422 on Build GRCh38
rs750621625 Chr.11: 114582314-114582314 on Build GRCh38
rs763144364 Chr.11: 114582421-114582421 on Build GRCh38
rs749798709 Chr.11: 114582629-114582629 on Build GRCh38
rs776448352 Chr.11: 114582568-114582568 on Build GRCh38
rs769575246 Chr.11: 114582656-114582656 on Build GRCh38
rs752391000 Chr.11: 114582680-114582680 on Build GRCh38
rs374528575 Chr.11: 114582306-114582306 on Build GRCh38
rs777788051 Chr.11: 114582264-114582264 on Build GRCh38
rs752576681 Chr.11: 114582255-114582255 on Build GRCh38
rs755950047 Chr.11: 114582335-114582335 on Build GRCh38
rs778437339 Chr.11: 114582521-114582521 on Build GRCh38
rs779707730 Chr.11: 114582376-114582376 on Build GRCh38
rs565925125 Chr.11: 114582638-114582638 on Build GRCh38
rs762404214 Chr.11: 114582611-114582611 on Build GRCh38
rs1046500009 Chr.11: 114582467-114582467 on Build GRCh38
rs778460503 Chr.11: 114582279-114582279 on Build GRCh38
rs775589425 Chr.11: 114582651-114582651 on Build GRCh38
rs773471773 Chr.11: 114582322-114582322 on Build GRCh38
rs747432647 Chr.11: 114582305-114582305 on Build GRCh38
rs762842355 Chr.11: 114582662-114582662 on Build GRCh38
rs1007079319 Chr.11: 114582274-114582274 on Build GRCh38
rs749884864 Chr.11: 114582588-114582589 on Build GRCh38
rs1050840025 Chr.11: 114582256-114582256 on Build GRCh38
rs761232288 Chr.11: 114582489-114582489 on Build GRCh38
rs770461413 Chr.11: 114582407-114582407 on Build GRCh38
rs759783347 Chr.11: 114582559-114582559 on Build GRCh38
rs770477263 Chr.11: 114582272-114582272 on Build GRCh38
rs372964345 Chr.11: 114582628-114582628 on Build GRCh38
rs773381710 Chr.11: 114582416-114582416 on Build GRCh38
rs748664367 Chr.11: 114582406-114582406 on Build GRCh38
rs775135867 Chr.11: 114582258-114582258 on Build GRCh38
rs778259672 Chr.11: 114582684-114582684 on Build GRCh38
rs1002876793 Chr.11: 114582379-114582379 on Build GRCh38
rs921407024 Chr.11: 114582410-114582410 on Build GRCh38
rs763868602 Chr.11: 114582339-114582339 on Build GRCh38
rs371329608 Chr.11: 114582249-114582249 on Build GRCh38
rs775800914 Chr.11: 114582303-114582303 on Build GRCh38
rs772290293 Chr.11: 114582649-114582649 on Build GRCh38
rs527329057 Chr.11: 114582483-114582483 on Build GRCh38
rs373086041 Chr.11: 114582613-114582613 on Build GRCh38
rs750368161 Chr.11: 114582586-114582586 on Build GRCh38
rs763231355 Chr.11: 114582323-114582323 on Build GRCh38
rs757244762 Chr.11: 114582358-114582358 on Build GRCh38
rs945368020 Chr.11: 114582465-114582465 on Build GRCh38
rs1034598429 Chr.11: 114582582-114582582 on Build GRCh38
rs771129856 Chr.11: 114582547-114582547 on Build GRCh38
rs779508520 Chr.11: 114582485-114582485 on Build GRCh38
rs560475261 Chr.11: 114582253-114582253 on Build GRCh38
rs773053522 Chr.11: 114582658-114582658 on Build GRCh38
rs368421021 Chr.11: 114582484-114582484 on Build GRCh38
rs564705857 Chr.11: 114582436-114582436 on Build GRCh38
rs762300115 Chr.11: 114582271-114582271 on Build GRCh38
rs765104948 Chr.11: 114582578-114582578 on Build GRCh38
rs201300075 Chr.11: 114582637-114582637 on Build GRCh38
rs377136099 Chr.11: 114582545-114582545 on Build GRCh38
rs367730614 Chr.11: 114582382-114582382 on Build GRCh38
rs371571845 Chr.11: 114582431-114582431 on Build GRCh38
rs545121703 Chr.11: 114582435-114582435 on Build GRCh38
rs750681617 Chr.11: 114582245-114582245 on Build GRCh38
rs112731981 Chr.11: 114582504-114582504 on Build GRCh38
rs767129579 Chr.11: 114582669-114582669 on Build GRCh38

View Full Product Details