Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73126175-73126703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEO1
Entrez Gene ID
4756
Gene Name
neogenin 1
Gene Aliases
IGDCC2, NGN, NTN1R2
UniGene
Hs.388613
Gene Chromosome Location
Chr.15: 73051715-73305206 on Build GRCh38
Transcripts
NM_001172623.1 NM_001172624.1 NM_002499.3 XM_005254408.1 XM_011521628.2 XM_011521629.2 XM_011521630.2 XM_011521632.2 XM_011521633.2 XM_017022231.1 XM_017022232.1 XM_017022233.1 XM_017022234.1 XM_017022235.1 XM_017022236.1 XM_017022237.1 XM_017022238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6892660 c.770C>T p.P257L Chr.15: 73126462-73126462 on Build GRCh38
4056712 c.812T>C p.V271A Chr.15: 73126504-73126504 on Build GRCh38
4961493 c.858G>A p.E286E Chr.15: 73126550-73126550 on Build GRCh38
4056711 c.729T>C p.P243P Chr.15: 73126421-73126421 on Build GRCh38
4757409 c.779G>T p.R260I Chr.15: 73126471-73126471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048521112 Chr.15: 73126497-73126497 on Build GRCh38
rs769324767 Chr.15: 73126401-73126401 on Build GRCh38
rs571063879 Chr.15: 73126674-73126674 on Build GRCh38
rs747845315 Chr.15: 73126400-73126401 on Build GRCh38
rs923180392 Chr.15: 73126316-73126316 on Build GRCh38
rs760012395 Chr.15: 73126522-73126522 on Build GRCh38
rs754082961 Chr.15: 73126510-73126510 on Build GRCh38
rs888605772 Chr.15: 73126515-73126515 on Build GRCh38
rs767851153 Chr.15: 73126368-73126368 on Build GRCh38
rs189717185 Chr.15: 73126238-73126238 on Build GRCh38
rs905640712 Chr.15: 73126395-73126395 on Build GRCh38
rs749035439 Chr.15: 73126580-73126580 on Build GRCh38
rs776703615 Chr.15: 73126609-73126609 on Build GRCh38
rs376666423 Chr.15: 73126493-73126493 on Build GRCh38
rs779479382 Chr.15: 73126426-73126426 on Build GRCh38
rs936549959 Chr.15: 73126386-73126386 on Build GRCh38
rs779666296 Chr.15: 73126448-73126448 on Build GRCh38
rs192950211 Chr.15: 73126554-73126554 on Build GRCh38
rs780640636 Chr.15: 73126552-73126552 on Build GRCh38
rs200006363 Chr.15: 73126456-73126456 on Build GRCh38
rs780801893 Chr.15: 73126429-73126429 on Build GRCh38
rs964460613 Chr.15: 73126586-73126586 on Build GRCh38
rs765646925 Chr.15: 73126524-73126524 on Build GRCh38
rs964797136 Chr.15: 73126675-73126675 on Build GRCh38
rs745561049 Chr.15: 73126439-73126439 on Build GRCh38
rs1007100782 Chr.15: 73126518-73126518 on Build GRCh38
rs897438912 Chr.15: 73126240-73126240 on Build GRCh38
rs751062730 Chr.15: 73126427-73126427 on Build GRCh38
rs777505204 Chr.15: 73126401-73126402 on Build GRCh38
rs941731470 Chr.15: 73126291-73126291 on Build GRCh38
rs766609680 Chr.15: 73126405-73126405 on Build GRCh38
rs755667951 Chr.15: 73126570-73126570 on Build GRCh38
rs977911279 Chr.15: 73126337-73126337 on Build GRCh38
rs761236337 Chr.15: 73126281-73126281 on Build GRCh38
rs772315254 Chr.15: 73126472-73126472 on Build GRCh38
rs761088379 Chr.15: 73126391-73126391 on Build GRCh38
rs369836149 Chr.15: 73126543-73126543 on Build GRCh38
rs532293372 Chr.15: 73126246-73126246 on Build GRCh38
rs748055227 Chr.15: 73126620-73126620 on Build GRCh38
rs979424125 Chr.15: 73126601-73126601 on Build GRCh38
rs774499694 Chr.15: 73126470-73126470 on Build GRCh38
rs566139889 Chr.15: 73126311-73126311 on Build GRCh38
rs1027585696 Chr.15: 73126270-73126270 on Build GRCh38
rs547292719 Chr.15: 73126280-73126280 on Build GRCh38
rs768556798 Chr.15: 73126598-73126598 on Build GRCh38
rs539606269 Chr.15: 73126411-73126411 on Build GRCh38
rs755381737 Chr.15: 73126419-73126419 on Build GRCh38
rs781069335 Chr.15: 73126399-73126399 on Build GRCh38
rs964346427 Chr.15: 73126284-73126284 on Build GRCh38
rs766633111 Chr.15: 73126496-73126496 on Build GRCh38
rs756817434 Chr.15: 73126428-73126428 on Build GRCh38
rs62016813 Chr.15: 73126237-73126237 on Build GRCh38
rs933607398 Chr.15: 73126266-73126266 on Build GRCh38
rs1029955546 Chr.15: 73126660-73126660 on Build GRCh38
rs530247893 Chr.15: 73126344-73126344 on Build GRCh38
rs537735808 Chr.15: 73126484-73126484 on Build GRCh38
rs370484291 Chr.15: 73126208-73126208 on Build GRCh38
rs769517564 Chr.15: 73126442-73126442 on Build GRCh38
rs773455045 Chr.15: 73126479-73126479 on Build GRCh38
rs372320512 Chr.15: 73126461-73126461 on Build GRCh38
rs143129579 Chr.15: 73126471-73126471 on Build GRCh38
rs367560647 Chr.15: 73126640-73126640 on Build GRCh38
rs1001360982 Chr.15: 73126400-73126400 on Build GRCh38
rs756731569 Chr.15: 73126548-73126548 on Build GRCh38
rs966615073 Chr.15: 73126321-73126321 on Build GRCh38
rs1010579828 Chr.15: 73126520-73126520 on Build GRCh38
rs750720410 Chr.15: 73126378-73126378 on Build GRCh38
rs779765912 Chr.15: 73126573-73126573 on Build GRCh38

View Full Product Details